Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42685916-42686189 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTNAP1
Entrez Gene ID
8506
Gene Name
contactin associated protein 1
Gene Aliases
CASPR, CNTNAP, NRXN4, P190
UniGene
Hs.408730
Gene Chromosome Location
Chr.17: 42679963-42699993 on Build GRCh38
Transcripts
NM_003632.2 XM_005257748.4 XM_017025238.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6258642 c.747C>A p.H249Q Chr.17: 42685988-42685988 on Build GRCh38
5473892 c.853C>T p.R285C Chr.17: 42686094-42686094 on Build GRCh38
5006898 c.766G>A p.G256R Chr.17: 42686007-42686007 on Build GRCh38
6631878 c.841G>A p.G281S Chr.17: 42686082-42686082 on Build GRCh38
7107770 c.900G>A p.E300E Chr.17: 42686141-42686141 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775401828 Chr.17: 42686039-42686039 on Build GRCh38
rs776632940 Chr.17: 42686023-42686023 on Build GRCh38
rs765634738 Chr.17: 42686096-42686096 on Build GRCh38
rs773250909 Chr.17: 42686055-42686055 on Build GRCh38
rs756856715 Chr.17: 42686144-42686144 on Build GRCh38
rs910448156 Chr.17: 42686129-42686129 on Build GRCh38
rs989250604 Chr.17: 42686063-42686063 on Build GRCh38
rs762112480 Chr.17: 42685944-42685944 on Build GRCh38
rs779237919 Chr.17: 42685991-42685991 on Build GRCh38
rs377261961 Chr.17: 42686074-42686074 on Build GRCh38
rs765055513 Chr.17: 42685938-42685938 on Build GRCh38
rs770286337 Chr.17: 42686017-42686017 on Build GRCh38
rs747850851 Chr.17: 42685994-42685994 on Build GRCh38
rs758709669 Chr.17: 42686117-42686117 on Build GRCh38
rs369854764 Chr.17: 42685941-42685941 on Build GRCh38
rs762165875 Chr.17: 42686040-42686040 on Build GRCh38
rs373203275 Chr.17: 42686037-42686037 on Build GRCh38
rs373476917 Chr.17: 42686025-42686025 on Build GRCh38
rs760814838 Chr.17: 42686064-42686064 on Build GRCh38
rs753095573 Chr.17: 42686107-42686107 on Build GRCh38
rs865820745 Chr.17: 42685974-42685974 on Build GRCh38
rs767754345 Chr.17: 42685947-42685947 on Build GRCh38
rs74967402 Chr.17: 42686136-42686136 on Build GRCh38
rs62001916 Chr.17: 42686006-42686006 on Build GRCh38
rs879206017 Chr.17: 42685945-42685945 on Build GRCh38
rs139692372 Chr.17: 42686130-42686130 on Build GRCh38
rs766366397 Chr.17: 42686087-42686087 on Build GRCh38
rs780625509 Chr.17: 42685976-42685976 on Build GRCh38
rs527466366 Chr.17: 42686092-42686092 on Build GRCh38
rs922624124 Chr.17: 42686081-42686081 on Build GRCh38
rs560060977 Chr.17: 42686086-42686086 on Build GRCh38
rs780551692 Chr.17: 42686154-42686154 on Build GRCh38
rs746547089 Chr.17: 42686016-42686016 on Build GRCh38
rs754233759 Chr.17: 42685980-42685980 on Build GRCh38
rs755403805 Chr.17: 42685990-42685990 on Build GRCh38
rs769767609 Chr.17: 42686035-42686035 on Build GRCh38
rs1051701748 Chr.17: 42686110-42686110 on Build GRCh38
rs756098225 Chr.17: 42685964-42685964 on Build GRCh38
rs1049892466 Chr.17: 42686001-42686001 on Build GRCh38
rs562643035 Chr.17: 42685951-42685951 on Build GRCh38
rs767875480 Chr.17: 42686050-42686050 on Build GRCh38
rs900882401 Chr.17: 42685946-42685946 on Build GRCh38
rs545191038 Chr.17: 42686013-42686013 on Build GRCh38
rs745866278 Chr.17: 42686168-42686168 on Build GRCh38
rs754429609 Chr.17: 42686091-42686091 on Build GRCh38

View Full Product Details