Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 50365356-50365589 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNA2D2
Entrez Gene ID
9254
Gene Name
calcium voltage-gated channel auxiliary subunit alpha2delta 2
Gene Aliases
CACNA2D
UniGene
Hs.476273
Gene Chromosome Location
Chr.3: 50362613-50504244 on Build GRCh38
Transcripts
NM_001005505.2 NM_001174051.2 NM_001291101.1 NM_006030.3 XM_005265581.4 XM_011534243.2 NR_111912.1 NR_111913.1 NR_111914.1

Gene Symbol
CYB561D2
Entrez Gene ID
11068
Gene Name
cytochrome b561 family member D2
Gene Aliases
101F6, TSP10, XXcos-LUCA11.4
UniGene
Hs.149443
Gene Chromosome Location
Chr.3: 50350695-50368197 on Build GRCh38
Transcripts
NM_001005505.2 NM_001174051.2 NM_001291101.1 NM_006030.3 XM_005265581.4 XM_011534243.2 NR_111912.1 NR_111913.1 NR_111914.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4771160 c.2987A>C p.E996A Chr.3: 50365467-50365467 on Build GRCh38
6618466 c.3082T>G p.S1028A Chr.3: 50365393-50365393 on Build GRCh38
731198 c.3095C>T p.S1032F Chr.3: 50365359-50365359 on Build GRCh38
6618467 c.3061T>G p.S1021A Chr.3: 50365393-50365393 on Build GRCh38
6304316 c.3018C>A p.C1006* Chr.3: 50365436-50365436 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145135167 Chr.3: 50365393-50365393 on Build GRCh38
rs778726117 Chr.3: 50365457-50365457 on Build GRCh38
rs587669753 Chr.3: 50365552-50365552 on Build GRCh38
rs201154283 Chr.3: 50365394-50365394 on Build GRCh38
rs202246703 Chr.3: 50365436-50365436 on Build GRCh38
rs767534935 Chr.3: 50365384-50365384 on Build GRCh38
rs758090213 Chr.3: 50365461-50365461 on Build GRCh38
rs774826852 Chr.3: 50365425-50365425 on Build GRCh38
rs755249188 Chr.3: 50365383-50365383 on Build GRCh38
rs587667347 Chr.3: 50365521-50365521 on Build GRCh38
rs200217699 Chr.3: 50365451-50365451 on Build GRCh38
rs773874066 Chr.3: 50365415-50365415 on Build GRCh38
rs775839909 Chr.3: 50365516-50365516 on Build GRCh38
rs774959195 Chr.3: 50365407-50365407 on Build GRCh38
rs777488651 Chr.3: 50365449-50365449 on Build GRCh38
rs771544610 Chr.3: 50365490-50365490 on Build GRCh38
rs146587089 Chr.3: 50365459-50365459 on Build GRCh38
rs1046005359 Chr.3: 50365560-50365560 on Build GRCh38
rs767988859 Chr.3: 50365412-50365412 on Build GRCh38
rs745556824 Chr.3: 50365404-50365404 on Build GRCh38
rs766918496 Chr.3: 50365422-50365422 on Build GRCh38
rs756230551 Chr.3: 50365445-50365445 on Build GRCh38
rs764066785 Chr.3: 50365450-50365450 on Build GRCh38
rs749853324 Chr.3: 50365487-50365487 on Build GRCh38
rs202063356 Chr.3: 50365460-50365460 on Build GRCh38
rs201100743 Chr.3: 50365508-50365508 on Build GRCh38
rs761829248 Chr.3: 50365525-50365525 on Build GRCh38
rs587771455 Chr.3: 50365440-50365440 on Build GRCh38
rs375525172 Chr.3: 50365379-50365379 on Build GRCh38
rs587774877 Chr.3: 50365396-50365396 on Build GRCh38
rs182425252 Chr.3: 50365498-50365498 on Build GRCh38
rs780845562 Chr.3: 50365486-50365486 on Build GRCh38
rs142269127 Chr.3: 50365520-50365520 on Build GRCh38
rs587621769 Chr.3: 50365539-50365539 on Build GRCh38
rs757097781 Chr.3: 50365398-50365398 on Build GRCh38
rs753890492 Chr.3: 50365523-50365523 on Build GRCh38
rs190120215 Chr.3: 50365410-50365410 on Build GRCh38
rs768270735 Chr.3: 50365485-50365485 on Build GRCh38
rs772267031 Chr.3: 50365513-50365513 on Build GRCh38
rs149440809 Chr.3: 50365397-50365397 on Build GRCh38
rs765192901 Chr.3: 50365526-50365526 on Build GRCh38
rs746588781 Chr.3: 50365468-50365468 on Build GRCh38
rs368065286 Chr.3: 50365427-50365427 on Build GRCh38
rs587747300 Chr.3: 50365538-50365538 on Build GRCh38
rs750296867 Chr.3: 50365527-50365527 on Build GRCh38
rs778670979 Chr.3: 50365403-50365403 on Build GRCh38
rs950369121 Chr.3: 50365550-50365550 on Build GRCh38
rs140865285 Chr.3: 50365435-50365435 on Build GRCh38
rs587631845 Chr.3: 50365375-50365375 on Build GRCh38
rs148079551 Chr.3: 50365419-50365419 on Build GRCh38

View Full Product Details