Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 44787317-44787576 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BEST4
Entrez Gene ID
266675
Gene Name
bestrophin 4
Gene Aliases
VMD2L2
UniGene
Hs.302513
Gene Chromosome Location
Chr.1: 44781836-44792828 on Build GRCh38
Transcripts
NM_153274.2 XM_017001020.1 XM_017001021.1 XM_017001022.1 XM_017001023.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1342998 c.212G>A p.R71H Chr.1: 44787407-44787407 on Build GRCh38
5655204 c.180C>T p.Y60Y Chr.1: 44787439-44787439 on Build GRCh38
681205 c.199C>T p.R67W Chr.1: 44787420-44787420 on Build GRCh38
6611367 c.159G>A p.L53L Chr.1: 44787460-44787460 on Build GRCh38
4828547 c.220G>A p.D74N Chr.1: 44787399-44787399 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs16832245 Chr.1: 44787434-44787434 on Build GRCh38
rs369584291 Chr.1: 44787482-44787482 on Build GRCh38
rs1021538486 Chr.1: 44787506-44787506 on Build GRCh38
rs752957577 Chr.1: 44787420-44787420 on Build GRCh38
rs761014440 Chr.1: 44787539-44787539 on Build GRCh38
rs374657428 Chr.1: 44787382-44787382 on Build GRCh38
rs760112648 Chr.1: 44787528-44787528 on Build GRCh38
rs755410207 Chr.1: 44787414-44787414 on Build GRCh38
rs75504010 Chr.1: 44787555-44787555 on Build GRCh38
rs933903611 Chr.1: 44787407-44787407 on Build GRCh38
rs374914946 Chr.1: 44787452-44787452 on Build GRCh38
rs553816047 Chr.1: 44787523-44787523 on Build GRCh38
rs771862311 Chr.1: 44787465-44787465 on Build GRCh38
rs754111733 Chr.1: 44787450-44787450 on Build GRCh38
rs762964399 Chr.1: 44787428-44787428 on Build GRCh38
rs372975497 Chr.1: 44787394-44787394 on Build GRCh38
rs746721992 Chr.1: 44787477-44787477 on Build GRCh38
rs779917577 Chr.1: 44787470-44787470 on Build GRCh38
rs147375981 Chr.1: 44787438-44787438 on Build GRCh38
rs745854022 Chr.1: 44787460-44787460 on Build GRCh38
rs761244505 Chr.1: 44787379-44787379 on Build GRCh38
rs763602126 Chr.1: 44787366-44787366 on Build GRCh38
rs745783966 Chr.1: 44787339-44787339 on Build GRCh38
rs771948591 Chr.1: 44787341-44787341 on Build GRCh38
rs776332966 Chr.1: 44787510-44787510 on Build GRCh38
rs1017669383 Chr.1: 44787439-44787439 on Build GRCh38
rs79680251 Chr.1: 44787454-44787454 on Build GRCh38
rs138830699 Chr.1: 44787429-44787429 on Build GRCh38
rs753113343 Chr.1: 44787530-44787530 on Build GRCh38
rs138395354 Chr.1: 44787419-44787419 on Build GRCh38
rs148576399 Chr.1: 44787554-44787554 on Build GRCh38
rs761440318 Chr.1: 44787517-44787517 on Build GRCh38
rs567017432 Chr.1: 44787350-44787350 on Build GRCh38
rs769791491 Chr.1: 44787350-44787351 on Build GRCh38
rs368542422 Chr.1: 44787519-44787519 on Build GRCh38
rs776130069 Chr.1: 44787375-44787375 on Build GRCh38
rs760404755 Chr.1: 44787360-44787360 on Build GRCh38

View Full Product Details