Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109671444-109671681 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSTM2
Entrez Gene ID
2946
Gene Name
glutathione S-transferase mu 2 (muscle)
Gene Aliases
GST4, GSTM, GSTM2-2, GTHMUS
UniGene
Hs.279837
Gene Chromosome Location
Chr.1: 109668022-109683997 on Build GRCh38
Transcripts
NM_000848.3 NM_001142368.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4550868 c.511G>T p.E171* Chr.1: 109671527-109671527 on Build GRCh38
6057616 c.487G>A p.V163I Chr.1: 109671503-109671503 on Build GRCh38
6676597 c.567G>A p.E189E Chr.1: 109671583-109671583 on Build GRCh38
5469484 c.559C>T p.R187* Chr.1: 109671575-109671575 on Build GRCh38
6028680 c.532T>A p.F178I Chr.1: 109671548-109671548 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372310109 Chr.1: 109671545-109671545 on Build GRCh38
rs775408664 Chr.1: 109671520-109671520 on Build GRCh38
rs200146590 Chr.1: 109671494-109671494 on Build GRCh38
rs756812334 Chr.1: 109671550-109671550 on Build GRCh38
rs565065268 Chr.1: 109671641-109671641 on Build GRCh38
rs764914762 Chr.1: 109671508-109671508 on Build GRCh38
rs141100983 Chr.1: 109671498-109671498 on Build GRCh38
rs753066473 Chr.1: 109671582-109671582 on Build GRCh38
rs745588550 Chr.1: 109671464-109671464 on Build GRCh38
rs758767133 Chr.1: 109671593-109671593 on Build GRCh38
rs201869450 Chr.1: 109671583-109671583 on Build GRCh38
rs767202519 Chr.1: 109671573-109671573 on Build GRCh38
rs955200013 Chr.1: 109671602-109671602 on Build GRCh38
rs776822153 Chr.1: 109671500-109671500 on Build GRCh38
rs559293652 Chr.1: 109671499-109671499 on Build GRCh38
rs747573818 Chr.1: 109671604-109671604 on Build GRCh38
rs956425312 Chr.1: 109671625-109671625 on Build GRCh38
rs778196904 Chr.1: 109671601-109671601 on Build GRCh38
rs772580173 Chr.1: 109671493-109671493 on Build GRCh38
rs755291871 Chr.1: 109671469-109671469 on Build GRCh38
rs574231286 Chr.1: 109671475-109671475 on Build GRCh38
rs755839132 Chr.1: 109671575-109671575 on Build GRCh38
rs776843576 Chr.1: 109671610-109671611 on Build GRCh38
rs770997349 Chr.1: 109671606-109671606 on Build GRCh38
rs141364640 Chr.1: 109671574-109671574 on Build GRCh38
rs200799476 Chr.1: 109671558-109671558 on Build GRCh38
rs199628405 Chr.1: 109671577-109671577 on Build GRCh38
rs138914594 Chr.1: 109671533-109671533 on Build GRCh38
rs896073087 Chr.1: 109671539-109671539 on Build GRCh38
rs904676004 Chr.1: 109671576-109671576 on Build GRCh38
rs781291859 Chr.1: 109671624-109671624 on Build GRCh38
rs759871533 Chr.1: 109671502-109671502 on Build GRCh38
rs2229050 Chr.1: 109671534-109671534 on Build GRCh38
rs376698398 Chr.1: 109671481-109671481 on Build GRCh38
rs746059575 Chr.1: 109671630-109671630 on Build GRCh38
rs867771179 Chr.1: 109671477-109671477 on Build GRCh38
rs200842177 Chr.1: 109671587-109671587 on Build GRCh38
rs770013047 Chr.1: 109671631-109671631 on Build GRCh38
rs113580534 Chr.1: 109671633-109671633 on Build GRCh38
rs267597910 Chr.1: 109671505-109671505 on Build GRCh38
rs762684019 Chr.1: 109671522-109671522 on Build GRCh38

View Full Product Details