Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 46373151-46373404 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCR5
Entrez Gene ID
1234
Gene Name
C-C motif chemokine receptor 5 (gene/pseudogene)
Gene Aliases
CC-CKR-5, CCCKR5, CCR-5, CD195, CKR-5, CKR5, CMKBR5, IDDM22
UniGene
Hs.450802
Gene Chromosome Location
Chr.3: 46370142-46376206 on Build GRCh38
Transcripts
NM_000579.3 NM_001100168.1 NR_125406.1

Gene Symbol
LOC102724297
Entrez Gene ID
102724297
Gene Name
uncharacterized LOC102724297
Gene Aliases
-
UniGene
Hs.622021
Gene Chromosome Location
Chr.3: 46364660-46407059 on Build GRCh38
Transcripts
NM_000579.3 NM_001100168.1 NR_125406.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3068108 c.305A>G p.Q102R Chr.3: 46373207-46373207 on Build GRCh38
6584347 c.304C>G p.Q102E Chr.3: 46373206-46373206 on Build GRCh38
1559661 c.275C>A p.A92D Chr.3: 46373177-46373177 on Build GRCh38
419996 c.249C>T p.V83V Chr.3: 46373151-46373151 on Build GRCh38
319324 c.375T>A p.D125E Chr.3: 46373277-46373277 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371901382 Chr.3: 46373324-46373324 on Build GRCh38
rs201179081 Chr.3: 46373284-46373284 on Build GRCh38
rs1026398876 Chr.3: 46373367-46373367 on Build GRCh38
rs751252593 Chr.3: 46373233-46373233 on Build GRCh38
rs771244514 Chr.3: 46373280-46373280 on Build GRCh38
rs200108729 Chr.3: 46373262-46373262 on Build GRCh38
rs1041177039 Chr.3: 46373346-46373346 on Build GRCh38
rs764747692 Chr.3: 46373210-46373210 on Build GRCh38
rs541069027 Chr.3: 46373290-46373290 on Build GRCh38
rs754799423 Chr.3: 46373325-46373325 on Build GRCh38
rs149975182 Chr.3: 46373185-46373185 on Build GRCh38
rs752265311 Chr.3: 46373245-46373245 on Build GRCh38
rs55639502 Chr.3: 46373379-46373379 on Build GRCh38
rs773818071 Chr.3: 46373277-46373277 on Build GRCh38
rs865910667 Chr.3: 46373246-46373246 on Build GRCh38
rs559783442 Chr.3: 46373292-46373292 on Build GRCh38
rs767205045 Chr.3: 46373289-46373289 on Build GRCh38
rs757918238 Chr.3: 46373175-46373175 on Build GRCh38
rs746300015 Chr.3: 46373378-46373378 on Build GRCh38
rs765063799 Chr.3: 46373313-46373313 on Build GRCh38
rs757294302 Chr.3: 46373307-46373307 on Build GRCh38
rs138380934 Chr.3: 46373274-46373274 on Build GRCh38
rs777744606 Chr.3: 46373359-46373359 on Build GRCh38
rs766082963 Chr.3: 46373176-46373176 on Build GRCh38
rs769009812 Chr.3: 46373272-46373272 on Build GRCh38
rs558483026 Chr.3: 46373229-46373229 on Build GRCh38
rs901409670 Chr.3: 46373174-46373174 on Build GRCh38
rs759216579 Chr.3: 46373238-46373238 on Build GRCh38
rs757171495 Chr.3: 46373261-46373261 on Build GRCh38
rs971864402 Chr.3: 46373227-46373227 on Build GRCh38
rs754530124 Chr.3: 46373182-46373182 on Build GRCh38
rs762167048 Chr.3: 46373224-46373224 on Build GRCh38
rs772422768 Chr.3: 46373204-46373204 on Build GRCh38
rs756257015 Chr.3: 46373332-46373332 on Build GRCh38
rs748699284 Chr.3: 46373249-46373251 on Build GRCh38
rs747196580 Chr.3: 46373270-46373270 on Build GRCh38
rs776790089 Chr.3: 46373212-46373213 on Build GRCh38
rs867670777 Chr.3: 46373241-46373241 on Build GRCh38
rs781530599 Chr.3: 46373330-46373330 on Build GRCh38
rs766967187 Chr.3: 46373239-46373239 on Build GRCh38
rs770335913 Chr.3: 46373225-46373225 on Build GRCh38
rs138174483 Chr.3: 46373221-46373221 on Build GRCh38
rs756101518 Chr.3: 46373188-46373188 on Build GRCh38
rs765993128 Chr.3: 46373231-46373231 on Build GRCh38
rs749113400 Chr.3: 46373196-46373196 on Build GRCh38
rs1800560 Chr.3: 46373205-46373205 on Build GRCh38
rs200815713 Chr.3: 46373240-46373240 on Build GRCh38
rs763433060 Chr.3: 46373230-46373230 on Build GRCh38
rs183662584 Chr.3: 46373218-46373218 on Build GRCh38
rs771443021 Chr.3: 46373364-46373364 on Build GRCh38
rs759132997 Chr.3: 46373286-46373286 on Build GRCh38
rs747065935 Chr.3: 46373208-46373208 on Build GRCh38
rs370409212 Chr.3: 46373260-46373260 on Build GRCh38
rs574865763 Chr.3: 46373361-46373361 on Build GRCh38
rs150497029 Chr.3: 46373264-46373264 on Build GRCh38
rs755938296 Chr.3: 46373254-46373254 on Build GRCh38
rs376364792 Chr.3: 46373179-46373179 on Build GRCh38
rs56068070 Chr.3: 46373211-46373211 on Build GRCh38
rs146209111 Chr.3: 46373275-46373275 on Build GRCh38
rs748154509 Chr.3: 46373331-46373331 on Build GRCh38
rs542780938 Chr.3: 46373370-46373370 on Build GRCh38
rs34418657 Chr.3: 46373293-46373293 on Build GRCh38
rs777374937 Chr.3: 46373190-46373190 on Build GRCh38

View Full Product Details