Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 43981070-43981299 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B4GALT2
Entrez Gene ID
8704
Gene Name
beta-1,4-galactosyltransferase 2
Gene Aliases
B4Gal-T2, B4Gal-T3, beta4Gal-T2
UniGene
Hs.632403
Gene Chromosome Location
Chr.1: 43979202-43991171 on Build GRCh38
Transcripts
NM_001005417.2 NM_003780.4 NM_030587.2 XM_017002716.1 XM_017002717.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5984746 c.126C>T p.F42F Chr.1: 43981286-43981286 on Build GRCh38
3944109 c.46G>T p.G16* Chr.1: 43981119-43981119 on Build GRCh38
4664952 c.131C>T p.A44V Chr.1: 43981204-43981204 on Build GRCh38
6501664 c.218G>A p.R73H Chr.1: 43981291-43981291 on Build GRCh38
4664953 c.44C>T p.A15V Chr.1: 43981204-43981204 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs943486521 Chr.1: 43981189-43981189 on Build GRCh38
rs536172060 Chr.1: 43981192-43981192 on Build GRCh38
rs111233359 Chr.1: 43981132-43981132 on Build GRCh38
rs774767449 Chr.1: 43981152-43981152 on Build GRCh38
rs376991548 Chr.1: 43981256-43981256 on Build GRCh38
rs757948149 Chr.1: 43981139-43981139 on Build GRCh38
rs746116651 Chr.1: 43981171-43981171 on Build GRCh38
rs757515250 Chr.1: 43981242-43981242 on Build GRCh38
rs376096161 Chr.1: 43981119-43981119 on Build GRCh38
rs1021080409 Chr.1: 43981236-43981236 on Build GRCh38
rs767540187 Chr.1: 43981112-43981112 on Build GRCh38
rs779452219 Chr.1: 43981246-43981246 on Build GRCh38
rs139961353 Chr.1: 43981260-43981260 on Build GRCh38
rs762724852 Chr.1: 43981091-43981091 on Build GRCh38
rs775520119 Chr.1: 43981118-43981118 on Build GRCh38
rs746022331 Chr.1: 43981252-43981252 on Build GRCh38
rs759490732 Chr.1: 43981175-43981175 on Build GRCh38
rs748611876 Chr.1: 43981141-43981141 on Build GRCh38
rs1056470275 Chr.1: 43981199-43981199 on Build GRCh38
rs375735099 Chr.1: 43981241-43981241 on Build GRCh38
rs368225760 Chr.1: 43981136-43981136 on Build GRCh38
rs780266757 Chr.1: 43981262-43981262 on Build GRCh38
rs756580465 Chr.1: 43981146-43981146 on Build GRCh38
rs749684547 Chr.1: 43981151-43981151 on Build GRCh38
rs141394710 Chr.1: 43981126-43981126 on Build GRCh38
rs371553773 Chr.1: 43981194-43981194 on Build GRCh38
rs756643777 Chr.1: 43981210-43981210 on Build GRCh38
rs894020408 Chr.1: 43981204-43981204 on Build GRCh38
rs1010989612 Chr.1: 43981221-43981221 on Build GRCh38
rs529773880 Chr.1: 43981158-43981158 on Build GRCh38
rs766166962 Chr.1: 43981102-43981102 on Build GRCh38
rs567524191 Chr.1: 43981183-43981183 on Build GRCh38
rs547278358 Chr.1: 43981176-43981176 on Build GRCh38
rs776480818 Chr.1: 43981276-43981276 on Build GRCh38
rs41303407 Chr.1: 43981265-43981265 on Build GRCh38
rs765806648 Chr.1: 43981193-43981193 on Build GRCh38
rs750026175 Chr.1: 43981184-43981184 on Build GRCh38
rs375329179 Chr.1: 43981140-43981140 on Build GRCh38
rs571755531 Chr.1: 43981107-43981107 on Build GRCh38
rs776479895 Chr.1: 43981180-43981180 on Build GRCh38
rs759733436 Chr.1: 43981105-43981105 on Build GRCh38
rs960237219 Chr.1: 43981266-43981266 on Build GRCh38
rs867144565 Chr.1: 43981125-43981125 on Build GRCh38
rs756246445 Chr.1: 43981268-43981268 on Build GRCh38
rs761552271 Chr.1: 43981181-43981181 on Build GRCh38
rs527442282 Chr.1: 43981175-43981175 on Build GRCh38
rs968474262 Chr.1: 43981157-43981157 on Build GRCh38
rs753587540 Chr.1: 43981127-43981127 on Build GRCh38
rs774216783 Chr.1: 43981174-43981175 on Build GRCh38

View Full Product Details