Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 224973952-224974225 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH14
Entrez Gene ID
127602
Gene Name
dynein axonemal heavy chain 14
Gene Aliases
C1orf67, Dnahc14, HL-18, HL18
UniGene
Hs.133977
Gene Chromosome Location
Chr.1: 224928422-225399294 on Build GRCh38
Transcripts
NM_001145154.1 NM_001373.1 XM_006711735.3 XM_011544057.2 XM_011544058.2 XM_011544059.2 XM_011544061.2 XM_011544062.2 XM_011544063.2 XM_011544064.2 XM_011544065.2 XM_011544066.2 XM_011544067.2 XM_011544069.2 XM_011544070.2 XM_011544071.2 XM_011544072.2 XM_017000295.1 XM_017000296.1 XM_017000297.1 XM_017000298.1 XM_017000300.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5379795 c.791G>A p.W264* Chr.1: 224974114-224974114 on Build GRCh38
5773500 c.830+4A>G p.? Chr.1: 224974157-224974157 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs139426229 Chr.1: 224973989-224973989 on Build GRCh38
rs774780065 Chr.1: 224973997-224973997 on Build GRCh38
rs1011645158 Chr.1: 224974169-224974169 on Build GRCh38
rs374048564 Chr.1: 224974132-224974132 on Build GRCh38
rs760251351 Chr.1: 224974168-224974168 on Build GRCh38
rs907406276 Chr.1: 224974077-224974077 on Build GRCh38
rs192523416 Chr.1: 224974153-224974153 on Build GRCh38
rs926625824 Chr.1: 224974024-224974024 on Build GRCh38
rs982542092 Chr.1: 224973985-224973985 on Build GRCh38
rs755080221 Chr.1: 224974115-224974115 on Build GRCh38
rs187541101 Chr.1: 224974047-224974047 on Build GRCh38
rs772039638 Chr.1: 224974171-224974171 on Build GRCh38
rs749476043 Chr.1: 224974088-224974088 on Build GRCh38
rs779022678 Chr.1: 224974158-224974158 on Build GRCh38
rs1045034033 Chr.1: 224974064-224974064 on Build GRCh38
rs745460033 Chr.1: 224974045-224974046 on Build GRCh38
rs748057987 Chr.1: 224974167-224974167 on Build GRCh38
rs948979993 Chr.1: 224974058-224974058 on Build GRCh38
rs72470459 Chr.1: 224974175-224974175 on Build GRCh38
rs532921958 Chr.1: 224974048-224974048 on Build GRCh38
rs757843550 Chr.1: 224974020-224974020 on Build GRCh38
rs560186049 Chr.1: 224974123-224974123 on Build GRCh38
rs551138984 Chr.1: 224974076-224974076 on Build GRCh38
rs973726062 Chr.1: 224973990-224973990 on Build GRCh38

View Full Product Details