Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219575024-219575271 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INHA
Entrez Gene ID
3623
Gene Name
inhibin alpha subunit
Gene Aliases
-
UniGene
Hs.407506
Gene Chromosome Location
Chr.2: 219572232-219575713 on Build GRCh38
Transcripts
NM_002191.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3045526 c.706C>G p.L236V Chr.2: 219575131-219575131 on Build GRCh38
5861862 c.601C>T p.P201S Chr.2: 219575026-219575026 on Build GRCh38
3045527 c.729C>T p.P243P Chr.2: 219575154-219575154 on Build GRCh38
4429230 c.807_809delCTC p.S270delS Chr.2: 219575232-219575234 on Build GRCh38
3045524 c.694C>T p.R232C Chr.2: 219575119-219575119 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs903261520 Chr.2: 219575133-219575133 on Build GRCh38
rs937278645 Chr.2: 219575217-219575217 on Build GRCh38
rs759530572 Chr.2: 219575247-219575247 on Build GRCh38
rs768319710 Chr.2: 219575114-219575114 on Build GRCh38
rs368930117 Chr.2: 219575083-219575083 on Build GRCh38
rs746289128 Chr.2: 219575175-219575175 on Build GRCh38
rs1023513888 Chr.2: 219575130-219575130 on Build GRCh38
rs200600232 Chr.2: 219575131-219575131 on Build GRCh38
rs772979889 Chr.2: 219575214-219575214 on Build GRCh38
rs761632829 Chr.2: 219575048-219575048 on Build GRCh38
rs762659098 Chr.2: 219575045-219575045 on Build GRCh38
rs773831859 Chr.2: 219575116-219575116 on Build GRCh38
rs760514578 Chr.2: 219575220-219575220 on Build GRCh38
rs139608664 Chr.2: 219575100-219575100 on Build GRCh38
rs147900292 Chr.2: 219575159-219575159 on Build GRCh38
rs140014760 Chr.2: 219575172-219575172 on Build GRCh38
rs928419389 Chr.2: 219575216-219575216 on Build GRCh38
rs12720062 Chr.2: 219575194-219575194 on Build GRCh38
rs757555967 Chr.2: 219575170-219575170 on Build GRCh38
rs750272026 Chr.2: 219575183-219575183 on Build GRCh38
rs766309905 Chr.2: 219575224-219575224 on Build GRCh38
rs142337821 Chr.2: 219575084-219575084 on Build GRCh38
rs1056094778 Chr.2: 219575250-219575250 on Build GRCh38
rs376758084 Chr.2: 219575153-219575153 on Build GRCh38
rs369278709 Chr.2: 219575184-219575184 on Build GRCh38
rs200421240 Chr.2: 219575119-219575119 on Build GRCh38
rs752787964 Chr.2: 219575061-219575061 on Build GRCh38
rs768005523 Chr.2: 219575193-219575193 on Build GRCh38
rs993568545 Chr.2: 219575095-219575095 on Build GRCh38
rs761479412 Chr.2: 219575117-219575117 on Build GRCh38
rs111982983 Chr.2: 219575047-219575047 on Build GRCh38
rs12720061 Chr.2: 219575104-219575104 on Build GRCh38
rs148567617 Chr.2: 219575060-219575060 on Build GRCh38
rs771782116 Chr.2: 219575201-219575201 on Build GRCh38
rs747734566 Chr.2: 219575199-219575199 on Build GRCh38
rs565471809 Chr.2: 219575232-219575232 on Build GRCh38
rs761596421 Chr.2: 219575113-219575113 on Build GRCh38
rs148455844 Chr.2: 219575156-219575156 on Build GRCh38
rs781700503 Chr.2: 219575085-219575085 on Build GRCh38
rs758448367 Chr.2: 219575164-219575164 on Build GRCh38
rs375844945 Chr.2: 219575058-219575058 on Build GRCh38
rs777906518 Chr.2: 219575074-219575074 on Build GRCh38
rs1037869771 Chr.2: 219575063-219575063 on Build GRCh38
rs758635243 Chr.2: 219575062-219575062 on Build GRCh38
rs751673614 Chr.2: 219575165-219575165 on Build GRCh38
rs750315309 Chr.2: 219575051-219575051 on Build GRCh38
rs762416671 Chr.2: 219575109-219575109 on Build GRCh38
rs373504529 Chr.2: 219575192-219575192 on Build GRCh38

View Full Product Details