Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 43592556-43592807 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRGQ
Entrez Gene ID
126298
Gene Name
immunity related GTPase Q
Gene Aliases
FKSG27, IRGQ1
UniGene
Hs.6217
Gene Chromosome Location
Chr.19: 43584367-43596234 on Build GRCh38
Transcripts
NM_001007561.2 XM_005258515.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5856045 c.1315C>T p.L439F Chr.19: 43592583-43592583 on Build GRCh38
6889185 c.1276G>A p.E426K Chr.19: 43592622-43592622 on Build GRCh38
1197279 c.1282C>T p.P428S Chr.19: 43592616-43592616 on Build GRCh38
6687349 c.1233C>T p.A411A Chr.19: 43592665-43592665 on Build GRCh38
3422893 c.1131G>T p.S377S Chr.19: 43592767-43592767 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755832653 Chr.19: 43592776-43592776 on Build GRCh38
rs766377558 Chr.19: 43592734-43592734 on Build GRCh38
rs780856578 Chr.19: 43592659-43592659 on Build GRCh38
rs749810122 Chr.19: 43592614-43592614 on Build GRCh38
rs140503327 Chr.19: 43592725-43592725 on Build GRCh38
rs1019493590 Chr.19: 43592580-43592580 on Build GRCh38
rs200332851 Chr.19: 43592665-43592665 on Build GRCh38
rs267605527 Chr.19: 43592782-43592782 on Build GRCh38
rs765791839 Chr.19: 43592645-43592645 on Build GRCh38
rs773661245 Chr.19: 43592610-43592610 on Build GRCh38
rs765308965 Chr.19: 43592574-43592574 on Build GRCh38
rs748552507 Chr.19: 43592604-43592604 on Build GRCh38
rs376275493 Chr.19: 43592700-43592700 on Build GRCh38
rs754715550 Chr.19: 43592597-43592597 on Build GRCh38
rs150440122 Chr.19: 43592751-43592751 on Build GRCh38
rs1046325224 Chr.19: 43592664-43592664 on Build GRCh38
rs767028534 Chr.19: 43592765-43592765 on Build GRCh38
rs372501578 Chr.19: 43592757-43592757 on Build GRCh38
rs919574068 Chr.19: 43592617-43592617 on Build GRCh38
rs752280093 Chr.19: 43592775-43592775 on Build GRCh38
rs369394427 Chr.19: 43592702-43592702 on Build GRCh38
rs750405377 Chr.19: 43592586-43592586 on Build GRCh38
rs781116385 Chr.19: 43592660-43592660 on Build GRCh38
rs114508854 Chr.19: 43592620-43592620 on Build GRCh38
rs757677370 Chr.19: 43592780-43592780 on Build GRCh38
rs770175442 Chr.19: 43592606-43592606 on Build GRCh38
rs148317528 Chr.19: 43592632-43592632 on Build GRCh38
rs201312534 Chr.19: 43592745-43592745 on Build GRCh38
rs199964549 Chr.19: 43592595-43592595 on Build GRCh38
rs76873184 Chr.19: 43592602-43592602 on Build GRCh38
rs750660114 Chr.19: 43592653-43592659 on Build GRCh38
rs756361598 Chr.19: 43592671-43592672 on Build GRCh38
rs767833187 Chr.19: 43592619-43592619 on Build GRCh38
rs778098867 Chr.19: 43592777-43592777 on Build GRCh38
rs940829130 Chr.19: 43592657-43592657 on Build GRCh38
rs779438724 Chr.19: 43592783-43592783 on Build GRCh38
rs1056089632 Chr.19: 43592722-43592722 on Build GRCh38
rs746634025 Chr.19: 43592596-43592596 on Build GRCh38
rs202164946 Chr.19: 43592639-43592639 on Build GRCh38
rs750315910 Chr.19: 43592638-43592638 on Build GRCh38
rs372736629 Chr.19: 43592615-43592615 on Build GRCh38
rs760978733 Chr.19: 43592711-43592711 on Build GRCh38
rs778117125 Chr.19: 43592667-43592667 on Build GRCh38
rs751128429 Chr.19: 43592651-43592651 on Build GRCh38
rs111946192 Chr.19: 43592703-43592703 on Build GRCh38
rs987042144 Chr.19: 43592575-43592575 on Build GRCh38
rs762775998 Chr.19: 43592732-43592732 on Build GRCh38
rs965499629 Chr.19: 43592670-43592670 on Build GRCh38
rs780413995 Chr.19: 43592721-43592747 on Build GRCh38
rs575906968 Chr.19: 43592779-43592779 on Build GRCh38
rs773197023 Chr.19: 43592729-43592729 on Build GRCh38
rs774560118 Chr.19: 43592618-43592618 on Build GRCh38
rs553066545 Chr.19: 43592587-43592587 on Build GRCh38

View Full Product Details