Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 36284591-36284864 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EEPD1
Entrez Gene ID
80820
Gene Name
endonuclease/exonuclease/phosphatase family domain containing 1
Gene Aliases
HSPC107
UniGene
Hs.487994
Gene Chromosome Location
Chr.7: 36153149-36301543 on Build GRCh38
Transcripts
NM_030636.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1622818 c.1097A>T p.D366V Chr.7: 36284741-36284741 on Build GRCh38
3228778 c.1100C>T p.A367V Chr.7: 36284744-36284744 on Build GRCh38
3228779 c.1119G>A p.S373S Chr.7: 36284763-36284763 on Build GRCh38
1450540 c.1073C>T p.A358V Chr.7: 36284717-36284717 on Build GRCh38
6651824 c.1081G>A p.G361S Chr.7: 36284725-36284725 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764050568 Chr.7: 36284717-36284717 on Build GRCh38
rs946229491 Chr.7: 36284656-36284656 on Build GRCh38
rs752444320 Chr.7: 36284687-36284687 on Build GRCh38
rs771283188 Chr.7: 36284791-36284791 on Build GRCh38
rs556314080 Chr.7: 36284801-36284801 on Build GRCh38
rs978373499 Chr.7: 36284667-36284667 on Build GRCh38
rs748521659 Chr.7: 36284780-36284780 on Build GRCh38
rs748343830 Chr.7: 36284657-36284657 on Build GRCh38
rs751170853 Chr.7: 36284757-36284757 on Build GRCh38
rs762976339 Chr.7: 36284809-36284809 on Build GRCh38
rs144203625 Chr.7: 36284779-36284779 on Build GRCh38
rs1043312235 Chr.7: 36284666-36284666 on Build GRCh38
rs780708131 Chr.7: 36284709-36284709 on Build GRCh38
rs781417606 Chr.7: 36284792-36284792 on Build GRCh38
rs756797390 Chr.7: 36284702-36284702 on Build GRCh38
rs199764578 Chr.7: 36284825-36284825 on Build GRCh38
rs757410837 Chr.7: 36284745-36284745 on Build GRCh38
rs769023084 Chr.7: 36284646-36284646 on Build GRCh38
rs768388217 Chr.7: 36284812-36284812 on Build GRCh38
rs922244268 Chr.7: 36284805-36284805 on Build GRCh38
rs554647485 Chr.7: 36284715-36284715 on Build GRCh38
rs747171982 Chr.7: 36284790-36284790 on Build GRCh38
rs760893877 Chr.7: 36284677-36284677 on Build GRCh38
rs763707452 Chr.7: 36284746-36284746 on Build GRCh38
rs375949207 Chr.7: 36284679-36284679 on Build GRCh38
rs758871973 Chr.7: 36284785-36284785 on Build GRCh38
rs765158676 Chr.7: 36284742-36284742 on Build GRCh38
rs915177114 Chr.7: 36284628-36284628 on Build GRCh38
rs758069836 Chr.7: 36284689-36284689 on Build GRCh38
rs933747868 Chr.7: 36284625-36284625 on Build GRCh38
rs759452624 Chr.7: 36284725-36284725 on Build GRCh38
rs199655154 Chr.7: 36284808-36284808 on Build GRCh38
rs752797063 Chr.7: 36284641-36284641 on Build GRCh38
rs540878995 Chr.7: 36284760-36284760 on Build GRCh38
rs946157357 Chr.7: 36284831-36284831 on Build GRCh38
rs111320816 Chr.7: 36284724-36284724 on Build GRCh38
rs196589 Chr.7: 36284826-36284826 on Build GRCh38
rs773418360 Chr.7: 36284673-36284673 on Build GRCh38
rs766952625 Chr.7: 36284763-36284763 on Build GRCh38
rs751050303 Chr.7: 36284700-36284700 on Build GRCh38
rs760150444 Chr.7: 36284836-36284836 on Build GRCh38
rs747268053 Chr.7: 36284723-36284723 on Build GRCh38
rs149572443 Chr.7: 36284778-36284778 on Build GRCh38
rs3815682 Chr.7: 36284716-36284716 on Build GRCh38
rs576833376 Chr.7: 36284762-36284762 on Build GRCh38
rs201752562 Chr.7: 36284642-36284642 on Build GRCh38
rs778306668 Chr.7: 36284789-36284789 on Build GRCh38
rs749902092 Chr.7: 36284772-36284772 on Build GRCh38
rs200791900 Chr.7: 36284744-36284744 on Build GRCh38
rs774601220 Chr.7: 36284648-36284648 on Build GRCh38
rs371637404 Chr.7: 36284743-36284743 on Build GRCh38
rs376690347 Chr.7: 36284795-36284795 on Build GRCh38
rs774142838 Chr.7: 36284824-36284824 on Build GRCh38
rs201653478 Chr.7: 36284668-36284668 on Build GRCh38
rs542099954 Chr.7: 36284663-36284664 on Build GRCh38
rs373413738 Chr.7: 36284837-36284837 on Build GRCh38
rs13232200 Chr.7: 36284827-36284827 on Build GRCh38
rs773614952 Chr.7: 36284719-36284719 on Build GRCh38
rs765024509 Chr.7: 36284684-36284684 on Build GRCh38
rs368547576 Chr.7: 36284703-36284703 on Build GRCh38
rs746590280 Chr.7: 36284636-36284636 on Build GRCh38
rs772323105 Chr.7: 36284718-36284718 on Build GRCh38
rs372044928 Chr.7: 36284678-36284678 on Build GRCh38
rs763797410 Chr.7: 36284699-36284699 on Build GRCh38

View Full Product Details