Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7191796-7192067 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLG4
Entrez Gene ID
1742
Gene Name
discs large MAGUK scaffold protein 4
Gene Aliases
PSD95, SAP-90, SAP90
UniGene
Hs.463928
Gene Chromosome Location
Chr.17: 7189890-7220050 on Build GRCh38
Transcripts
NM_001128827.2 NM_001321074.1 NM_001321075.1 NM_001321076.1 NM_001321077.1 NM_001365.4 NR_135527.1 XM_005256491.1 XM_011523698.1 XM_011523699.2 XM_011523702.1 XM_017024288.1 XM_017024289.1 XM_017024290.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2976347 c.2022G>A p.S674S Chr.17: 7191976-7191976 on Build GRCh38
1385838 c.1942C>T p.R648C Chr.17: 7191918-7191918 on Build GRCh38
2976339 c.2064C>T p.P688P Chr.17: 7191934-7191934 on Build GRCh38
7364492 c.2032G>T p.V678L Chr.17: 7191966-7191966 on Build GRCh38
5387182 c.1953C>T p.S651S Chr.17: 7191907-7191907 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs314251 Chr.17: 7191879-7191879 on Build GRCh38
rs371808762 Chr.17: 7191950-7191950 on Build GRCh38
rs898455908 Chr.17: 7191826-7191826 on Build GRCh38
rs779906208 Chr.17: 7191931-7191931 on Build GRCh38
rs961013360 Chr.17: 7192006-7192006 on Build GRCh38
rs540087847 Chr.17: 7192028-7192028 on Build GRCh38
rs558578452 Chr.17: 7191836-7191836 on Build GRCh38
rs775912654 Chr.17: 7192024-7192024 on Build GRCh38
rs372407950 Chr.17: 7192023-7192023 on Build GRCh38
rs1007234978 Chr.17: 7191827-7191827 on Build GRCh38
rs747910074 Chr.17: 7192044-7192044 on Build GRCh38
rs767881790 Chr.17: 7191901-7191901 on Build GRCh38
rs773830473 Chr.17: 7191864-7191864 on Build GRCh38
rs754828513 Chr.17: 7192042-7192042 on Build GRCh38
rs1022393996 Chr.17: 7191940-7191940 on Build GRCh38
rs200866471 Chr.17: 7192026-7192026 on Build GRCh38
rs780965435 Chr.17: 7192043-7192043 on Build GRCh38
rs756267089 Chr.17: 7191911-7191911 on Build GRCh38
rs754740263 Chr.17: 7191949-7191949 on Build GRCh38
rs759932966 Chr.17: 7191895-7191895 on Build GRCh38
rs751479756 Chr.17: 7192040-7192040 on Build GRCh38
rs755735560 Chr.17: 7192045-7192045 on Build GRCh38
rs769168643 Chr.17: 7191863-7191863 on Build GRCh38
rs760016926 Chr.17: 7192021-7192021 on Build GRCh38
rs574653579 Chr.17: 7191985-7191985 on Build GRCh38
rs746737548 Chr.17: 7191851-7191851 on Build GRCh38
rs771589738 Chr.17: 7192012-7192012 on Build GRCh38
rs375420373 Chr.17: 7191873-7191873 on Build GRCh38
rs774961455 Chr.17: 7192017-7192017 on Build GRCh38
rs746825626 Chr.17: 7191934-7191934 on Build GRCh38
rs750438640 Chr.17: 7191917-7191917 on Build GRCh38
rs1051284141 Chr.17: 7191984-7191984 on Build GRCh38
rs532298680 Chr.17: 7192038-7192038 on Build GRCh38
rs375777526 Chr.17: 7191976-7191976 on Build GRCh38
rs930831412 Chr.17: 7192027-7192027 on Build GRCh38
rs375158297 Chr.17: 7192037-7192037 on Build GRCh38
rs752934560 Chr.17: 7191908-7191908 on Build GRCh38
rs1025977891 Chr.17: 7191848-7191848 on Build GRCh38
rs764066743 Chr.17: 7191912-7191912 on Build GRCh38
rs766843277 Chr.17: 7191874-7191874 on Build GRCh38
rs560404804 Chr.17: 7192032-7192032 on Build GRCh38
rs769417143 Chr.17: 7191967-7191967 on Build GRCh38
rs768308324 Chr.17: 7191853-7191853 on Build GRCh38
rs758269670 Chr.17: 7191921-7191921 on Build GRCh38
rs761354987 Chr.17: 7191861-7191861 on Build GRCh38
rs770580850 Chr.17: 7191860-7191860 on Build GRCh38
rs747747289 Chr.17: 7191960-7191960 on Build GRCh38

View Full Product Details