Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 153645141-153645366 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHTOP
Entrez Gene ID
26097
Gene Name
chromatin target of PRMT1
Gene Aliases
C1orf77, FL-SRAG, FOP, SRAG, SRAG-3, SRAG-5, pp7704
UniGene
Hs.611057
Gene Chromosome Location
Chr.1: 153633982-153646306 on Build GRCh38
Transcripts
NM_001206612.1 NM_001244664.1 NM_001317077.1 NM_015607.3 XM_017000993.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6903276 c.733delG p.E245fs*>4 Chr.1: 153645255-153645255 on Build GRCh38
4617136 c.622C>T p.R208C Chr.1: 153645144-153645144 on Build GRCh38
1661150 c.676A>C p.K226Q Chr.1: 153645198-153645198 on Build GRCh38
3474470 c.698C>T p.A233V Chr.1: 153645220-153645220 on Build GRCh38
4672409 c.623G>A p.R208H Chr.1: 153645145-153645145 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752122660 Chr.1: 153645170-153645170 on Build GRCh38
rs200566097 Chr.1: 153645252-153645252 on Build GRCh38
rs762639927 Chr.1: 153645310-153645310 on Build GRCh38
rs753050204 Chr.1: 153645278-153645278 on Build GRCh38
rs756441385 Chr.1: 153645279-153645279 on Build GRCh38
rs147051821 Chr.1: 153645277-153645277 on Build GRCh38
rs578097946 Chr.1: 153645185-153645185 on Build GRCh38
rs148846214 Chr.1: 153645260-153645260 on Build GRCh38
rs200014071 Chr.1: 153645284-153645284 on Build GRCh38
rs747907707 Chr.1: 153645303-153645303 on Build GRCh38
rs181391343 Chr.1: 153645299-153645299 on Build GRCh38
rs1007243255 Chr.1: 153645291-153645291 on Build GRCh38
rs146425380 Chr.1: 153645208-153645208 on Build GRCh38
rs200809784 Chr.1: 153645261-153645261 on Build GRCh38
rs770605014 Chr.1: 153645314-153645314 on Build GRCh38
rs765927765 Chr.1: 153645266-153645266 on Build GRCh38
rs11680 Chr.1: 153645297-153645297 on Build GRCh38
rs752034498 Chr.1: 153645271-153645271 on Build GRCh38
rs41313936 Chr.1: 153645273-153645273 on Build GRCh38
rs756314037 Chr.1: 153645197-153645197 on Build GRCh38
rs577747921 Chr.1: 153645308-153645308 on Build GRCh38
rs746992333 Chr.1: 153645248-153645248 on Build GRCh38
rs781003490 Chr.1: 153645301-153645301 on Build GRCh38
rs745975859 Chr.1: 153645298-153645298 on Build GRCh38
rs762442469 Chr.1: 153645262-153645262 on Build GRCh38
rs541090488 Chr.1: 153645300-153645300 on Build GRCh38
rs966726837 Chr.1: 153645264-153645264 on Build GRCh38
rs369983593 Chr.1: 153645179-153645179 on Build GRCh38
rs781652437 Chr.1: 153645180-153645180 on Build GRCh38
rs990675082 Chr.1: 153645312-153645312 on Build GRCh38
rs761533838 Chr.1: 153645255-153645255 on Build GRCh38
rs189791185 Chr.1: 153645242-153645242 on Build GRCh38
rs749314791 Chr.1: 153645224-153645224 on Build GRCh38
rs11555724 Chr.1: 153645236-153645236 on Build GRCh38
rs754171104 Chr.1: 153645282-153645282 on Build GRCh38
rs143160723 Chr.1: 153645254-153645254 on Build GRCh38

View Full Product Details