Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 95872260-95872497 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC38
Entrez Gene ID
120935
Gene Name
coiled-coil domain containing 38
Gene Aliases
-
UniGene
Hs.210377
Gene Chromosome Location
Chr.12: 95867048-95943301 on Build GRCh38
Transcripts
NM_182496.2 XM_006719229.2 XM_011537883.2 XM_011537884.2 XM_011537886.2 XM_011537887.2 XM_011537888.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5613639 c.1345G>A p.E449K Chr.12: 95872394-95872394 on Build GRCh38
7172119 c.1461G>A p.M487I Chr.12: 95872278-95872278 on Build GRCh38
6622128 c.1419C>T p.I473I Chr.12: 95872320-95872320 on Build GRCh38
5645621 c.1305A>G p.K435K Chr.12: 95872434-95872434 on Build GRCh38
1639274 c.1352A>C p.D451A Chr.12: 95872387-95872387 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762142680 Chr.12: 95872346-95872346 on Build GRCh38
rs546781954 Chr.12: 95872437-95872437 on Build GRCh38
rs767561976 Chr.12: 95872467-95872467 on Build GRCh38
rs758073256 Chr.12: 95872414-95872414 on Build GRCh38
rs757051516 Chr.12: 95872404-95872404 on Build GRCh38
rs763099926 Chr.12: 95872444-95872444 on Build GRCh38
rs549394904 Chr.12: 95872377-95872377 on Build GRCh38
rs770090309 Chr.12: 95872438-95872438 on Build GRCh38
rs200076553 Chr.12: 95872385-95872385 on Build GRCh38
rs777326778 Chr.12: 95872424-95872424 on Build GRCh38
rs766519386 Chr.12: 95872447-95872447 on Build GRCh38
rs373282670 Chr.12: 95872358-95872358 on Build GRCh38
rs764081481 Chr.12: 95872389-95872389 on Build GRCh38
rs1045355907 Chr.12: 95872315-95872315 on Build GRCh38
rs774539201 Chr.12: 95872332-95872332 on Build GRCh38
rs375981671 Chr.12: 95872312-95872312 on Build GRCh38
rs147823377 Chr.12: 95872386-95872386 on Build GRCh38
rs759676577 Chr.12: 95872408-95872409 on Build GRCh38
rs777275253 Chr.12: 95872345-95872345 on Build GRCh38
rs774549282 Chr.12: 95872450-95872450 on Build GRCh38
rs779718363 Chr.12: 95872415-95872415 on Build GRCh38
rs567568406 Chr.12: 95872406-95872406 on Build GRCh38
rs747533845 Chr.12: 95872325-95872325 on Build GRCh38
rs763198256 Chr.12: 95872373-95872373 on Build GRCh38
rs144956990 Chr.12: 95872288-95872288 on Build GRCh38
rs767516757 Chr.12: 95872437-95872437 on Build GRCh38
rs771521123 Chr.12: 95872319-95872319 on Build GRCh38
rs369322258 Chr.12: 95872320-95872320 on Build GRCh38
rs753797072 Chr.12: 95872392-95872392 on Build GRCh38
rs765617368 Chr.12: 95872354-95872354 on Build GRCh38
rs759666576 Chr.12: 95872454-95872454 on Build GRCh38
rs773580774 Chr.12: 95872441-95872441 on Build GRCh38
rs1048758318 Chr.12: 95872321-95872321 on Build GRCh38
rs201020728 Chr.12: 95872348-95872348 on Build GRCh38
rs530908291 Chr.12: 95872326-95872326 on Build GRCh38
rs201603971 Chr.12: 95872421-95872421 on Build GRCh38
rs964050536 Chr.12: 95872391-95872391 on Build GRCh38
rs778487658 Chr.12: 95872306-95872306 on Build GRCh38
rs373464624 Chr.12: 95872418-95872418 on Build GRCh38

View Full Product Details