Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 71862436-71862689 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD17B
Entrez Gene ID
51104
Gene Name
abhydrolase domain containing 17B
Gene Aliases
C9orf77, CGI-67, FAM108B1
UniGene
Hs.380389
Gene Chromosome Location
Chr.9: 71859461-71911245 on Build GRCh38
Transcripts
NM_016014.3 XM_006717134.3 XM_017014787.1 XM_017014790.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5467739 c.861G>A p.Q287Q Chr.9: 71862602-71862602 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs756118616 Chr.9: 71862533-71862533 on Build GRCh38
rs967737133 Chr.9: 71862508-71862508 on Build GRCh38
rs984501845 Chr.9: 71862463-71862463 on Build GRCh38
rs148748134 Chr.9: 71862607-71862607 on Build GRCh38
rs770232476 Chr.9: 71862566-71862566 on Build GRCh38
rs765881525 Chr.9: 71862551-71862552 on Build GRCh38
rs765813270 Chr.9: 71862631-71862631 on Build GRCh38
rs757785109 Chr.9: 71862630-71862630 on Build GRCh38
rs777792132 Chr.9: 71862627-71862627 on Build GRCh38
rs891868526 Chr.9: 71862659-71862659 on Build GRCh38
rs773145614 Chr.9: 71862575-71862575 on Build GRCh38
rs372360722 Chr.9: 71862645-71862645 on Build GRCh38
rs554523632 Chr.9: 71862620-71862620 on Build GRCh38
rs754387623 Chr.9: 71862550-71862550 on Build GRCh38
rs961908630 Chr.9: 71862498-71862498 on Build GRCh38
rs200993765 Chr.9: 71862644-71862644 on Build GRCh38
rs144986647 Chr.9: 71862598-71862598 on Build GRCh38
rs746719963 Chr.9: 71862633-71862635 on Build GRCh38
rs764820524 Chr.9: 71862592-71862593 on Build GRCh38
rs1018019333 Chr.9: 71862507-71862507 on Build GRCh38
rs761115944 Chr.9: 71862550-71862551 on Build GRCh38
rs770743854 Chr.9: 71862596-71862596 on Build GRCh38
rs769172410 Chr.9: 71862569-71862569 on Build GRCh38
rs201261766 Chr.9: 71862651-71862651 on Build GRCh38
rs754404407 Chr.9: 71862627-71862627 on Build GRCh38
rs753538865 Chr.9: 71862590-71862593 on Build GRCh38
rs752859664 Chr.9: 71862617-71862617 on Build GRCh38
rs760869817 Chr.9: 71862619-71862619 on Build GRCh38
rs752298641 Chr.9: 71862594-71862596 on Build GRCh38
rs762809626 Chr.9: 71862595-71862595 on Build GRCh38
rs778492903 Chr.9: 71862538-71862538 on Build GRCh38
rs747758610 Chr.9: 71862568-71862568 on Build GRCh38
rs371719974 Chr.9: 71862551-71862551 on Build GRCh38
rs373824532 Chr.9: 71862603-71862603 on Build GRCh38
rs758233008 Chr.9: 71862595-71862596 on Build GRCh38
rs780674266 Chr.9: 71862567-71862567 on Build GRCh38
rs780954493 Chr.9: 71862656-71862656 on Build GRCh38
rs533615278 Chr.9: 71862593-71862595 on Build GRCh38
rs746677517 Chr.9: 71862565-71862565 on Build GRCh38
rs779357539 Chr.9: 71862552-71862552 on Build GRCh38
rs907624551 Chr.9: 71862464-71862464 on Build GRCh38

View Full Product Details