Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 69176477-69176750 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRMD4B
Entrez Gene ID
23150
Gene Name
FERM domain containing 4B
Gene Aliases
6030440G05Rik, GRSP1
UniGene
Hs.371681
Gene Chromosome Location
Chr.3: 69168782-69541340 on Build GRCh38
Transcripts
NM_015123.2 XM_005264722.1 XM_005264723.1 XM_017005989.1 XM_017005990.1 XM_017005991.1 XM_017005992.1 XM_017005993.1 XM_017005994.1 XM_017005995.1 XM_017005996.1 XM_017005997.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1594089 c.2855C>A p.S952Y Chr.3: 69176653-69176653 on Build GRCh38
6454286 c.2790C>T p.G930G Chr.3: 69176556-69176556 on Build GRCh38
261013 c.2692T>C p.S898P Chr.3: 69176654-69176654 on Build GRCh38
5731334 c.2697T>C p.S899S Chr.3: 69176649-69176649 on Build GRCh38
1048307 c.2693C>A p.S898Y Chr.3: 69176653-69176653 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775940196 Chr.3: 69176679-69176680 on Build GRCh38
rs146925019 Chr.3: 69176673-69176673 on Build GRCh38
rs932947522 Chr.3: 69176675-69176675 on Build GRCh38
rs772262957 Chr.3: 69176679-69176682 on Build GRCh38
rs751055608 Chr.3: 69176689-69176689 on Build GRCh38
rs547298120 Chr.3: 69176595-69176595 on Build GRCh38
rs944355230 Chr.3: 69176627-69176627 on Build GRCh38
rs765092365 Chr.3: 69176543-69176543 on Build GRCh38
rs187666580 Chr.3: 69176602-69176602 on Build GRCh38
rs1040177691 Chr.3: 69176662-69176662 on Build GRCh38
rs760756473 Chr.3: 69176697-69176697 on Build GRCh38
rs752568667 Chr.3: 69176594-69176594 on Build GRCh38
rs752748573 Chr.3: 69176545-69176545 on Build GRCh38
rs780848580 Chr.3: 69176570-69176570 on Build GRCh38
rs376569409 Chr.3: 69176601-69176601 on Build GRCh38
rs765182088 Chr.3: 69176613-69176613 on Build GRCh38
rs372860431 Chr.3: 69176587-69176587 on Build GRCh38
rs752480807 Chr.3: 69176654-69176654 on Build GRCh38
rs764084566 Chr.3: 69176687-69176687 on Build GRCh38
rs758394679 Chr.3: 69176548-69176548 on Build GRCh38
rs780397205 Chr.3: 69176590-69176590 on Build GRCh38
rs767736684 Chr.3: 69176514-69176514 on Build GRCh38
rs749853829 Chr.3: 69176592-69176592 on Build GRCh38
rs368493743 Chr.3: 69176553-69176553 on Build GRCh38
rs746463031 Chr.3: 69176554-69176554 on Build GRCh38
rs770187956 Chr.3: 69176586-69176586 on Build GRCh38
rs745486824 Chr.3: 69176571-69176571 on Build GRCh38
rs754848099 Chr.3: 69176516-69176516 on Build GRCh38
rs527456469 Chr.3: 69176717-69176717 on Build GRCh38
rs760803226 Chr.3: 69176611-69176611 on Build GRCh38
rs756892809 Chr.3: 69176568-69176568 on Build GRCh38
rs929500672 Chr.3: 69176718-69176718 on Build GRCh38
rs761942521 Chr.3: 69176509-69176509 on Build GRCh38
rs1038574052 Chr.3: 69176622-69176622 on Build GRCh38
rs73835746 Chr.3: 69176722-69176722 on Build GRCh38
rs764229250 Chr.3: 69176507-69176507 on Build GRCh38
rs763423582 Chr.3: 69176506-69176506 on Build GRCh38
rs9872199 Chr.3: 69176724-69176724 on Build GRCh38
rs368983605 Chr.3: 69176678-69176678 on Build GRCh38
rs753593719 Chr.3: 69176515-69176515 on Build GRCh38
rs773519794 Chr.3: 69176604-69176604 on Build GRCh38
rs900076583 Chr.3: 69176636-69176636 on Build GRCh38
rs998861606 Chr.3: 69176719-69176719 on Build GRCh38

View Full Product Details