Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42460820-42461081 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V0A1
Entrez Gene ID
535
Gene Name
ATPase H+ transporting V0 subunit a1
Gene Aliases
ATP6N1, ATP6N1A, Stv1, VPP1, Vph1, a1
UniGene
Hs.463074
Gene Chromosome Location
Chr.17: 42458844-42522579 on Build GRCh38
Transcripts
NM_001130020.1 NM_001130021.1 NM_005177.3 XM_005257459.1 XM_005257461.1 XM_005257463.2 XM_011524908.1 XM_017024766.1 XM_017024767.1 XM_017024768.1 XM_017024769.1 XM_017024770.1 XM_017024771.1 XM_017024772.1 XM_017024773.1 XM_017024774.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3819628 c.112C>T p.R38C Chr.17: 42461006-42461006 on Build GRCh38
4267109 c.104T>C p.V35A Chr.17: 42460998-42460998 on Build GRCh38
5527145 c.12T>C p.L4L Chr.17: 42460906-42460906 on Build GRCh38
3188771 c.104T>C p.V35A Chr.17: 42460998-42460998 on Build GRCh38
5508644 c.83A>C p.E28A Chr.17: 42460977-42460977 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs980348829 Chr.17: 42460892-42460892 on Build GRCh38
rs770616441 Chr.17: 42460884-42460884 on Build GRCh38
rs750719265 Chr.17: 42460945-42460945 on Build GRCh38
rs142964773 Chr.17: 42460966-42460966 on Build GRCh38
rs779408907 Chr.17: 42461007-42461007 on Build GRCh38
rs144263882 Chr.17: 42460911-42460911 on Build GRCh38
rs796930148 Chr.17: 42460914-42460914 on Build GRCh38
rs1050799200 Chr.17: 42461006-42461006 on Build GRCh38
rs755485576 Chr.17: 42460997-42460997 on Build GRCh38
rs770669796 Chr.17: 42461031-42461031 on Build GRCh38
rs1007025615 Chr.17: 42461023-42461023 on Build GRCh38
rs181018431 Chr.17: 42460936-42460936 on Build GRCh38
rs753863830 Chr.17: 42460973-42460973 on Build GRCh38
rs776252382 Chr.17: 42460887-42460887 on Build GRCh38
rs372616123 Chr.17: 42461011-42461011 on Build GRCh38
rs769626654 Chr.17: 42461053-42461053 on Build GRCh38
rs760389071 Chr.17: 42460865-42460865 on Build GRCh38
rs779205570 Chr.17: 42460903-42460903 on Build GRCh38
rs765218836 Chr.17: 42460894-42460894 on Build GRCh38
rs762746790 Chr.17: 42460928-42460928 on Build GRCh38
rs371426599 Chr.17: 42461028-42461028 on Build GRCh38
rs759577257 Chr.17: 42460889-42460889 on Build GRCh38
rs376972869 Chr.17: 42461027-42461027 on Build GRCh38
rs367652271 Chr.17: 42461032-42461032 on Build GRCh38
rs780134569 Chr.17: 42460972-42460972 on Build GRCh38
rs949905693 Chr.17: 42460867-42460867 on Build GRCh38
rs927192514 Chr.17: 42460893-42460893 on Build GRCh38
rs772983401 Chr.17: 42460852-42460852 on Build GRCh38
rs907977079 Chr.17: 42460988-42460988 on Build GRCh38
rs772545104 Chr.17: 42461012-42461012 on Build GRCh38

View Full Product Details