Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 205526016-205526287 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK18
Entrez Gene ID
5129
Gene Name
cyclin dependent kinase 18
Gene Aliases
PCTAIRE, PCTAIRE3, PCTK3
UniGene
Hs.445402
Gene Chromosome Location
Chr.1: 205504556-205532793 on Build GRCh38
Transcripts
NM_002596.3 NM_212502.2 NM_212503.2 XM_011509602.2 XM_017001423.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3482233 c.566G>A p.R189Q Chr.1: 205526174-205526174 on Build GRCh38
3721211 c.636A>G p.G212G Chr.1: 205526154-205526154 on Build GRCh38
6223853 c.639G>A p.A213A Chr.1: 205526157-205526157 on Build GRCh38
7274618 c.655C>T p.R219* Chr.1: 205526173-205526173 on Build GRCh38
1247935 c.526C>T p.R176W Chr.1: 205526134-205526134 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138968428 Chr.1: 205526157-205526157 on Build GRCh38
rs368932000 Chr.1: 205526035-205526035 on Build GRCh38
rs368933855 Chr.1: 205526186-205526186 on Build GRCh38
rs754614155 Chr.1: 205526067-205526067 on Build GRCh38
rs111912303 Chr.1: 205526040-205526040 on Build GRCh38
rs778575826 Chr.1: 205526069-205526069 on Build GRCh38
rs114729679 Chr.1: 205526044-205526044 on Build GRCh38
rs753307031 Chr.1: 205526156-205526156 on Build GRCh38
rs757166609 Chr.1: 205526205-205526205 on Build GRCh38
rs537796194 Chr.1: 205526045-205526045 on Build GRCh38
rs774600930 Chr.1: 205526135-205526135 on Build GRCh38
rs897950163 Chr.1: 205526172-205526172 on Build GRCh38
rs527729195 Chr.1: 205526256-205526256 on Build GRCh38
rs768810662 Chr.1: 205526134-205526134 on Build GRCh38
rs775483117 Chr.1: 205526211-205526211 on Build GRCh38
rs984746875 Chr.1: 205526262-205526262 on Build GRCh38
rs753319990 Chr.1: 205526054-205526054 on Build GRCh38
rs969704704 Chr.1: 205526214-205526214 on Build GRCh38
rs761266287 Chr.1: 205526146-205526146 on Build GRCh38
rs199539966 Chr.1: 205526208-205526208 on Build GRCh38
rs765816566 Chr.1: 205526052-205526052 on Build GRCh38
rs746768264 Chr.1: 205526201-205526201 on Build GRCh38
rs772350123 Chr.1: 205526229-205526229 on Build GRCh38
rs769852951 Chr.1: 205526033-205526033 on Build GRCh38
rs1030677664 Chr.1: 205526187-205526187 on Build GRCh38
rs2297837 Chr.1: 205526264-205526264 on Build GRCh38
rs138491552 Chr.1: 205526145-205526145 on Build GRCh38
rs747980463 Chr.1: 205526074-205526074 on Build GRCh38
rs960503920 Chr.1: 205526190-205526190 on Build GRCh38
rs748394111 Chr.1: 205526228-205526228 on Build GRCh38
rs757990902 Chr.1: 205526179-205526179 on Build GRCh38
rs774601449 Chr.1: 205526043-205526043 on Build GRCh38
rs763091254 Chr.1: 205526036-205526036 on Build GRCh38
rs368064182 Chr.1: 205526090-205526090 on Build GRCh38
rs200706865 Chr.1: 205526116-205526116 on Build GRCh38
rs374754341 Chr.1: 205526112-205526112 on Build GRCh38
rs897145600 Chr.1: 205526092-205526092 on Build GRCh38
rs74141243 Chr.1: 205526034-205526034 on Build GRCh38
rs764869707 Chr.1: 205526173-205526173 on Build GRCh38
rs764338635 Chr.1: 205526038-205526038 on Build GRCh38
rs542319458 Chr.1: 205526143-205526143 on Build GRCh38
rs17850752 Chr.1: 205526105-205526105 on Build GRCh38
rs571421159 Chr.1: 205526051-205526051 on Build GRCh38
rs906684525 Chr.1: 205526118-205526118 on Build GRCh38
rs200339047 Chr.1: 205526048-205526048 on Build GRCh38
rs746909562 Chr.1: 205526071-205526071 on Build GRCh38
rs752337313 Chr.1: 205526174-205526174 on Build GRCh38
rs768496803 Chr.1: 205526093-205526093 on Build GRCh38
rs573405544 Chr.1: 205526106-205526106 on Build GRCh38
rs766867772 Chr.1: 205526151-205526151 on Build GRCh38
rs977030703 Chr.1: 205526249-205526249 on Build GRCh38
rs533719224 Chr.1: 205526087-205526087 on Build GRCh38
rs777748186 Chr.1: 205526093-205526093 on Build GRCh38
rs749368001 Chr.1: 205526209-205526209 on Build GRCh38
rs367897237 Chr.1: 205526227-205526227 on Build GRCh38

View Full Product Details