Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 73286418-73286643 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC42EP4
Entrez Gene ID
23580
Gene Name
CDC42 effector protein 4
Gene Aliases
BORG4, CEP4, KAIA1777
UniGene
Hs.3903
Gene Chromosome Location
Chr.17: 73283624-73312004 on Build GRCh38
Transcripts
NM_012121.4 XM_005257182.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6523843 c.69G>T p.T23T Chr.17: 73286432-73286432 on Build GRCh38
983837 c.55C>T p.R19* Chr.17: 73286446-73286446 on Build GRCh38
3787418 c.32C>T p.S11L Chr.17: 73286469-73286469 on Build GRCh38
5368307 c.55C>G p.R19G Chr.17: 73286446-73286446 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs757814799 Chr.17: 73286517-73286517 on Build GRCh38
rs762458530 Chr.17: 73286561-73286561 on Build GRCh38
rs140562239 Chr.17: 73286442-73286442 on Build GRCh38
rs539330908 Chr.17: 73286600-73286600 on Build GRCh38
rs761115198 Chr.17: 73286508-73286508 on Build GRCh38
rs917119113 Chr.17: 73286518-73286518 on Build GRCh38
rs17850665 Chr.17: 73286496-73286496 on Build GRCh38
rs753026305 Chr.17: 73286488-73286488 on Build GRCh38
rs770864839 Chr.17: 73286457-73286457 on Build GRCh38
rs779328295 Chr.17: 73286520-73286520 on Build GRCh38
rs761821374 Chr.17: 73286455-73286455 on Build GRCh38
rs11551235 Chr.17: 73286541-73286541 on Build GRCh38
rs776367705 Chr.17: 73286454-73286454 on Build GRCh38
rs569802841 Chr.17: 73286444-73286444 on Build GRCh38
rs767826005 Chr.17: 73286479-73286479 on Build GRCh38
rs768275306 Chr.17: 73286543-73286543 on Build GRCh38
rs935910589 Chr.17: 73286546-73286546 on Build GRCh38
rs1035198832 Chr.17: 73286525-73286525 on Build GRCh38
rs776640579 Chr.17: 73286572-73286572 on Build GRCh38
rs868508363 Chr.17: 73286446-73286446 on Build GRCh38
rs775038770 Chr.17: 73286549-73286549 on Build GRCh38
rs1033944853 Chr.17: 73286480-73286480 on Build GRCh38
rs958476998 Chr.17: 73286497-73286497 on Build GRCh38
rs768066533 Chr.17: 73286575-73286575 on Build GRCh38
rs921304998 Chr.17: 73286592-73286592 on Build GRCh38
rs376758429 Chr.17: 73286468-73286468 on Build GRCh38
rs371743473 Chr.17: 73286451-73286451 on Build GRCh38
rs746797027 Chr.17: 73286452-73286452 on Build GRCh38
rs754252387 Chr.17: 73286515-73286515 on Build GRCh38
rs11867607 Chr.17: 73286590-73286590 on Build GRCh38
rs1040481992 Chr.17: 73286609-73286609 on Build GRCh38
rs757964440 Chr.17: 73286533-73286533 on Build GRCh38
rs147217940 Chr.17: 73286441-73286441 on Build GRCh38
rs1051017823 Chr.17: 73286598-73286600 on Build GRCh38
rs771682160 Chr.17: 73286445-73286445 on Build GRCh38
rs368134711 Chr.17: 73286514-73286514 on Build GRCh38
rs371964654 Chr.17: 73286542-73286542 on Build GRCh38
rs199503438 Chr.17: 73286532-73286532 on Build GRCh38
rs79452902 Chr.17: 73286593-73286593 on Build GRCh38
rs774517149 Chr.17: 73286463-73286463 on Build GRCh38

View Full Product Details