Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 106239712-106239971 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CKAP4
Entrez Gene ID
10970
Gene Name
cytoskeleton associated protein 4
Gene Aliases
CLIMP-63, ERGIC-63, p63
UniGene
Hs.74368
Gene Chromosome Location
Chr.12: 106237881-106247935 on Build GRCh38
Transcripts
NM_006825.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4146504 c.1042G>C p.A348P Chr.12: 106239791-106239791 on Build GRCh38
5812336 c.925G>C p.A309P Chr.12: 106239908-106239908 on Build GRCh38
5653706 c.867A>T p.L289F Chr.12: 106239966-106239966 on Build GRCh38
260117 c.979G>A p.E327K Chr.12: 106239854-106239854 on Build GRCh38
691452 c.1043C>T p.A348V Chr.12: 106239790-106239790 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141394460 Chr.12: 106239739-106239739 on Build GRCh38
rs138167410 Chr.12: 106239807-106239807 on Build GRCh38
rs764573182 Chr.12: 106239806-106239806 on Build GRCh38
rs912285888 Chr.12: 106239894-106239894 on Build GRCh38
rs200691946 Chr.12: 106239743-106239743 on Build GRCh38
rs903178815 Chr.12: 106239790-106239790 on Build GRCh38
rs146992733 Chr.12: 106239879-106239879 on Build GRCh38
rs550924456 Chr.12: 106239801-106239801 on Build GRCh38
rs143926692 Chr.12: 106239740-106239740 on Build GRCh38
rs543189103 Chr.12: 106239823-106239823 on Build GRCh38
rs141311887 Chr.12: 106239864-106239864 on Build GRCh38
rs371832357 Chr.12: 106239759-106239759 on Build GRCh38
rs773194117 Chr.12: 106239855-106239855 on Build GRCh38
rs749961461 Chr.12: 106239813-106239813 on Build GRCh38
rs763063503 Chr.12: 106239747-106239747 on Build GRCh38
rs764344163 Chr.12: 106239872-106239872 on Build GRCh38
rs201305583 Chr.12: 106239763-106239763 on Build GRCh38
rs763436975 Chr.12: 106239914-106239914 on Build GRCh38
rs764077586 Chr.12: 106239744-106239744 on Build GRCh38
rs768253197 Chr.12: 106239831-106239831 on Build GRCh38
rs757922884 Chr.12: 106239819-106239819 on Build GRCh38
rs150919623 Chr.12: 106239779-106239779 on Build GRCh38
rs746848319 Chr.12: 106239787-106239787 on Build GRCh38
rs896971593 Chr.12: 106239818-106239818 on Build GRCh38
rs775153247 Chr.12: 106239742-106239742 on Build GRCh38
rs374042901 Chr.12: 106239797-106239797 on Build GRCh38
rs12818106 Chr.12: 106239939-106239939 on Build GRCh38
rs558578842 Chr.12: 106239737-106239737 on Build GRCh38
rs778463864 Chr.12: 106239774-106239774 on Build GRCh38
rs770325130 Chr.12: 106239867-106239867 on Build GRCh38
rs965086697 Chr.12: 106239905-106239905 on Build GRCh38
rs756588500 Chr.12: 106239734-106239734 on Build GRCh38
rs370136151 Chr.12: 106239757-106239757 on Build GRCh38
rs1033196684 Chr.12: 106239925-106239925 on Build GRCh38
rs200244385 Chr.12: 106239775-106239775 on Build GRCh38
rs200500476 Chr.12: 106239857-106239857 on Build GRCh38
rs3088113 Chr.12: 106239791-106239791 on Build GRCh38
rs751034013 Chr.12: 106239907-106239907 on Build GRCh38
rs369881444 Chr.12: 106239796-106239796 on Build GRCh38
rs756540458 Chr.12: 106239769-106239769 on Build GRCh38
rs150112073 Chr.12: 106239792-106239792 on Build GRCh38
rs77195855 Chr.12: 106239885-106239885 on Build GRCh38
rs553591002 Chr.12: 106239811-106239811 on Build GRCh38
rs142777548 Chr.12: 106239873-106239873 on Build GRCh38
rs757898039 Chr.12: 106239893-106239893 on Build GRCh38
rs776548460 Chr.12: 106239746-106239746 on Build GRCh38
rs766428918 Chr.12: 106239750-106239750 on Build GRCh38
rs146628636 Chr.12: 106239738-106239738 on Build GRCh38
rs201369395 Chr.12: 106239856-106239856 on Build GRCh38
rs765732692 Chr.12: 106239884-106239884 on Build GRCh38
rs899777535 Chr.12: 106239776-106239776 on Build GRCh38
rs757842402 Chr.12: 106239810-106239810 on Build GRCh38
rs748870625 Chr.12: 106239735-106239735 on Build GRCh38
rs769777070 Chr.12: 106239854-106239854 on Build GRCh38
rs369694343 Chr.12: 106239828-106239828 on Build GRCh38
rs775380868 Chr.12: 106239863-106239863 on Build GRCh38
rs771637414 Chr.12: 106239780-106239780 on Build GRCh38
rs779745599 Chr.12: 106239784-106239784 on Build GRCh38
rs781435735 Chr.12: 106239733-106239733 on Build GRCh38
rs540489869 Chr.12: 106239767-106239767 on Build GRCh38
rs187707537 Chr.12: 106239732-106239732 on Build GRCh38
rs567820155 Chr.12: 106239802-106239802 on Build GRCh38
rs934379141 Chr.12: 106239745-106239745 on Build GRCh38
rs765938952 Chr.12: 106239897-106239897 on Build GRCh38
rs781000102 Chr.12: 106239846-106239846 on Build GRCh38
rs747877489 Chr.12: 106239849-106239849 on Build GRCh38

View Full Product Details