Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 95586559-95586808 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARSK
Entrez Gene ID
153642
Gene Name
arylsulfatase family member K
Gene Aliases
TSULF
UniGene
Hs.585051
Gene Chromosome Location
Chr.5: 95555108-95605102 on Build GRCh38
Transcripts
NM_198150.2 XM_005271904.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
318788 c.757G>T p.V253L Chr.5: 95586619-95586619 on Build GRCh38
5587214 c.788G>T p.C263F Chr.5: 95586650-95586650 on Build GRCh38
5989115 c.839A>G p.Y280C Chr.5: 95586701-95586701 on Build GRCh38
1439259 c.780delA p.N262fs*39 Chr.5: 95586642-95586642 on Build GRCh38
5737297 c.804_805insA p.E271fs*3 Chr.5: 95586666-95586667 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs183499702 Chr.5: 95586658-95586658 on Build GRCh38
rs970995308 Chr.5: 95586726-95586726 on Build GRCh38
rs764719102 Chr.5: 95586727-95586727 on Build GRCh38
rs376096151 Chr.5: 95586633-95586633 on Build GRCh38
rs750316907 Chr.5: 95586591-95586591 on Build GRCh38
rs765274466 Chr.5: 95586613-95586613 on Build GRCh38
rs1042910107 Chr.5: 95586614-95586614 on Build GRCh38
rs764053353 Chr.5: 95586727-95586728 on Build GRCh38
rs763673609 Chr.5: 95586598-95586598 on Build GRCh38
rs768380609 Chr.5: 95586703-95586703 on Build GRCh38
rs141927922 Chr.5: 95586675-95586675 on Build GRCh38
rs765323769 Chr.5: 95586765-95586765 on Build GRCh38
rs201849642 Chr.5: 95586667-95586667 on Build GRCh38
rs369059088 Chr.5: 95586665-95586665 on Build GRCh38
rs755155962 Chr.5: 95586599-95586599 on Build GRCh38
rs779685757 Chr.5: 95586683-95586683 on Build GRCh38
rs200586202 Chr.5: 95586701-95586701 on Build GRCh38
rs370101671 Chr.5: 95586662-95586662 on Build GRCh38
rs764241401 Chr.5: 95586782-95586782 on Build GRCh38
rs760507991 Chr.5: 95586740-95586740 on Build GRCh38
rs761729151 Chr.5: 95586725-95586725 on Build GRCh38
rs947116810 Chr.5: 95586594-95586594 on Build GRCh38
rs1012922249 Chr.5: 95586691-95586691 on Build GRCh38
rs758567887 Chr.5: 95586636-95586636 on Build GRCh38
rs756760374 Chr.5: 95586693-95586693 on Build GRCh38
rs1024385168 Chr.5: 95586700-95586700 on Build GRCh38
rs200281345 Chr.5: 95586710-95586710 on Build GRCh38
rs998682024 Chr.5: 95586640-95586640 on Build GRCh38
rs373982219 Chr.5: 95586630-95586630 on Build GRCh38
rs769382487 Chr.5: 95586678-95586678 on Build GRCh38
rs199789005 Chr.5: 95586737-95586737 on Build GRCh38
rs55803959 Chr.5: 95586622-95586622 on Build GRCh38
rs774303564 Chr.5: 95586706-95586706 on Build GRCh38
rs146910298 Chr.5: 95586730-95586730 on Build GRCh38
rs752735612 Chr.5: 95586776-95586776 on Build GRCh38
rs188952316 Chr.5: 95586780-95586780 on Build GRCh38

View Full Product Details