Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 50312380-50312645 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND6B
Entrez Gene ID
414918
Gene Name
DENN domain containing 6B
Gene Aliases
AFI1B, FAM116B
UniGene
Hs.530380
Gene Chromosome Location
Chr.22: 50309032-50327088 on Build GRCh38
Transcripts
NM_001001794.3 XM_005261914.3 XM_011530692.2 XM_011530693.2 XM_017028801.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4002225 c.1572C>T p.T524T Chr.22: 50312406-50312406 on Build GRCh38
3555711 c.1566C>T p.I522I Chr.22: 50312412-50312412 on Build GRCh38
4682406 c.1563C>T p.N521N Chr.22: 50312415-50312415 on Build GRCh38
3555710 c.1568A>G p.E523G Chr.22: 50312410-50312410 on Build GRCh38
6642531 c.1590C>T p.S530S Chr.22: 50312388-50312388 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs777238809 Chr.22: 50312562-50312562 on Build GRCh38
rs764551395 Chr.22: 50312464-50312464 on Build GRCh38
rs766554763 Chr.22: 50312581-50312581 on Build GRCh38
rs772241822 Chr.22: 50312411-50312411 on Build GRCh38
rs1029805720 Chr.22: 50312560-50312560 on Build GRCh38
rs749252524 Chr.22: 50312524-50312524 on Build GRCh38
rs759191928 Chr.22: 50312448-50312448 on Build GRCh38
rs746060215 Chr.22: 50312432-50312432 on Build GRCh38
rs531277848 Chr.22: 50312550-50312550 on Build GRCh38
rs769684031 Chr.22: 50312423-50312423 on Build GRCh38
rs773951541 Chr.22: 50312447-50312447 on Build GRCh38
rs770731538 Chr.22: 50312435-50312435 on Build GRCh38
rs373383489 Chr.22: 50312530-50312530 on Build GRCh38
rs975985417 Chr.22: 50312431-50312431 on Build GRCh38
rs765731573 Chr.22: 50312479-50312479 on Build GRCh38
rs758712234 Chr.22: 50312503-50312503 on Build GRCh38
rs780172065 Chr.22: 50312505-50312505 on Build GRCh38
rs73439304 Chr.22: 50312450-50312450 on Build GRCh38
rs758612608 Chr.22: 50312580-50312580 on Build GRCh38
rs917173735 Chr.22: 50312506-50312506 on Build GRCh38
rs146358653 Chr.22: 50312621-50312622 on Build GRCh38
rs745505770 Chr.22: 50312544-50312544 on Build GRCh38
rs371327605 Chr.22: 50312552-50312552 on Build GRCh38
rs549475217 Chr.22: 50312623-50312623 on Build GRCh38
rs754232609 Chr.22: 50312473-50312473 on Build GRCh38
rs564493613 Chr.22: 50312624-50312624 on Build GRCh38
rs377542127 Chr.22: 50312549-50312549 on Build GRCh38
rs1004434370 Chr.22: 50312568-50312568 on Build GRCh38
rs756052169 Chr.22: 50312516-50312516 on Build GRCh38
rs773026431 Chr.22: 50312425-50312425 on Build GRCh38
rs774143211 Chr.22: 50312532-50312532 on Build GRCh38
rs375517464 Chr.22: 50312567-50312567 on Build GRCh38
rs771688525 Chr.22: 50312546-50312546 on Build GRCh38
rs754969072 Chr.22: 50312587-50312587 on Build GRCh38
rs955621416 Chr.22: 50312427-50312427 on Build GRCh38
rs750879765 Chr.22: 50312574-50312574 on Build GRCh38
rs991431288 Chr.22: 50312497-50312497 on Build GRCh38
rs202213925 Chr.22: 50312403-50312403 on Build GRCh38
rs937137759 Chr.22: 50312482-50312482 on Build GRCh38
rs897322208 Chr.22: 50312566-50312566 on Build GRCh38
rs775034393 Chr.22: 50312547-50312547 on Build GRCh38
rs750609768 Chr.22: 50312484-50312484 on Build GRCh38
rs781416715 Chr.22: 50312415-50312416 on Build GRCh38
rs751826381 Chr.22: 50312582-50312582 on Build GRCh38
rs781252238 Chr.22: 50312588-50312588 on Build GRCh38
rs752205349 Chr.22: 50312456-50312456 on Build GRCh38
rs777519916 Chr.22: 50312523-50312523 on Build GRCh38
rs117291362 Chr.22: 50312406-50312406 on Build GRCh38
rs762781086 Chr.22: 50312432-50312432 on Build GRCh38
rs1024580623 Chr.22: 50312579-50312579 on Build GRCh38
rs765641629 Chr.22: 50312569-50312569 on Build GRCh38
rs747095698 Chr.22: 50312507-50312507 on Build GRCh38
rs561164471 Chr.22: 50312478-50312478 on Build GRCh38
rs868219317 Chr.22: 50312551-50312551 on Build GRCh38
rs780005592 Chr.22: 50312412-50312412 on Build GRCh38

View Full Product Details