Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 51361136-51361409 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPS15
Entrez Gene ID
2060
Gene Name
epidermal growth factor receptor pathway substrate 15
Gene Aliases
AF-1P, AF1P, MLLT5
UniGene
Hs.83722
Gene Chromosome Location
Chr.1: 51354263-51519354 on Build GRCh38
Transcripts
NM_001159969.1 NM_001981.2 XM_005270618.2 XM_017000615.1 XM_017000616.1 XM_017000617.1 XM_017000618.1 XM_017000619.1 XM_017000620.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6848526 c.2499_2500insT p.T834fs*3 Chr.1: 51361215-51361216 on Build GRCh38
4808183 c.1569C>T p.F523F Chr.1: 51361177-51361177 on Build GRCh38
146508 c.2534A>G p.N845S Chr.1: 51361181-51361181 on Build GRCh38
6848527 c.1530_1531insT p.T511fs*3 Chr.1: 51361215-51361216 on Build GRCh38
5728633 c.1400C>T p.S467F Chr.1: 51361346-51361346 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760930252 Chr.1: 51361162-51361162 on Build GRCh38
rs745572941 Chr.1: 51361374-51361374 on Build GRCh38
rs552579354 Chr.1: 51361211-51361211 on Build GRCh38
rs778896057 Chr.1: 51361257-51361257 on Build GRCh38
rs1046245249 Chr.1: 51361175-51361175 on Build GRCh38
rs766972459 Chr.1: 51361167-51361167 on Build GRCh38
rs17567 Chr.1: 51361249-51361249 on Build GRCh38
rs901601410 Chr.1: 51361283-51361283 on Build GRCh38
rs755835490 Chr.1: 51361236-51361236 on Build GRCh38
rs1046971218 Chr.1: 51361197-51361197 on Build GRCh38
rs148821171 Chr.1: 51361184-51361184 on Build GRCh38
rs373517965 Chr.1: 51361274-51361274 on Build GRCh38
rs139788081 Chr.1: 51361312-51361312 on Build GRCh38
rs767319254 Chr.1: 51361313-51361313 on Build GRCh38
rs778804128 Chr.1: 51361373-51361373 on Build GRCh38
rs754848683 Chr.1: 51361351-51361351 on Build GRCh38
rs763660782 Chr.1: 51361190-51361190 on Build GRCh38
rs115367873 Chr.1: 51361310-51361310 on Build GRCh38
rs151081025 Chr.1: 51361181-51361181 on Build GRCh38
rs377729995 Chr.1: 51361282-51361282 on Build GRCh38
rs535318118 Chr.1: 51361293-51361293 on Build GRCh38
rs538305506 Chr.1: 51361219-51361219 on Build GRCh38
rs200593303 Chr.1: 51361382-51361382 on Build GRCh38
rs955088384 Chr.1: 51361355-51361355 on Build GRCh38
rs750299817 Chr.1: 51361231-51361231 on Build GRCh38
rs570863189 Chr.1: 51361214-51361214 on Build GRCh38
rs770865304 Chr.1: 51361207-51361208 on Build GRCh38
rs771175589 Chr.1: 51361279-51361279 on Build GRCh38
rs777963379 Chr.1: 51361278-51361278 on Build GRCh38
rs977130252 Chr.1: 51361372-51361372 on Build GRCh38
rs190799583 Chr.1: 51361255-51361255 on Build GRCh38
rs750022924 Chr.1: 51361320-51361320 on Build GRCh38
rs753815525 Chr.1: 51361337-51361337 on Build GRCh38
rs760543648 Chr.1: 51361328-51361328 on Build GRCh38
rs1005501345 Chr.1: 51361260-51361260 on Build GRCh38
rs145914495 Chr.1: 51361273-51361273 on Build GRCh38
rs749132893 Chr.1: 51361254-51361254 on Build GRCh38

View Full Product Details