Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 196240384-196240657 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HECW2
Entrez Gene ID
57520
Gene Name
HECT, C2 and WW domain containing E3 ubiquitin protein ligase 2
Gene Aliases
NEDL2
UniGene
Hs.654742
Gene Chromosome Location
Chr.2: 196194382-196593692 on Build GRCh38
Transcripts
NM_001304840.1 NM_020760.2 XM_006712646.3 XM_006712648.3 XM_017004563.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1404214 c.3728G>T p.R1243I Chr.2: 196240485-196240485 on Build GRCh38
281664 c.3662G>A p.R1221Q Chr.2: 196240551-196240551 on Build GRCh38
3771231 c.3731A>G p.N1244S Chr.2: 196240482-196240482 on Build GRCh38
442051 c.3726G>C p.Q1242H Chr.2: 196240487-196240487 on Build GRCh38
1014378 c.3750C>T p.F1250F Chr.2: 196240463-196240463 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370667360 Chr.2: 196240512-196240512 on Build GRCh38
rs574725402 Chr.2: 196240592-196240592 on Build GRCh38
rs139510372 Chr.2: 196240510-196240510 on Build GRCh38
rs1000366695 Chr.2: 196240602-196240602 on Build GRCh38
rs769718927 Chr.2: 196240551-196240551 on Build GRCh38
rs554758208 Chr.2: 196240547-196240547 on Build GRCh38
rs759499873 Chr.2: 196240420-196240420 on Build GRCh38
rs763021044 Chr.2: 196240565-196240565 on Build GRCh38
rs760965096 Chr.2: 196240483-196240483 on Build GRCh38
rs576853190 Chr.2: 196240610-196240610 on Build GRCh38
rs142674505 Chr.2: 196240509-196240509 on Build GRCh38
rs776539916 Chr.2: 196240543-196240543 on Build GRCh38
rs775442248 Chr.2: 196240552-196240552 on Build GRCh38
rs770449771 Chr.2: 196240419-196240420 on Build GRCh38
rs368146245 Chr.2: 196240593-196240593 on Build GRCh38
rs201600229 Chr.2: 196240437-196240437 on Build GRCh38
rs745820704 Chr.2: 196240546-196240546 on Build GRCh38
rs757684316 Chr.2: 196240471-196240471 on Build GRCh38
rs746650525 Chr.2: 196240532-196240532 on Build GRCh38
rs777637521 Chr.2: 196240520-196240520 on Build GRCh38
rs751923902 Chr.2: 196240463-196240463 on Build GRCh38
rs1037498952 Chr.2: 196240527-196240527 on Build GRCh38
rs906908096 Chr.2: 196240529-196240529 on Build GRCh38
rs764153467 Chr.2: 196240570-196240570 on Build GRCh38
rs371020934 Chr.2: 196240574-196240574 on Build GRCh38
rs141522728 Chr.2: 196240406-196240407 on Build GRCh38
rs369177409 Chr.2: 196240418-196240418 on Build GRCh38
rs766845247 Chr.2: 196240598-196240598 on Build GRCh38
rs767854683 Chr.2: 196240472-196240472 on Build GRCh38
rs35704479 Chr.2: 196240619-196240620 on Build GRCh38
rs778497175 Chr.2: 196240611-196240616 on Build GRCh38
rs373067310 Chr.2: 196240432-196240432 on Build GRCh38
rs377412926 Chr.2: 196240462-196240462 on Build GRCh38
rs374428183 Chr.2: 196240461-196240461 on Build GRCh38
rs767899851 Chr.2: 196240578-196240578 on Build GRCh38
rs764241293 Chr.2: 196240443-196240443 on Build GRCh38
rs555159490 Chr.2: 196240535-196240535 on Build GRCh38
rs201072645 Chr.2: 196240599-196240599 on Build GRCh38
rs771542620 Chr.2: 196240542-196240542 on Build GRCh38
rs370052088 Chr.2: 196240438-196240438 on Build GRCh38
rs144631612 Chr.2: 196240475-196240475 on Build GRCh38

View Full Product Details