Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 53905950-53906185 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIO1
Entrez Gene ID
1733
Gene Name
deiodinase, iodothyronine type I
Gene Aliases
5DI, TXDI1
UniGene
Hs.251415
Gene Chromosome Location
Chr.1: 53894187-53911086 on Build GRCh38
Transcripts
NM_000792.6 NM_001039715.2 NM_001039716.2 NM_001324316.1 NM_213593.4 NR_136692.1 NR_136693.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
358119 c.570G>A p.R190R Chr.1: 53906183-53906183 on Build GRCh38
4895384 c.403G>A p.A135T Chr.1: 53906160-53906160 on Build GRCh38
4895383 c.547G>A p.A183T Chr.1: 53906160-53906160 on Build GRCh38
1284498 c.412C>A p.L138I Chr.1: 53906169-53906169 on Build GRCh38
3419323 c.394C>T p.R132C Chr.1: 53906151-53906151 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752617046 Chr.1: 53906064-53906064 on Build GRCh38
rs998078095 Chr.1: 53906103-53906103 on Build GRCh38
rs762243504 Chr.1: 53906151-53906151 on Build GRCh38
rs777476215 Chr.1: 53906071-53906071 on Build GRCh38
rs371590559 Chr.1: 53906080-53906080 on Build GRCh38
rs902145311 Chr.1: 53906165-53906165 on Build GRCh38
rs759168610 Chr.1: 53906166-53906166 on Build GRCh38
rs149919521 Chr.1: 53906106-53906106 on Build GRCh38
rs758344350 Chr.1: 53906067-53906067 on Build GRCh38
rs764720671 Chr.1: 53906063-53906063 on Build GRCh38
rs746117169 Chr.1: 53906096-53906096 on Build GRCh38
rs554503130 Chr.1: 53906105-53906105 on Build GRCh38
rs111776012 Chr.1: 53906053-53906053 on Build GRCh38
rs749757590 Chr.1: 53906111-53906111 on Build GRCh38
rs781454937 Chr.1: 53906092-53906092 on Build GRCh38
rs769293758 Chr.1: 53906119-53906119 on Build GRCh38
rs143295134 Chr.1: 53906164-53906164 on Build GRCh38
rs753478762 Chr.1: 53906050-53906050 on Build GRCh38
rs145006861 Chr.1: 53906152-53906152 on Build GRCh38
rs116474429 Chr.1: 53906072-53906072 on Build GRCh38
rs774804349 Chr.1: 53906142-53906142 on Build GRCh38

View Full Product Details