Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6843615-6843842 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNB3
Entrez Gene ID
2784
Gene Name
G protein subunit beta 3
Gene Aliases
CSNB1H
UniGene
Hs.631657
Gene Chromosome Location
Chr.12: 6840854-6847400 on Build GRCh38
Transcripts
NM_001297571.1 NM_002075.3 XM_011520953.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
942957 c.525G>A p.Q175Q Chr.12: 6843804-6843804 on Build GRCh38
5841803 c.487G>A p.D163N Chr.12: 6843688-6843688 on Build GRCh38
7119871 c.436C>G p.L146V Chr.12: 6843637-6843637 on Build GRCh38
1208412 c.512T>G p.I171S Chr.12: 6843791-6843791 on Build GRCh38
1241219 c.457G>T p.D153Y Chr.12: 6843658-6843658 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782530958 Chr.12: 6843741-6843741 on Build GRCh38
rs782683626 Chr.12: 6843778-6843778 on Build GRCh38
rs201972812 Chr.12: 6843766-6843766 on Build GRCh38
rs782199265 Chr.12: 6843641-6843641 on Build GRCh38
rs147291332 Chr.12: 6843820-6843820 on Build GRCh38
rs782269397 Chr.12: 6843649-6843649 on Build GRCh38
rs782392907 Chr.12: 6843801-6843801 on Build GRCh38
rs782321891 Chr.12: 6843824-6843824 on Build GRCh38
rs200309858 Chr.12: 6843639-6843639 on Build GRCh38
rs782199199 Chr.12: 6843765-6843765 on Build GRCh38
rs782054708 Chr.12: 6843673-6843673 on Build GRCh38
rs75977062 Chr.12: 6843684-6843684 on Build GRCh38
rs372997527 Chr.12: 6843742-6843742 on Build GRCh38
rs28395779 Chr.12: 6843756-6843756 on Build GRCh38
rs782351294 Chr.12: 6843663-6843663 on Build GRCh38
rs782317079 Chr.12: 6843775-6843775 on Build GRCh38
rs781988924 Chr.12: 6843657-6843657 on Build GRCh38
rs782432503 Chr.12: 6843642-6843642 on Build GRCh38
rs781918900 Chr.12: 6843825-6843825 on Build GRCh38
rs200864177 Chr.12: 6843660-6843660 on Build GRCh38
rs782615277 Chr.12: 6843763-6843763 on Build GRCh38
rs781988763 Chr.12: 6843659-6843659 on Build GRCh38
rs533829744 Chr.12: 6843739-6843739 on Build GRCh38
rs368740450 Chr.12: 6843648-6843648 on Build GRCh38
rs781898297 Chr.12: 6843710-6843710 on Build GRCh38
rs782697401 Chr.12: 6843701-6843701 on Build GRCh38
rs138319468 Chr.12: 6843783-6843783 on Build GRCh38
rs782022033 Chr.12: 6843690-6843690 on Build GRCh38
rs782313482 Chr.12: 6843792-6843793 on Build GRCh38
rs144008405 Chr.12: 6843791-6843791 on Build GRCh38
rs782486451 Chr.12: 6843757-6843757 on Build GRCh38
rs781940063 Chr.12: 6843666-6843666 on Build GRCh38
rs782388688 Chr.12: 6843650-6843650 on Build GRCh38
rs782130029 Chr.12: 6843695-6843695 on Build GRCh38

View Full Product Details