Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82722727-82723000 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FN3KRP
Entrez Gene ID
79672
Gene Name
fructosamine 3 kinase related protein
Gene Aliases
FN3KL
UniGene
Hs.31431
Gene Chromosome Location
Chr.17: 82716706-82728017 on Build GRCh38
Transcripts
NM_024619.3 NR_046408.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3821071 c.389G>A p.R130K Chr.17: 82722807-82722807 on Build GRCh38
3362419 c.399G>A p.G133G Chr.17: 82722817-82722817 on Build GRCh38
5853848 c.446C>T p.T149M Chr.17: 82722864-82722864 on Build GRCh38
252507 c.439G>A p.V147M Chr.17: 82722857-82722857 on Build GRCh38
4070938 c.468+2T>C p.? Chr.17: 82722888-82722888 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748074498 Chr.17: 82722883-82722883 on Build GRCh38
rs113865480 Chr.17: 82722887-82722887 on Build GRCh38
rs986981194 Chr.17: 82722898-82722898 on Build GRCh38
rs745498949 Chr.17: 82722800-82722800 on Build GRCh38
rs1038296991 Chr.17: 82722809-82722809 on Build GRCh38
rs779522933 Chr.17: 82722875-82722875 on Build GRCh38
rs751143747 Chr.17: 82722789-82722789 on Build GRCh38
rs753523680 Chr.17: 82722791-82722791 on Build GRCh38
rs778375179 Chr.17: 82722856-82722857 on Build GRCh38
rs527876447 Chr.17: 82722965-82722965 on Build GRCh38
rs1018547256 Chr.17: 82722869-82722869 on Build GRCh38
rs139582864 Chr.17: 82722807-82722807 on Build GRCh38
rs753672186 Chr.17: 82722937-82722937 on Build GRCh38
rs181065000 Chr.17: 82722820-82722820 on Build GRCh38
rs761463348 Chr.17: 82722859-82722859 on Build GRCh38
rs369912926 Chr.17: 82722900-82722900 on Build GRCh38
rs367983743 Chr.17: 82722895-82722895 on Build GRCh38
rs191683466 Chr.17: 82722884-82722884 on Build GRCh38
rs375407644 Chr.17: 82722847-82722847 on Build GRCh38
rs777846344 Chr.17: 82722886-82722886 on Build GRCh38
rs763557239 Chr.17: 82722909-82722909 on Build GRCh38
rs367615058 Chr.17: 82722941-82722941 on Build GRCh38
rs372108103 Chr.17: 82722896-82722896 on Build GRCh38
rs755524907 Chr.17: 82722784-82722784 on Build GRCh38
rs566764496 Chr.17: 82722957-82722957 on Build GRCh38
rs139863997 Chr.17: 82722864-82722864 on Build GRCh38
rs376489615 Chr.17: 82722935-82722935 on Build GRCh38
rs749970833 Chr.17: 82722925-82722925 on Build GRCh38
rs772127645 Chr.17: 82722834-82722834 on Build GRCh38
rs778881619 Chr.17: 82722882-82722882 on Build GRCh38
rs893353540 Chr.17: 82722876-82722876 on Build GRCh38
rs971954746 Chr.17: 82722958-82722958 on Build GRCh38
rs185031375 Chr.17: 82722750-82722750 on Build GRCh38
rs991689809 Chr.17: 82722905-82722905 on Build GRCh38
rs967016381 Chr.17: 82722885-82722885 on Build GRCh38
rs753708144 Chr.17: 82722880-82722880 on Build GRCh38
rs146131242 Chr.17: 82722856-82722856 on Build GRCh38
rs765616082 Chr.17: 82722787-82722787 on Build GRCh38
rs768409975 Chr.17: 82722753-82722753 on Build GRCh38
rs555615326 Chr.17: 82722865-82722865 on Build GRCh38
rs761086947 Chr.17: 82722768-82722768 on Build GRCh38
rs897156457 Chr.17: 82722812-82722812 on Build GRCh38
rs761843187 Chr.17: 82722756-82722756 on Build GRCh38
rs781037348 Chr.17: 82722796-82722796 on Build GRCh38
rs531592908 Chr.17: 82722844-82722844 on Build GRCh38
rs187091105 Chr.17: 82722769-82722769 on Build GRCh38
rs557222571 Chr.17: 82722892-82722892 on Build GRCh38
rs750014981 Chr.17: 82722884-82722893 on Build GRCh38
rs372059941 Chr.17: 82722827-82722827 on Build GRCh38
rs374288236 Chr.17: 82722843-82722843 on Build GRCh38
rs893316509 Chr.17: 82722920-82722920 on Build GRCh38
rs1002908912 Chr.17: 82722961-82722961 on Build GRCh38
rs144422621 Chr.17: 82722842-82722842 on Build GRCh38
rs376402941 Chr.17: 82722899-82722899 on Build GRCh38
rs748278731 Chr.17: 82722833-82722833 on Build GRCh38
rs767081360 Chr.17: 82722785-82722785 on Build GRCh38
rs756963029 Chr.17: 82722807-82722809 on Build GRCh38
rs373324194 Chr.17: 82722910-82722910 on Build GRCh38
rs764636360 Chr.17: 82722916-82722916 on Build GRCh38
rs371167020 Chr.17: 82722754-82722754 on Build GRCh38
rs757981368 Chr.17: 82722915-82722916 on Build GRCh38
rs921513464 Chr.17: 82722755-82722755 on Build GRCh38
rs754088836 Chr.17: 82722773-82722773 on Build GRCh38
rs773401531 Chr.17: 82722763-82722763 on Build GRCh38
rs201092965 Chr.17: 82722828-82722828 on Build GRCh38

View Full Product Details