Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 46941112-46941365 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP4A11
Entrez Gene ID
1579
Gene Name
cytochrome P450 family 4 subfamily A member 11
Gene Aliases
CP4Y, CYP4A2, CYP4AII
UniGene
Hs.1645
Gene Chromosome Location
Chr.1: 46929174-46941484 on Build GRCh38
Transcripts
NM_000778.3 NM_001319155.1 NR_134988.1 NR_134989.1 NR_134990.1 NR_134991.1 NR_134992.1 NR_134993.1 NR_134994.1 XM_005270539.1 XM_017000464.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5758840 c.176T>C p.L59P Chr.1: 46941258-46941258 on Build GRCh38
5381150 c.153C>T p.F51F Chr.1: 46941281-46941281 on Build GRCh38
2150851 c.165C>T p.P55P Chr.1: 46941269-46941269 on Build GRCh38
4921532 c.157T>C p.C53R Chr.1: 46941277-46941277 on Build GRCh38
305488 c.113A>G p.Y38C Chr.1: 46941321-46941321 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370538155 Chr.1: 46941253-46941253 on Build GRCh38
rs771237083 Chr.1: 46941322-46941322 on Build GRCh38
rs531728999 Chr.1: 46941203-46941203 on Build GRCh38
rs547380491 Chr.1: 46941148-46941148 on Build GRCh38
rs868630045 Chr.1: 46941297-46941297 on Build GRCh38
rs770769451 Chr.1: 46941237-46941237 on Build GRCh38
rs533214883 Chr.1: 46941201-46941201 on Build GRCh38
rs886713669 Chr.1: 46941137-46941137 on Build GRCh38
rs746393531 Chr.1: 46941206-46941206 on Build GRCh38
rs764259809 Chr.1: 46941256-46941256 on Build GRCh38
rs746748752 Chr.1: 46941234-46941234 on Build GRCh38
rs761075285 Chr.1: 46941314-46941314 on Build GRCh38
rs555853875 Chr.1: 46941278-46941278 on Build GRCh38
rs770625688 Chr.1: 46941309-46941309 on Build GRCh38
rs536828497 Chr.1: 46941273-46941273 on Build GRCh38
rs372477140 Chr.1: 46941194-46941194 on Build GRCh38
rs747664532 Chr.1: 46941211-46941211 on Build GRCh38
rs528916568 Chr.1: 46941138-46941138 on Build GRCh38
rs746660787 Chr.1: 46941304-46941304 on Build GRCh38
rs375391044 Chr.1: 46941271-46941271 on Build GRCh38
rs370654389 Chr.1: 46941218-46941218 on Build GRCh38
rs781095828 Chr.1: 46941310-46941310 on Build GRCh38
rs149204978 Chr.1: 46941330-46941330 on Build GRCh38
rs757551399 Chr.1: 46941265-46941265 on Build GRCh38
rs763129685 Chr.1: 46941254-46941254 on Build GRCh38
rs778258571 Chr.1: 46941274-46941274 on Build GRCh38
rs771896015 Chr.1: 46941222-46941222 on Build GRCh38
rs777506430 Chr.1: 46941299-46941299 on Build GRCh38
rs751960488 Chr.1: 46941198-46941198 on Build GRCh38
rs765216120 Chr.1: 46941244-46941244 on Build GRCh38
rs1005075622 Chr.1: 46941165-46941165 on Build GRCh38
rs375549495 Chr.1: 46941223-46941223 on Build GRCh38
rs764419430 Chr.1: 46941196-46941196 on Build GRCh38
rs759581767 Chr.1: 46941241-46941241 on Build GRCh38
rs561743612 Chr.1: 46941131-46941131 on Build GRCh38
rs767753685 Chr.1: 46941344-46941344 on Build GRCh38
rs762154121 Chr.1: 46941331-46941331 on Build GRCh38
rs775436920 Chr.1: 46941316-46941316 on Build GRCh38
rs758007868 Chr.1: 46941279-46941279 on Build GRCh38
rs762963432 Chr.1: 46941317-46941317 on Build GRCh38
rs776363366 Chr.1: 46941238-46941238 on Build GRCh38
rs751794931 Chr.1: 46941258-46941258 on Build GRCh38
rs745767999 Chr.1: 46941312-46941312 on Build GRCh38

View Full Product Details