Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 36923201-36923460 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSF2RB
Entrez Gene ID
1439
Gene Name
colony stimulating factor 2 receptor beta common subunit
Gene Aliases
CD131, CDw131, IL3RB, IL5RB, SMDP5
UniGene
Hs.592192
Gene Chromosome Location
Chr.22: 36913574-36940449 on Build GRCh38
Transcripts
NM_000395.2 XM_005261340.3 XM_011529903.2 XM_011529904.2 XM_011529905.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4004926 c.118A>C p.T40P Chr.22: 36923285-36923285 on Build GRCh38
5755745 c.128T>C p.I43T Chr.22: 36923295-36923295 on Build GRCh38
1416095 c.175G>A p.V59M Chr.22: 36923342-36923342 on Build GRCh38
1416096 c.199G>C p.E67Q Chr.22: 36923366-36923366 on Build GRCh38
3149942 c.92A>G p.Q31R Chr.22: 36923259-36923259 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202215481 Chr.22: 36923355-36923355 on Build GRCh38
rs778622632 Chr.22: 36923320-36923320 on Build GRCh38
rs758285488 Chr.22: 36923396-36923396 on Build GRCh38
rs776032106 Chr.22: 36923347-36923347 on Build GRCh38
rs750431877 Chr.22: 36923288-36923288 on Build GRCh38
rs768128646 Chr.22: 36923274-36923276 on Build GRCh38
rs750524951 Chr.22: 36923275-36923275 on Build GRCh38
rs199932177 Chr.22: 36923284-36923284 on Build GRCh38
rs2072713 Chr.22: 36923383-36923383 on Build GRCh38
rs975721272 Chr.22: 36923434-36923434 on Build GRCh38
rs376449769 Chr.22: 36923267-36923267 on Build GRCh38
rs748834535 Chr.22: 36923235-36923235 on Build GRCh38
rs756196738 Chr.22: 36923290-36923290 on Build GRCh38
rs781297912 Chr.22: 36923268-36923268 on Build GRCh38
rs760174329 Chr.22: 36923232-36923232 on Build GRCh38
rs768978226 Chr.22: 36923415-36923415 on Build GRCh38
rs371220018 Chr.22: 36923238-36923238 on Build GRCh38
rs763415308 Chr.22: 36923354-36923354 on Build GRCh38
rs370731851 Chr.22: 36923378-36923378 on Build GRCh38
rs912415224 Chr.22: 36923423-36923423 on Build GRCh38
rs775877225 Chr.22: 36923282-36923282 on Build GRCh38
rs746697642 Chr.22: 36923403-36923403 on Build GRCh38
rs756766865 Chr.22: 36923408-36923408 on Build GRCh38
rs116757501 Chr.22: 36923394-36923394 on Build GRCh38
rs377561426 Chr.22: 36923365-36923365 on Build GRCh38
rs149609232 Chr.22: 36923316-36923316 on Build GRCh38
rs192760202 Chr.22: 36923251-36923251 on Build GRCh38
rs374971985 Chr.22: 36923239-36923239 on Build GRCh38
rs373031830 Chr.22: 36923360-36923360 on Build GRCh38
rs751793975 Chr.22: 36923261-36923261 on Build GRCh38
rs751783111 Chr.22: 36923283-36923283 on Build GRCh38
rs746462818 Chr.22: 36923341-36923341 on Build GRCh38
rs919723011 Chr.22: 36923234-36923234 on Build GRCh38
rs201476440 Chr.22: 36923285-36923285 on Build GRCh38
rs771689873 Chr.22: 36923280-36923280 on Build GRCh38
rs777557495 Chr.22: 36923397-36923397 on Build GRCh38
rs368852888 Chr.22: 36923336-36923336 on Build GRCh38
rs1026652277 Chr.22: 36923407-36923407 on Build GRCh38
rs772800054 Chr.22: 36923281-36923281 on Build GRCh38
rs138299199 Chr.22: 36923278-36923278 on Build GRCh38
rs770407372 Chr.22: 36923342-36923342 on Build GRCh38
rs771814455 Chr.22: 36923335-36923335 on Build GRCh38
rs372145171 Chr.22: 36923254-36923254 on Build GRCh38
rs753787293 Chr.22: 36923309-36923309 on Build GRCh38
rs370532273 Chr.22: 36923433-36923433 on Build GRCh38
rs373253443 Chr.22: 36923379-36923379 on Build GRCh38
rs370015700 Chr.22: 36923410-36923410 on Build GRCh38
rs372621269 Chr.22: 36923411-36923411 on Build GRCh38
rs777250876 Chr.22: 36923228-36923228 on Build GRCh38
rs1037349119 Chr.22: 36923356-36923356 on Build GRCh38
rs778626873 Chr.22: 36923279-36923279 on Build GRCh38
rs370000670 Chr.22: 36923331-36923331 on Build GRCh38
rs774534225 Chr.22: 36923357-36923357 on Build GRCh38
rs538023924 Chr.22: 36923422-36923422 on Build GRCh38
rs780025665 Chr.22: 36923292-36923292 on Build GRCh38
rs1033966655 Chr.22: 36923395-36923395 on Build GRCh38
rs756002403 Chr.22: 36923277-36923277 on Build GRCh38
rs754822869 Chr.22: 36923384-36923384 on Build GRCh38

View Full Product Details