Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 73085126-73085369 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD17
Entrez Gene ID
26057
Gene Name
ankyrin repeat domain 17
Gene Aliases
GTAR, MASK2, NY-BR-16
UniGene
Hs.601206
Gene Chromosome Location
Chr.4: 73073966-73259142 on Build GRCh38
Transcripts
NM_001286771.2 NM_015574.1 NM_032217.4 NM_198889.2 XM_005265671.3 XM_017008011.1 XM_017008012.1 XM_017008013.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7090310 c.7121C>A p.P2374Q Chr.4: 73085287-73085287 on Build GRCh38
5544478 c.7138C>T p.P2380S Chr.4: 73085270-73085270 on Build GRCh38
334604 c.7121C>T p.P2374L Chr.4: 73085287-73085287 on Build GRCh38
5995556 c.7055A>C p.Y2352S Chr.4: 73085353-73085353 on Build GRCh38
1173064 c.7087G>A p.A2363T Chr.4: 73085321-73085321 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761087814 Chr.4: 73085285-73085285 on Build GRCh38
rs762523913 Chr.4: 73085321-73085321 on Build GRCh38
rs532129013 Chr.4: 73085204-73085204 on Build GRCh38
rs773760971 Chr.4: 73085284-73085284 on Build GRCh38
rs1030656209 Chr.4: 73085346-73085346 on Build GRCh38
rs749474501 Chr.4: 73085236-73085236 on Build GRCh38
rs115181919 Chr.4: 73085192-73085192 on Build GRCh38
rs756020462 Chr.4: 73085341-73085341 on Build GRCh38
rs757166478 Chr.4: 73085327-73085327 on Build GRCh38
rs752354539 Chr.4: 73085320-73085320 on Build GRCh38
rs751455851 Chr.4: 73085323-73085323 on Build GRCh38
rs769029980 Chr.4: 73085240-73085240 on Build GRCh38
rs906833364 Chr.4: 73085301-73085301 on Build GRCh38
rs989878826 Chr.4: 73085232-73085232 on Build GRCh38
rs764850426 Chr.4: 73085319-73085319 on Build GRCh38
rs374501403 Chr.4: 73085268-73085268 on Build GRCh38
rs771668773 Chr.4: 73085162-73085162 on Build GRCh38
rs146616951 Chr.4: 73085286-73085286 on Build GRCh38
rs540568077 Chr.4: 73085172-73085172 on Build GRCh38
rs781149975 Chr.4: 73085233-73085233 on Build GRCh38
rs759938048 Chr.4: 73085287-73085287 on Build GRCh38
rs777757286 Chr.4: 73085215-73085215 on Build GRCh38
rs781094933 Chr.4: 73085328-73085328 on Build GRCh38
rs191971603 Chr.4: 73085212-73085212 on Build GRCh38
rs1030559579 Chr.4: 73085182-73085182 on Build GRCh38
rs370454898 Chr.4: 73085227-73085227 on Build GRCh38
rs774840532 Chr.4: 73085256-73085256 on Build GRCh38
rs972302882 Chr.4: 73085163-73085163 on Build GRCh38
rs150785652 Chr.4: 73085337-73085337 on Build GRCh38
rs986450112 Chr.4: 73085208-73085208 on Build GRCh38

View Full Product Details