Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 4571482-4571753 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf96
Entrez Gene ID
342346
Gene Name
chromosome 16 open reading frame 96
Gene Aliases
-
UniGene
Hs.684212
Gene Chromosome Location
Chr.16: 4538709-4600455 on Build GRCh38
Transcripts
NM_001145011.1 XM_005255298.3 XM_005255301.3 XM_011522475.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3712032 c.522C>A p.D174E Chr.16: 4571662-4571662 on Build GRCh38
2920639 c.481G>A p.A161T Chr.16: 4571621-4571621 on Build GRCh38
4496875 c.484C>T p.L162F Chr.16: 4571624-4571624 on Build GRCh38
6842469 c.498G>A p.V166V Chr.16: 4571638-4571638 on Build GRCh38
4749049 c.455T>G p.L152R Chr.16: 4571595-4571595 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs202099268 Chr.16: 4571725-4571725 on Build GRCh38
rs553232591 Chr.16: 4571616-4571616 on Build GRCh38
rs773256941 Chr.16: 4571515-4571515 on Build GRCh38
rs1050364962 Chr.16: 4571608-4571608 on Build GRCh38
rs191330425 Chr.16: 4571641-4571641 on Build GRCh38
rs773449144 Chr.16: 4571701-4571701 on Build GRCh38
rs35531180 Chr.16: 4571725-4571725 on Build GRCh38
rs114176089 Chr.16: 4571710-4571710 on Build GRCh38
rs931988804 Chr.16: 4571598-4571598 on Build GRCh38
rs554999273 Chr.16: 4571677-4571677 on Build GRCh38
rs375062189 Chr.16: 4571530-4571530 on Build GRCh38
rs940991081 Chr.16: 4571558-4571558 on Build GRCh38
rs777699842 Chr.16: 4571724-4571725 on Build GRCh38
rs550610802 Chr.16: 4571528-4571528 on Build GRCh38
rs767903079 Chr.16: 4571571-4571571 on Build GRCh38
rs80324612 Chr.16: 4571692-4571692 on Build GRCh38
rs1010887171 Chr.16: 4571633-4571633 on Build GRCh38
rs903961415 Chr.16: 4571636-4571636 on Build GRCh38
rs896087207 Chr.16: 4571605-4571605 on Build GRCh38
rs763051053 Chr.16: 4571518-4571518 on Build GRCh38
rs904523717 Chr.16: 4571705-4571705 on Build GRCh38
rs201040249 Chr.16: 4571721-4571721 on Build GRCh38
rs753142119 Chr.16: 4571575-4571575 on Build GRCh38
rs376409573 Chr.16: 4571602-4571602 on Build GRCh38
rs971280097 Chr.16: 4571712-4571712 on Build GRCh38
rs891621416 Chr.16: 4571675-4571675 on Build GRCh38
rs1041017437 Chr.16: 4571564-4571564 on Build GRCh38
rs756410611 Chr.16: 4571597-4571597 on Build GRCh38
rs371188041 Chr.16: 4571645-4571645 on Build GRCh38
rs779657343 Chr.16: 4571653-4571653 on Build GRCh38
rs147839625 Chr.16: 4571539-4571539 on Build GRCh38

View Full Product Details