Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 130470875-130471140 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS15
Entrez Gene ID
170689
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 15
Gene Aliases
-
UniGene
Hs.534221
Gene Chromosome Location
Chr.11: 130448876-130478215 on Build GRCh38
Transcripts
NM_139055.3 XM_005271419.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6593932 c.1838G>A p.R613Q Chr.11: 130471037-130471037 on Build GRCh38
7237682 c.1868A>G p.N623S Chr.11: 130471067-130471067 on Build GRCh38
5368283 c.1765G>A p.G589S Chr.11: 130470964-130470964 on Build GRCh38
3967419 c.1801G>A p.V601M Chr.11: 130471000-130471000 on Build GRCh38
226700 c.1897C>T p.P633S Chr.11: 130471096-130471096 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs555562930 Chr.11: 130470986-130470986 on Build GRCh38
rs754674583 Chr.11: 130470976-130470976 on Build GRCh38
rs762711694 Chr.11: 130470923-130470923 on Build GRCh38
rs755760662 Chr.11: 130471073-130471073 on Build GRCh38
rs373000118 Chr.11: 130470985-130470985 on Build GRCh38
rs377624869 Chr.11: 130471110-130471110 on Build GRCh38
rs936236779 Chr.11: 130471109-130471109 on Build GRCh38
rs755615911 Chr.11: 130471068-130471068 on Build GRCh38
rs757668783 Chr.11: 130470960-130470960 on Build GRCh38
rs200393200 Chr.11: 130470963-130470963 on Build GRCh38
rs765474969 Chr.11: 130471037-130471037 on Build GRCh38
rs201640202 Chr.11: 130470902-130470902 on Build GRCh38
rs758979259 Chr.11: 130471053-130471053 on Build GRCh38
rs150734073 Chr.11: 130471017-130471017 on Build GRCh38
rs1017560645 Chr.11: 130470999-130470999 on Build GRCh38
rs778530267 Chr.11: 130471094-130471094 on Build GRCh38
rs375413440 Chr.11: 130471023-130471023 on Build GRCh38
rs927601598 Chr.11: 130471098-130471098 on Build GRCh38
rs867034669 Chr.11: 130471022-130471022 on Build GRCh38
rs766547214 Chr.11: 130471078-130471078 on Build GRCh38
rs745647777 Chr.11: 130471100-130471100 on Build GRCh38
rs751977582 Chr.11: 130471060-130471060 on Build GRCh38
rs776200888 Chr.11: 130470924-130470924 on Build GRCh38
rs141261317 Chr.11: 130470948-130470948 on Build GRCh38
rs764405782 Chr.11: 130471026-130471026 on Build GRCh38
rs769794337 Chr.11: 130470908-130470908 on Build GRCh38
rs760756603 Chr.11: 130471024-130471024 on Build GRCh38
rs369119460 Chr.11: 130470950-130470950 on Build GRCh38
rs146430548 Chr.11: 130471097-130471097 on Build GRCh38
rs750965110 Chr.11: 130471046-130471046 on Build GRCh38
rs765811154 Chr.11: 130470962-130470962 on Build GRCh38
rs777266263 Chr.11: 130471072-130471072 on Build GRCh38
rs749289528 Chr.11: 130471011-130471011 on Build GRCh38
rs760951777 Chr.11: 130470938-130470938 on Build GRCh38
rs79839754 Chr.11: 130470922-130470922 on Build GRCh38
rs758826755 Chr.11: 130470964-130470964 on Build GRCh38
rs766792716 Chr.11: 130471059-130471059 on Build GRCh38
rs759166118 Chr.11: 130470996-130470996 on Build GRCh38
rs148676964 Chr.11: 130470944-130470944 on Build GRCh38
rs11222114 Chr.11: 130471067-130471067 on Build GRCh38
rs780632888 Chr.11: 130470968-130470968 on Build GRCh38
rs746576565 Chr.11: 130470969-130470969 on Build GRCh38
rs762717576 Chr.11: 130471085-130471085 on Build GRCh38
rs147190472 Chr.11: 130470925-130470925 on Build GRCh38
rs371359279 Chr.11: 130471113-130471113 on Build GRCh38
rs780874749 Chr.11: 130471061-130471061 on Build GRCh38
rs377184006 Chr.11: 130470997-130470997 on Build GRCh38
rs775682090 Chr.11: 130470937-130470937 on Build GRCh38
rs764019858 Chr.11: 130470941-130470941 on Build GRCh38
rs776761397 Chr.11: 130471027-130471027 on Build GRCh38
rs139100319 Chr.11: 130471036-130471036 on Build GRCh38
rs370391190 Chr.11: 130471006-130471006 on Build GRCh38
rs374196474 Chr.11: 130471089-130471089 on Build GRCh38
rs776903816 Chr.11: 130471112-130471112 on Build GRCh38

View Full Product Details