Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 92717730-92717999 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BICD2
Entrez Gene ID
23299
Gene Name
BICD cargo adaptor 2
Gene Aliases
SMALED2, bA526D8.1
UniGene
Hs.436939
Gene Chromosome Location
Chr.9: 92711363-92764841 on Build GRCh38
Transcripts
NM_001003800.1 NM_015250.3 XM_017014551.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4478383 c.2234C>T p.S745L Chr.9: 92717821-92717821 on Build GRCh38
4820387 c.2240G>A p.R747H Chr.9: 92717815-92717815 on Build GRCh38
4665702 c.2170G>A p.E724K Chr.9: 92717885-92717885 on Build GRCh38
4794226 c.2119G>A p.A707T Chr.9: 92717936-92717936 on Build GRCh38
4794227 c.2119G>A p.A707T Chr.9: 92717936-92717936 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs554612134 Chr.9: 92717780-92717780 on Build GRCh38
rs759865700 Chr.9: 92717939-92717939 on Build GRCh38
rs541181084 Chr.9: 92717766-92717766 on Build GRCh38
rs746627069 Chr.9: 92717966-92717966 on Build GRCh38
rs566572599 Chr.9: 92717791-92717791 on Build GRCh38
rs376177099 Chr.9: 92717779-92717779 on Build GRCh38
rs750839993 Chr.9: 92717790-92717790 on Build GRCh38
rs774315888 Chr.9: 92717772-92717772 on Build GRCh38
rs145923602 Chr.9: 92717810-92717810 on Build GRCh38
rs780658966 Chr.9: 92717970-92717970 on Build GRCh38
rs142140690 Chr.9: 92717838-92717838 on Build GRCh38
rs910755631 Chr.9: 92717761-92717761 on Build GRCh38
rs770798582 Chr.9: 92717770-92717770 on Build GRCh38
rs79574551 Chr.9: 92717969-92717969 on Build GRCh38
rs755995595 Chr.9: 92717760-92717760 on Build GRCh38
rs757247225 Chr.9: 92717955-92717955 on Build GRCh38
rs771619173 Chr.9: 92717871-92717871 on Build GRCh38
rs781440350 Chr.9: 92717821-92717821 on Build GRCh38
rs368924457 Chr.9: 92717964-92717964 on Build GRCh38
rs777927601 Chr.9: 92717764-92717764 on Build GRCh38
rs563633956 Chr.9: 92717877-92717877 on Build GRCh38
rs201335175 Chr.9: 92717883-92717883 on Build GRCh38
rs531116133 Chr.9: 92717961-92717961 on Build GRCh38
rs929825881 Chr.9: 92717786-92717786 on Build GRCh38
rs779967012 Chr.9: 92717749-92717749 on Build GRCh38
rs745733355 Chr.9: 92717867-92717867 on Build GRCh38
rs373990508 Chr.9: 92717820-92717820 on Build GRCh38
rs763445408 Chr.9: 92717904-92717904 on Build GRCh38
rs777986224 Chr.9: 92717913-92717913 on Build GRCh38
rs747166870 Chr.9: 92717752-92717752 on Build GRCh38
rs878944571 Chr.9: 92717815-92717815 on Build GRCh38
rs916266142 Chr.9: 92717769-92717769 on Build GRCh38
rs201695979 Chr.9: 92717963-92717963 on Build GRCh38
rs745474543 Chr.9: 92717774-92717774 on Build GRCh38
rs766685844 Chr.9: 92717802-92717802 on Build GRCh38
rs1049337357 Chr.9: 92717751-92717751 on Build GRCh38
rs756538071 Chr.9: 92717863-92717863 on Build GRCh38
rs943974428 Chr.9: 92717866-92717866 on Build GRCh38
rs200133999 Chr.9: 92717862-92717862 on Build GRCh38
rs969083172 Chr.9: 92717775-92717775 on Build GRCh38
rs767670894 Chr.9: 92717943-92717943 on Build GRCh38
rs1042399429 Chr.9: 92717910-92717910 on Build GRCh38
rs371707778 Chr.9: 92717947-92717947 on Build GRCh38
rs776152202 Chr.9: 92717885-92717885 on Build GRCh38
rs140501870 Chr.9: 92717782-92717782 on Build GRCh38
rs750330886 Chr.9: 92717962-92717962 on Build GRCh38
rs770335133 Chr.9: 92717886-92717886 on Build GRCh38
rs746654103 Chr.9: 92717880-92717880 on Build GRCh38
rs367722713 Chr.9: 92717859-92717859 on Build GRCh38
rs765546977 Chr.9: 92717784-92717784 on Build GRCh38
rs751750400 Chr.9: 92717919-92717919 on Build GRCh38

View Full Product Details