Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 49019983-49020256 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSHR
Entrez Gene ID
2492
Gene Name
follicle stimulating hormone receptor
Gene Aliases
FSHR1, FSHRO, LGR1, ODG1
UniGene
Hs.1428
Gene Chromosome Location
Chr.2: 48953161-49154527 on Build GRCh38
Transcripts
NM_000145.3 NM_181446.2 XM_011532733.2 XM_011532734.2 XM_011532740.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6091884 c.288A>T p.K96N Chr.2: 49020097-49020097 on Build GRCh38
5810989 c.294T>A p.H98Q Chr.2: 49020091-49020091 on Build GRCh38
1408404 c.255G>T p.V85V Chr.2: 49020130-49020130 on Build GRCh38
3682661 c.269T>G p.V90G Chr.2: 49020116-49020116 on Build GRCh38
3186547 c.259G>A p.E87K Chr.2: 49020126-49020126 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780215158 Chr.2: 49020057-49020057 on Build GRCh38
rs140960768 Chr.2: 49020120-49020120 on Build GRCh38
rs1019057992 Chr.2: 49020008-49020008 on Build GRCh38
rs145775250 Chr.2: 49020122-49020122 on Build GRCh38
rs190759257 Chr.2: 49020066-49020066 on Build GRCh38
rs749571389 Chr.2: 49020145-49020145 on Build GRCh38
rs374642025 Chr.2: 49020041-49020041 on Build GRCh38
rs537551534 Chr.2: 49020204-49020204 on Build GRCh38
rs199548127 Chr.2: 49020175-49020175 on Build GRCh38
rs574070235 Chr.2: 49020115-49020115 on Build GRCh38
rs767592976 Chr.2: 49020185-49020185 on Build GRCh38
rs201949594 Chr.2: 49020156-49020156 on Build GRCh38
rs761775278 Chr.2: 49020183-49020183 on Build GRCh38
rs556289091 Chr.2: 49020174-49020174 on Build GRCh38
rs914924401 Chr.2: 49020189-49020189 on Build GRCh38
rs750192853 Chr.2: 49020190-49020190 on Build GRCh38
rs768583889 Chr.2: 49020157-49020157 on Build GRCh38
rs762349327 Chr.2: 49020082-49020082 on Build GRCh38
rs888330143 Chr.2: 49020060-49020060 on Build GRCh38
rs561658647 Chr.2: 49020014-49020014 on Build GRCh38
rs772576285 Chr.2: 49020169-49020169 on Build GRCh38
rs753998909 Chr.2: 49020199-49020199 on Build GRCh38
rs755948857 Chr.2: 49020197-49020197 on Build GRCh38
rs965264401 Chr.2: 49020067-49020067 on Build GRCh38
rs779111613 Chr.2: 49020158-49020158 on Build GRCh38
rs774975627 Chr.2: 49020179-49020179 on Build GRCh38
rs187697780 Chr.2: 49020218-49020218 on Build GRCh38
rs773583739 Chr.2: 49020068-49020068 on Build GRCh38
rs780363421 Chr.2: 49020178-49020178 on Build GRCh38
rs773563862 Chr.2: 49020169-49020169 on Build GRCh38
rs200981622 Chr.2: 49020076-49020076 on Build GRCh38
rs147234312 Chr.2: 49020021-49020021 on Build GRCh38
rs748549199 Chr.2: 49020159-49020159 on Build GRCh38
rs951973760 Chr.2: 49020038-49020039 on Build GRCh38
rs267599405 Chr.2: 49020124-49020124 on Build GRCh38
rs769155566 Chr.2: 49020061-49020061 on Build GRCh38
rs770383357 Chr.2: 49020168-49020168 on Build GRCh38
rs371646619 Chr.2: 49020038-49020038 on Build GRCh38
rs750246214 Chr.2: 49020128-49020128 on Build GRCh38
rs147298183 Chr.2: 49020109-49020109 on Build GRCh38
rs202122664 Chr.2: 49020058-49020058 on Build GRCh38
rs948282963 Chr.2: 49020214-49020214 on Build GRCh38
rs751093894 Chr.2: 49020182-49020182 on Build GRCh38
rs78784563 Chr.2: 49020012-49020012 on Build GRCh38
rs781199640 Chr.2: 49020050-49020050 on Build GRCh38
rs755363484 Chr.2: 49020225-49020225 on Build GRCh38
rs749891700 Chr.2: 49020063-49020063 on Build GRCh38
rs755282386 Chr.2: 49020201-49020201 on Build GRCh38
rs770152782 Chr.2: 49020054-49020054 on Build GRCh38
rs756362810 Chr.2: 49020139-49020139 on Build GRCh38
rs989065378 Chr.2: 49020161-49020161 on Build GRCh38
rs555904948 Chr.2: 49020099-49020099 on Build GRCh38
rs767034426 Chr.2: 49020037-49020037 on Build GRCh38
rs550320631 Chr.2: 49020023-49020023 on Build GRCh38
rs759126590 Chr.2: 49020069-49020069 on Build GRCh38
rs183931186 Chr.2: 49020072-49020072 on Build GRCh38
rs939415623 Chr.2: 49020015-49020015 on Build GRCh38
rs377550562 Chr.2: 49020092-49020092 on Build GRCh38
rs886056152 Chr.2: 49020171-49020171 on Build GRCh38
rs758319261 Chr.2: 49020036-49020036 on Build GRCh38
rs201628139 Chr.2: 49020205-49020205 on Build GRCh38
rs756972314 Chr.2: 49020045-49020045 on Build GRCh38
rs369073966 Chr.2: 49020046-49020046 on Build GRCh38
rs1026267352 Chr.2: 49020033-49020033 on Build GRCh38
rs528312254 Chr.2: 49020004-49020004 on Build GRCh38
rs763159868 Chr.2: 49020168-49020169 on Build GRCh38
rs140715301 Chr.2: 49020053-49020053 on Build GRCh38

View Full Product Details