Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 37374032-37374291 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELFN2
Entrez Gene ID
114794
Gene Name
extracellular leucine rich repeat and fibronectin type III domain containing 2
Gene Aliases
LRRC62, PPP1R29
UniGene
Hs.344170
Gene Chromosome Location
Chr.22: 37340644-37427470 on Build GRCh38
Transcripts
NM_052906.4 NR_110512.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7189900 c.1498G>A p.G500S Chr.22: 37374037-37374037 on Build GRCh38
5563750 c.1369C>A p.Q457K Chr.22: 37374166-37374166 on Build GRCh38
6653269 c.1349G>A p.G450D Chr.22: 37374186-37374186 on Build GRCh38
7098123 c.1348G>A p.G450S Chr.22: 37374187-37374187 on Build GRCh38
5638241 c.1366G>A p.A456T Chr.22: 37374169-37374169 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760924048 Chr.22: 37374152-37374152 on Build GRCh38
rs767275235 Chr.22: 37374267-37374267 on Build GRCh38
rs201073943 Chr.22: 37374086-37374086 on Build GRCh38
rs780661723 Chr.22: 37374130-37374130 on Build GRCh38
rs777615510 Chr.22: 37374128-37374128 on Build GRCh38
rs774091407 Chr.22: 37374244-37374244 on Build GRCh38
rs145219811 Chr.22: 37374173-37374173 on Build GRCh38
rs372926360 Chr.22: 37374270-37374270 on Build GRCh38
rs376193256 Chr.22: 37374072-37374072 on Build GRCh38
rs764989723 Chr.22: 37374226-37374228 on Build GRCh38
rs568828697 Chr.22: 37374136-37374136 on Build GRCh38
rs748721475 Chr.22: 37374139-37374139 on Build GRCh38
rs779577089 Chr.22: 37374137-37374137 on Build GRCh38
rs776247026 Chr.22: 37374064-37374064 on Build GRCh38
rs1001070569 Chr.22: 37374271-37374271 on Build GRCh38
rs771132559 Chr.22: 37374210-37374210 on Build GRCh38
rs776311867 Chr.22: 37374063-37374063 on Build GRCh38
rs749986197 Chr.22: 37374187-37374187 on Build GRCh38
rs772753160 Chr.22: 37374202-37374202 on Build GRCh38
rs764482250 Chr.22: 37374060-37374060 on Build GRCh38
rs751396260 Chr.22: 37374174-37374174 on Build GRCh38
rs756964166 Chr.22: 37374176-37374176 on Build GRCh38
rs745351154 Chr.22: 37374067-37374067 on Build GRCh38
rs750226418 Chr.22: 37374268-37374268 on Build GRCh38
rs201026516 Chr.22: 37374155-37374155 on Build GRCh38
rs777403678 Chr.22: 37374051-37374051 on Build GRCh38
rs761684294 Chr.22: 37374254-37374254 on Build GRCh38
rs759482053 Chr.22: 37374103-37374103 on Build GRCh38
rs771204594 Chr.22: 37374154-37374154 on Build GRCh38
rs142330872 Chr.22: 37374205-37374205 on Build GRCh38
rs61743097 Chr.22: 37374068-37374068 on Build GRCh38
rs776922187 Chr.22: 37374211-37374211 on Build GRCh38
rs559019194 Chr.22: 37374206-37374206 on Build GRCh38
rs762273969 Chr.22: 37374077-37374077 on Build GRCh38
rs529899777 Chr.22: 37374151-37374151 on Build GRCh38
rs201382396 Chr.22: 37374114-37374114 on Build GRCh38
rs779274527 Chr.22: 37374161-37374161 on Build GRCh38
rs753998297 Chr.22: 37374092-37374092 on Build GRCh38
rs773698358 Chr.22: 37374081-37374081 on Build GRCh38
rs142413918 Chr.22: 37374104-37374104 on Build GRCh38
rs964842465 Chr.22: 37374214-37374214 on Build GRCh38
rs199993017 Chr.22: 37374238-37374238 on Build GRCh38
rs570843257 Chr.22: 37374235-37374235 on Build GRCh38
rs759661421 Chr.22: 37374157-37374157 on Build GRCh38
rs746725384 Chr.22: 37374052-37374052 on Build GRCh38
rs373313833 Chr.22: 37374135-37374135 on Build GRCh38
rs150864906 Chr.22: 37374115-37374115 on Build GRCh38
rs748776432 Chr.22: 37374197-37374197 on Build GRCh38
rs370109757 Chr.22: 37374129-37374129 on Build GRCh38
rs765239864 Chr.22: 37374169-37374169 on Build GRCh38
rs759747163 Chr.22: 37374220-37374220 on Build GRCh38
rs996593605 Chr.22: 37374237-37374237 on Build GRCh38
rs780903748 Chr.22: 37374177-37374177 on Build GRCh38
rs755815695 Chr.22: 37374188-37374188 on Build GRCh38
rs762922500 Chr.22: 37374172-37374172 on Build GRCh38
rs779776054 Chr.22: 37374196-37374196 on Build GRCh38
rs139310200 Chr.22: 37374146-37374146 on Build GRCh38
rs375959244 Chr.22: 37374236-37374236 on Build GRCh38
rs7292855 Chr.22: 37374170-37374170 on Build GRCh38
rs140163106 Chr.22: 37374085-37374085 on Build GRCh38

View Full Product Details