Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 68468619-68468892 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8orf34
Entrez Gene ID
116328
Gene Name
chromosome 8 open reading frame 34
Gene Aliases
VEST-1, VEST1
UniGene
Hs.491941
Gene Chromosome Location
Chr.8: 68330722-68819023 on Build GRCh38
Transcripts
NM_001195639.1 NM_052958.2 XM_011517445.2 XM_011517446.2 XM_011517448.2 XM_011517449.2 XM_011517450.2 XM_017013019.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7420200 c.386T>A p.L129H Chr.8: 68468803-68468803 on Build GRCh38
4966202 c.285A>C p.S95S Chr.8: 68468702-68468702 on Build GRCh38
6616705 c.332A>T p.D111V Chr.8: 68468749-68468749 on Build GRCh38
3942731 c.357G>T p.R119S Chr.8: 68468699-68468699 on Build GRCh38
5580464 c.424C>T p.H142Y Chr.8: 68468766-68468766 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs923363019 Chr.8: 68468870-68468870 on Build GRCh38
rs143049728 Chr.8: 68468755-68468755 on Build GRCh38
rs933848949 Chr.8: 68468859-68468859 on Build GRCh38
rs200920530 Chr.8: 68468743-68468743 on Build GRCh38
rs370223851 Chr.8: 68468717-68468717 on Build GRCh38
rs763529734 Chr.8: 68468761-68468761 on Build GRCh38
rs867590263 Chr.8: 68468762-68468762 on Build GRCh38
rs537514672 Chr.8: 68468723-68468723 on Build GRCh38
rs61736081 Chr.8: 68468818-68468818 on Build GRCh38
rs759948704 Chr.8: 68468745-68468745 on Build GRCh38
rs3210875 Chr.8: 68468790-68468790 on Build GRCh38
rs746340923 Chr.8: 68468860-68468860 on Build GRCh38
rs771359309 Chr.8: 68468736-68468736 on Build GRCh38
rs535057811 Chr.8: 68468704-68468704 on Build GRCh38
rs985574387 Chr.8: 68468866-68468866 on Build GRCh38
rs777718252 Chr.8: 68468821-68468821 on Build GRCh38
rs999022082 Chr.8: 68468768-68468768 on Build GRCh38
rs764655541 Chr.8: 68468863-68468863 on Build GRCh38
rs756826811 Chr.8: 68468792-68468792 on Build GRCh38
rs753745947 Chr.8: 68468664-68468664 on Build GRCh38
rs755624929 Chr.8: 68468655-68468655 on Build GRCh38
rs753326129 Chr.8: 68468771-68468771 on Build GRCh38
rs957664985 Chr.8: 68468810-68468811 on Build GRCh38
rs868113803 Chr.8: 68468748-68468748 on Build GRCh38
rs761197839 Chr.8: 68468675-68468676 on Build GRCh38
rs866337168 Chr.8: 68468775-68468775 on Build GRCh38
rs895900501 Chr.8: 68468682-68468682 on Build GRCh38
rs771981922 Chr.8: 68468702-68468702 on Build GRCh38
rs554947856 Chr.8: 68468710-68468710 on Build GRCh38
rs745725860 Chr.8: 68468699-68468699 on Build GRCh38
rs142064557 Chr.8: 68468746-68468746 on Build GRCh38
rs750364553 Chr.8: 68468797-68468797 on Build GRCh38
rs748338743 Chr.8: 68468817-68468817 on Build GRCh38
rs760329354 Chr.8: 68468759-68468759 on Build GRCh38
rs757206278 Chr.8: 68468683-68468683 on Build GRCh38
rs1023007134 Chr.8: 68468708-68468708 on Build GRCh38
rs1014719154 Chr.8: 68468696-68468696 on Build GRCh38
rs779860135 Chr.8: 68468816-68468816 on Build GRCh38
rs777300568 Chr.8: 68468656-68468656 on Build GRCh38
rs773480488 Chr.8: 68468676-68468676 on Build GRCh38
rs758310264 Chr.8: 68468799-68468799 on Build GRCh38
rs770981093 Chr.8: 68468854-68468854 on Build GRCh38
rs778858551 Chr.8: 68468684-68468684 on Build GRCh38
rs765323437 Chr.8: 68468793-68468793 on Build GRCh38
rs763462550 Chr.8: 68468729-68468729 on Build GRCh38
rs766775499 Chr.8: 68468826-68468826 on Build GRCh38
rs749436126 Chr.8: 68468853-68468853 on Build GRCh38
rs150118220 Chr.8: 68468856-68468856 on Build GRCh38

View Full Product Details