Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41973768-41974003 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNP
Entrez Gene ID
1267
Gene Name
2',3'-cyclic nucleotide 3' phosphodiesterase
Gene Aliases
CNP1
UniGene
Hs.273621
Gene Chromosome Location
Chr.17: 41966741-41977736 on Build GRCh38
Transcripts
NM_033133.4 XM_006721701.3 XM_011524340.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4130076 c.1194C>A p.Y398* Chr.17: 41973852-41973852 on Build GRCh38
3755547 c.1128G>A p.G376G Chr.17: 41973846-41973846 on Build GRCh38
4066492 c.1181C>G p.A394G Chr.17: 41973899-41973899 on Build GRCh38
7332121 c.1086G>A p.L362L Chr.17: 41973804-41973804 on Build GRCh38
4130077 c.1134C>A p.Y378* Chr.17: 41973852-41973852 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782020383 Chr.17: 41973919-41973919 on Build GRCh38
rs367704141 Chr.17: 41973852-41973852 on Build GRCh38
rs529263304 Chr.17: 41973946-41973946 on Build GRCh38
rs782456858 Chr.17: 41973812-41973812 on Build GRCh38
rs782800795 Chr.17: 41973880-41973880 on Build GRCh38
rs200980179 Chr.17: 41973876-41973876 on Build GRCh38
rs782439529 Chr.17: 41973909-41973909 on Build GRCh38
rs782266279 Chr.17: 41973920-41973920 on Build GRCh38
rs781852887 Chr.17: 41973949-41973949 on Build GRCh38
rs372142717 Chr.17: 41973887-41973887 on Build GRCh38
rs782178419 Chr.17: 41973901-41973901 on Build GRCh38
rs781813740 Chr.17: 41973964-41973964 on Build GRCh38
rs782007705 Chr.17: 41973872-41973872 on Build GRCh38
rs782063304 Chr.17: 41973932-41973932 on Build GRCh38
rs782695579 Chr.17: 41973935-41973935 on Build GRCh38
rs782166990 Chr.17: 41973859-41973859 on Build GRCh38
rs782433845 Chr.17: 41973823-41973823 on Build GRCh38
rs1016737269 Chr.17: 41973977-41973977 on Build GRCh38
rs575459583 Chr.17: 41973832-41973832 on Build GRCh38
rs782730083 Chr.17: 41973962-41973962 on Build GRCh38
rs540321367 Chr.17: 41973886-41973886 on Build GRCh38
rs781975778 Chr.17: 41973843-41973843 on Build GRCh38
rs782661998 Chr.17: 41973829-41973829 on Build GRCh38
rs374583295 Chr.17: 41973808-41973808 on Build GRCh38
rs201242451 Chr.17: 41973834-41973834 on Build GRCh38
rs2070106 Chr.17: 41973846-41973846 on Build GRCh38
rs781883394 Chr.17: 41973881-41973881 on Build GRCh38
rs1016790393 Chr.17: 41973805-41973805 on Build GRCh38
rs1052794149 Chr.17: 41973903-41973903 on Build GRCh38
rs889680920 Chr.17: 41973972-41973972 on Build GRCh38
rs370234532 Chr.17: 41973849-41973849 on Build GRCh38
rs782096149 Chr.17: 41973875-41973875 on Build GRCh38
rs776942033 Chr.17: 41973948-41973948 on Build GRCh38
rs34301655 Chr.17: 41973897-41973897 on Build GRCh38
rs782443159 Chr.17: 41973973-41973973 on Build GRCh38
rs782233811 Chr.17: 41973888-41973888 on Build GRCh38
rs782811736 Chr.17: 41973871-41973871 on Build GRCh38
rs1043847540 Chr.17: 41973894-41973894 on Build GRCh38
rs782033287 Chr.17: 41973937-41973937 on Build GRCh38
rs2229932 Chr.17: 41973951-41973951 on Build GRCh38
rs782736241 Chr.17: 41973925-41973925 on Build GRCh38
rs782378608 Chr.17: 41973842-41973842 on Build GRCh38
rs782498838 Chr.17: 41973960-41973960 on Build GRCh38
rs782657028 Chr.17: 41973793-41973793 on Build GRCh38
rs782587919 Chr.17: 41973814-41973814 on Build GRCh38

View Full Product Details