Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 73758871-73759096 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD109
Entrez Gene ID
135228
Gene Name
CD109 molecule
Gene Aliases
CPAMD7, p180, r150
UniGene
Hs.399891
Gene Chromosome Location
Chr.6: 73679192-73828317 on Build GRCh38
Transcripts
NM_001159587.2 NM_001159588.2 NM_133493.4 XM_005248659.3 XM_011535473.1 XM_017010283.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6504451 c.685T>C p.F229L Chr.6: 73758955-73758955 on Build GRCh38
1445864 c.752C>T p.T251M Chr.6: 73759022-73759022 on Build GRCh38
1162327 c.735T>C p.H245H Chr.6: 73759005-73759005 on Build GRCh38
7157431 c.738A>G p.L246L Chr.6: 73759008-73759008 on Build GRCh38
6234666 c.758+7G>T p.? Chr.6: 73759035-73759035 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs944064070 Chr.6: 73758952-73758952 on Build GRCh38
rs376967090 Chr.6: 73759026-73759026 on Build GRCh38
rs757459134 Chr.6: 73759031-73759031 on Build GRCh38
rs1032186191 Chr.6: 73758949-73758949 on Build GRCh38
rs908625669 Chr.6: 73758934-73758934 on Build GRCh38
rs746778680 Chr.6: 73759044-73759044 on Build GRCh38
rs762625249 Chr.6: 73758991-73758991 on Build GRCh38
rs769620718 Chr.6: 73759053-73759053 on Build GRCh38
rs766199209 Chr.6: 73759001-73759001 on Build GRCh38
rs143388686 Chr.6: 73758900-73758900 on Build GRCh38
rs755441690 Chr.6: 73759012-73759012 on Build GRCh38
rs141066052 Chr.6: 73758953-73758953 on Build GRCh38
rs779158159 Chr.6: 73759034-73759034 on Build GRCh38
rs41265527 Chr.6: 73759050-73759050 on Build GRCh38
rs764752232 Chr.6: 73758987-73758987 on Build GRCh38
rs36019774 Chr.6: 73758963-73758964 on Build GRCh38
rs751957646 Chr.6: 73759010-73759010 on Build GRCh38
rs748154629 Chr.6: 73758905-73758905 on Build GRCh38
rs746868295 Chr.6: 73758971-73758971 on Build GRCh38
rs770209787 Chr.6: 73759036-73759036 on Build GRCh38
rs371377745 Chr.6: 73758974-73758974 on Build GRCh38
rs775263523 Chr.6: 73758948-73758948 on Build GRCh38
rs767880549 Chr.6: 73759016-73759016 on Build GRCh38
rs373548964 Chr.6: 73758982-73758982 on Build GRCh38
rs749802252 Chr.6: 73758966-73758967 on Build GRCh38
rs772607561 Chr.6: 73758989-73758989 on Build GRCh38
rs199660260 Chr.6: 73759022-73759022 on Build GRCh38
rs910779180 Chr.6: 73759068-73759068 on Build GRCh38
rs903553438 Chr.6: 73759063-73759063 on Build GRCh38
rs777843417 Chr.6: 73759024-73759024 on Build GRCh38
rs761682929 Chr.6: 73758983-73758983 on Build GRCh38
rs769939827 Chr.6: 73758909-73758910 on Build GRCh38
rs777738448 Chr.6: 73758917-73758917 on Build GRCh38
rs373245474 Chr.6: 73759054-73759054 on Build GRCh38
rs749471297 Chr.6: 73758946-73758946 on Build GRCh38
rs748057531 Chr.6: 73759052-73759052 on Build GRCh38
rs756592994 Chr.6: 73759023-73759023 on Build GRCh38
rs768396255 Chr.6: 73759049-73759049 on Build GRCh38
rs931317273 Chr.6: 73758901-73758901 on Build GRCh38
rs775395134 Chr.6: 73758911-73758911 on Build GRCh38

View Full Product Details