Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 245155294-245155537 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF26B
Entrez Gene ID
55083
Gene Name
kinesin family member 26B
Gene Aliases
-
UniGene
Hs.143134
Gene Chromosome Location
Chr.1: 245154985-245703126 on Build GRCh38
Transcripts
NM_018012.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5820572 c.63+3A>G p.? Chr.1: 245155490-245155490 on Build GRCh38
6460296 c.46A>G p.R16G Chr.1: 245155470-245155470 on Build GRCh38
6257267 c.3G>T p.M1I Chr.1: 245155427-245155427 on Build GRCh38
1662486 c.26A>T p.E9V Chr.1: 245155450-245155450 on Build GRCh38
4165117 c.23A>C p.K8T Chr.1: 245155447-245155447 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs772411690 Chr.1: 245155387-245155387 on Build GRCh38
rs373174058 Chr.1: 245155370-245155370 on Build GRCh38
rs898677524 Chr.1: 245155342-245155342 on Build GRCh38
rs532381084 Chr.1: 245155338-245155338 on Build GRCh38
rs969420572 Chr.1: 245155423-245155423 on Build GRCh38
rs755441267 Chr.1: 245155459-245155459 on Build GRCh38
rs567828648 Chr.1: 245155395-245155395 on Build GRCh38
rs12409851 Chr.1: 245155386-245155386 on Build GRCh38
rs756761116 Chr.1: 245155433-245155433 on Build GRCh38
rs748567281 Chr.1: 245155467-245155467 on Build GRCh38
rs530762023 Chr.1: 245155369-245155369 on Build GRCh38
rs1029831798 Chr.1: 245155489-245155489 on Build GRCh38
rs377742075 Chr.1: 245155518-245155518 on Build GRCh38
rs377067786 Chr.1: 245155412-245155412 on Build GRCh38
rs749896175 Chr.1: 245155516-245155516 on Build GRCh38
rs761596817 Chr.1: 245155504-245155504 on Build GRCh38
rs760443089 Chr.1: 245155500-245155500 on Build GRCh38
rs747382313 Chr.1: 245155376-245155376 on Build GRCh38
rs560413028 Chr.1: 245155337-245155337 on Build GRCh38
rs773112020 Chr.1: 245155473-245155473 on Build GRCh38
rs760531064 Chr.1: 245155400-245155400 on Build GRCh38
rs774614684 Chr.1: 245155499-245155499 on Build GRCh38
rs1034496862 Chr.1: 245155402-245155402 on Build GRCh38
rs780280038 Chr.1: 245155503-245155503 on Build GRCh38
rs779916680 Chr.1: 245155451-245155451 on Build GRCh38
rs773376327 Chr.1: 245155389-245155389 on Build GRCh38
rs761927732 Chr.1: 245155383-245155383 on Build GRCh38
rs747360254 Chr.1: 245155458-245155458 on Build GRCh38
rs370230230 Chr.1: 245155441-245155441 on Build GRCh38
rs374748538 Chr.1: 245155517-245155517 on Build GRCh38
rs369604504 Chr.1: 245155394-245155394 on Build GRCh38
rs769277526 Chr.1: 245155378-245155378 on Build GRCh38
rs763918100 Chr.1: 245155502-245155502 on Build GRCh38
rs978712912 Chr.1: 245155469-245155469 on Build GRCh38
rs867476545 Chr.1: 245155349-245155349 on Build GRCh38
rs770358602 Chr.1: 245155468-245155468 on Build GRCh38
rs372845660 Chr.1: 245155408-245155408 on Build GRCh38
rs967662718 Chr.1: 245155455-245155456 on Build GRCh38
rs757922173 Chr.1: 245155444-245155444 on Build GRCh38
rs749393962 Chr.1: 245155487-245155487 on Build GRCh38
rs764602468 Chr.1: 245155440-245155440 on Build GRCh38
rs776115981 Chr.1: 245155404-245155405 on Build GRCh38
rs185652778 Chr.1: 245155345-245155345 on Build GRCh38
rs199501502 Chr.1: 245155496-245155496 on Build GRCh38
rs571814392 Chr.1: 245155352-245155352 on Build GRCh38
rs769801247 Chr.1: 245155388-245155388 on Build GRCh38
rs536972752 Chr.1: 245155460-245155460 on Build GRCh38
rs777330878 Chr.1: 245155382-245155382 on Build GRCh38
rs540020834 Chr.1: 245155333-245155333 on Build GRCh38

View Full Product Details