Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 138182417-138182676 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF20A
Entrez Gene ID
10112
Gene Name
kinesin family member 20A
Gene Aliases
MKLP2, RAB6KIFL
UniGene
Hs.718626
Gene Chromosome Location
Chr.5: 138178728-138187715 on Build GRCh38
Transcripts
NM_005733.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5346428 c.538C>T p.L180F Chr.5: 138182609-138182609 on Build GRCh38
3826832 c.491C>G p.S164* Chr.5: 138182438-138182438 on Build GRCh38
1061250 c.507G>A p.T169T Chr.5: 138182454-138182454 on Build GRCh38
6695005 c.506C>T p.T169M Chr.5: 138182453-138182453 on Build GRCh38
6440467 c.530G>A p.G177E Chr.5: 138182601-138182601 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371671153 Chr.5: 138182626-138182626 on Build GRCh38
rs942270617 Chr.5: 138182637-138182637 on Build GRCh38
rs773589238 Chr.5: 138182478-138182478 on Build GRCh38
rs771538037 Chr.5: 138182536-138182537 on Build GRCh38
rs772349108 Chr.5: 138182471-138182471 on Build GRCh38
rs974579974 Chr.5: 138182562-138182562 on Build GRCh38
rs185063701 Chr.5: 138182480-138182480 on Build GRCh38
rs756990948 Chr.5: 138182565-138182565 on Build GRCh38
rs759613418 Chr.5: 138182508-138182508 on Build GRCh38
rs148216162 Chr.5: 138182616-138182616 on Build GRCh38
rs759703354 Chr.5: 138182622-138182622 on Build GRCh38
rs748489732 Chr.5: 138182519-138182519 on Build GRCh38
rs751420729 Chr.5: 138182564-138182564 on Build GRCh38
rs758291436 Chr.5: 138182546-138182546 on Build GRCh38
rs780966779 Chr.5: 138182579-138182579 on Build GRCh38
rs200026092 Chr.5: 138182612-138182612 on Build GRCh38
rs374271585 Chr.5: 138182625-138182625 on Build GRCh38
rs761024664 Chr.5: 138182510-138182511 on Build GRCh38
rs141777523 Chr.5: 138182643-138182643 on Build GRCh38
rs747521180 Chr.5: 138182516-138182516 on Build GRCh38
rs771130761 Chr.5: 138182615-138182615 on Build GRCh38
rs370724756 Chr.5: 138182481-138182481 on Build GRCh38
rs748700722 Chr.5: 138182606-138182606 on Build GRCh38
rs761612274 Chr.5: 138182591-138182591 on Build GRCh38
rs374601490 Chr.5: 138182511-138182511 on Build GRCh38
rs765241898 Chr.5: 138182510-138182510 on Build GRCh38
rs954833139 Chr.5: 138182603-138182603 on Build GRCh38
rs779665695 Chr.5: 138182604-138182604 on Build GRCh38
rs559088548 Chr.5: 138182558-138182558 on Build GRCh38
rs200872203 Chr.5: 138182453-138182453 on Build GRCh38
rs773574505 Chr.5: 138182611-138182611 on Build GRCh38
rs886785789 Chr.5: 138182538-138182538 on Build GRCh38
rs900917059 Chr.5: 138182496-138182496 on Build GRCh38
rs375606679 Chr.5: 138182454-138182454 on Build GRCh38
rs367756689 Chr.5: 138182599-138182599 on Build GRCh38
rs762354018 Chr.5: 138182455-138182455 on Build GRCh38
rs753999115 Chr.5: 138182492-138182492 on Build GRCh38
rs768827865 Chr.5: 138182550-138182550 on Build GRCh38
rs200853662 Chr.5: 138182640-138182640 on Build GRCh38
rs374761244 Chr.5: 138182649-138182649 on Build GRCh38
rs933766516 Chr.5: 138182504-138182504 on Build GRCh38
rs772680195 Chr.5: 138182609-138182609 on Build GRCh38
rs769445007 Chr.5: 138182602-138182602 on Build GRCh38

View Full Product Details