Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 112087376-112087601 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM229B
Entrez Gene ID
619208
Gene Name
family with sequence similarity 229 member B
Gene Aliases
C6orf225
UniGene
Hs.591340
Gene Chromosome Location
Chr.6: 112087471-112102790 on Build GRCh38
Transcripts
NM_001033564.2 NM_016262.4 XM_011535875.2 XM_017010918.1

Gene Symbol
TUBE1
Entrez Gene ID
51175
Gene Name
tubulin epsilon 1
Gene Aliases
TUBE, dJ142L7.2
UniGene
Hs.34851
Gene Chromosome Location
Chr.6: 112070657-112087548 on Build GRCh38
Transcripts
NM_001033564.2 NM_016262.4 XM_011535875.2 XM_017010918.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7210699 c.3G>A p.M1I Chr.6: 112087432-112087432 on Build GRCh38
7240579 c.25+3A>G p.? Chr.6: 112087407-112087407 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs782014165 Chr.6: 112087424-112087424 on Build GRCh38
rs199513778 Chr.6: 112087425-112087425 on Build GRCh38
rs894623940 Chr.6: 112087470-112087470 on Build GRCh38
rs202220859 Chr.6: 112087474-112087474 on Build GRCh38
rs1031653110 Chr.6: 112087552-112087552 on Build GRCh38
rs1057243564 Chr.6: 112087457-112087457 on Build GRCh38
rs1013205132 Chr.6: 112087490-112087490 on Build GRCh38
rs587623185 Chr.6: 112087449-112087449 on Build GRCh38
rs782485988 Chr.6: 112087475-112087475 on Build GRCh38
rs759580602 Chr.6: 112087510-112087510 on Build GRCh38
rs6568714 Chr.6: 112087407-112087407 on Build GRCh38
rs782704182 Chr.6: 112087434-112087434 on Build GRCh38
rs587594929 Chr.6: 112087499-112087499 on Build GRCh38
rs771138356 Chr.6: 112087454-112087454 on Build GRCh38
rs782103653 Chr.6: 112087417-112087417 on Build GRCh38
rs768611246 Chr.6: 112087551-112087551 on Build GRCh38
rs781862050 Chr.6: 112087532-112087532 on Build GRCh38
rs782670287 Chr.6: 112087479-112087479 on Build GRCh38
rs904737943 Chr.6: 112087505-112087505 on Build GRCh38
rs147330886 Chr.6: 112087436-112087436 on Build GRCh38
rs200357896 Chr.6: 112087423-112087423 on Build GRCh38
rs781862571 Chr.6: 112087460-112087460 on Build GRCh38
rs1046680469 Chr.6: 112087437-112087437 on Build GRCh38
rs587642779 Chr.6: 112087406-112087406 on Build GRCh38
rs182369129 Chr.6: 112087456-112087456 on Build GRCh38
rs986649833 Chr.6: 112087396-112087396 on Build GRCh38

View Full Product Details