Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 16500637-16500878 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf44
Entrez Gene ID
84167
Gene Name
chromosome 19 open reading frame 44
Gene Aliases
-
UniGene
Hs.631627
Gene Chromosome Location
Chr.19: 16496311-16521369 on Build GRCh38
Transcripts
NM_001288834.1 NM_032207.3 XM_006722920.3 XM_011528354.2 XM_017027350.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
711360 c.69G>T p.L23F Chr.19: 16500861-16500861 on Build GRCh38
3530027 c.65C>T p.S22F Chr.19: 16500857-16500857 on Build GRCh38
3937916 c.34C>T p.R12C Chr.19: 16500826-16500826 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs769541837 Chr.19: 16500700-16500700 on Build GRCh38
rs759197090 Chr.19: 16500827-16500827 on Build GRCh38
rs776435313 Chr.19: 16500826-16500826 on Build GRCh38
rs370638399 Chr.19: 16500750-16500750 on Build GRCh38
rs377450443 Chr.19: 16500779-16500779 on Build GRCh38
rs769935343 Chr.19: 16500794-16500794 on Build GRCh38
rs767850185 Chr.19: 16500825-16500825 on Build GRCh38
rs892767341 Chr.19: 16500667-16500667 on Build GRCh38
rs566396315 Chr.19: 16500693-16500693 on Build GRCh38
rs779934479 Chr.19: 16500796-16500796 on Build GRCh38
rs764833711 Chr.19: 16500831-16500831 on Build GRCh38
rs762331456 Chr.19: 16500818-16500818 on Build GRCh38
rs62118143 Chr.19: 16500716-16500716 on Build GRCh38
rs757016013 Chr.19: 16500761-16500761 on Build GRCh38
rs774396285 Chr.19: 16500832-16500833 on Build GRCh38
rs143291289 Chr.19: 16500802-16500802 on Build GRCh38
rs746079130 Chr.19: 16500788-16500788 on Build GRCh38
rs902912535 Chr.19: 16500704-16500704 on Build GRCh38
rs958545949 Chr.19: 16500769-16500769 on Build GRCh38
rs975456923 Chr.19: 16500688-16500688 on Build GRCh38
rs1041420473 Chr.19: 16500703-16500703 on Build GRCh38
rs201897764 Chr.19: 16500801-16500801 on Build GRCh38
rs373189022 Chr.19: 16500756-16500756 on Build GRCh38
rs758229082 Chr.19: 16500742-16500742 on Build GRCh38
rs768766721 Chr.19: 16500797-16500798 on Build GRCh38
rs963325764 Chr.19: 16500813-16500813 on Build GRCh38
rs752101001 Chr.19: 16500841-16500841 on Build GRCh38
rs1051061120 Chr.19: 16500724-16500724 on Build GRCh38
rs774965695 Chr.19: 16500817-16500817 on Build GRCh38

View Full Product Details