Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 41921502-41921773 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BYSL
Entrez Gene ID
705
Gene Name
bystin like
Gene Aliases
BYSTIN
UniGene
Hs.106880
Gene Chromosome Location
Chr.6: 41921227-41933046 on Build GRCh38
Transcripts
NM_004053.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6284608 c.115G>T p.G39C Chr.6: 41921677-41921677 on Build GRCh38
6252590 c.139G>T p.E47* Chr.6: 41921701-41921701 on Build GRCh38
4951610 c.178C>A p.Q60K Chr.6: 41921740-41921740 on Build GRCh38
4940091 c.14A>T p.K5M Chr.6: 41921576-41921576 on Build GRCh38
4816346 c.61G>A p.D21N Chr.6: 41921623-41921623 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1020607730 Chr.6: 41921652-41921652 on Build GRCh38
rs866195599 Chr.6: 41921751-41921751 on Build GRCh38
rs147171521 Chr.6: 41921597-41921597 on Build GRCh38
rs747604397 Chr.6: 41921657-41921657 on Build GRCh38
rs202135065 Chr.6: 41921543-41921543 on Build GRCh38
rs563253509 Chr.6: 41921749-41921749 on Build GRCh38
rs758484450 Chr.6: 41921555-41921555 on Build GRCh38
rs754927384 Chr.6: 41921720-41921720 on Build GRCh38
rs768665277 Chr.6: 41921592-41921592 on Build GRCh38
rs751706469 Chr.6: 41921683-41921683 on Build GRCh38
rs759887573 Chr.6: 41921595-41921595 on Build GRCh38
rs143551764 Chr.6: 41921585-41921585 on Build GRCh38
rs556154714 Chr.6: 41921538-41921538 on Build GRCh38
rs761419679 Chr.6: 41921589-41921589 on Build GRCh38
rs745659269 Chr.6: 41921631-41921631 on Build GRCh38
rs751649565 Chr.6: 41921606-41921606 on Build GRCh38
rs747616822 Chr.6: 41921729-41921729 on Build GRCh38
rs376067308 Chr.6: 41921656-41921656 on Build GRCh38
rs772851955 Chr.6: 41921662-41921662 on Build GRCh38
rs747214685 Chr.6: 41921570-41921570 on Build GRCh38
rs779791479 Chr.6: 41921719-41921719 on Build GRCh38
rs757443744 Chr.6: 41921609-41921609 on Build GRCh38
rs769293717 Chr.6: 41921748-41921748 on Build GRCh38
rs776021157 Chr.6: 41921741-41921741 on Build GRCh38
rs761890603 Chr.6: 41921697-41921697 on Build GRCh38
rs3763274 Chr.6: 41921529-41921529 on Build GRCh38
rs138882886 Chr.6: 41921574-41921574 on Build GRCh38
rs768138610 Chr.6: 41921587-41921587 on Build GRCh38
rs776182011 Chr.6: 41921675-41921675 on Build GRCh38
rs763585422 Chr.6: 41921679-41921679 on Build GRCh38
rs750647786 Chr.6: 41921717-41921717 on Build GRCh38
rs945932099 Chr.6: 41921688-41921688 on Build GRCh38
rs769636909 Chr.6: 41921584-41921584 on Build GRCh38
rs761606399 Chr.6: 41921664-41921688 on Build GRCh38
rs755935646 Chr.6: 41921634-41921634 on Build GRCh38
rs935788463 Chr.6: 41921701-41921701 on Build GRCh38
rs377537689 Chr.6: 41921598-41921598 on Build GRCh38
rs748867329 Chr.6: 41921586-41921586 on Build GRCh38
rs545082479 Chr.6: 41921544-41921544 on Build GRCh38
rs61737728 Chr.6: 41921542-41921542 on Build GRCh38
rs200314949 Chr.6: 41921645-41921645 on Build GRCh38
rs374431794 Chr.6: 41921732-41921732 on Build GRCh38
rs763354121 Chr.6: 41921747-41921747 on Build GRCh38
rs774059290 Chr.6: 41921549-41921549 on Build GRCh38
rs773992123 Chr.6: 41921527-41921527 on Build GRCh38
rs3763273 Chr.6: 41921671-41921671 on Build GRCh38
rs767154156 Chr.6: 41921593-41921593 on Build GRCh38
rs766628254 Chr.6: 41921533-41921533 on Build GRCh38
rs559049242 Chr.6: 41921548-41921548 on Build GRCh38
rs778763224 Chr.6: 41921725-41921725 on Build GRCh38
rs1048420164 Chr.6: 41921534-41921534 on Build GRCh38
rs560983008 Chr.6: 41921659-41921659 on Build GRCh38
rs768192134 Chr.6: 41921646-41921646 on Build GRCh38
rs764648487 Chr.6: 41921680-41921680 on Build GRCh38
rs745631183 Chr.6: 41921580-41921580 on Build GRCh38
rs780880767 Chr.6: 41921577-41921577 on Build GRCh38
rs951489652 Chr.6: 41921525-41921525 on Build GRCh38
rs368935062 Chr.6: 41921636-41921636 on Build GRCh38
rs752836269 Chr.6: 41921552-41921552 on Build GRCh38
rs764240053 Chr.6: 41921602-41921602 on Build GRCh38
rs140361205 Chr.6: 41921715-41921715 on Build GRCh38
rs755907731 Chr.6: 41921718-41921718 on Build GRCh38
rs1034801599 Chr.6: 41921713-41921713 on Build GRCh38
rs779083404 Chr.6: 41921739-41921739 on Build GRCh38
rs777346715 Chr.6: 41921733-41921733 on Build GRCh38
rs774063141 Chr.6: 41921591-41921592 on Build GRCh38
rs904154825 Chr.6: 41921698-41921698 on Build GRCh38
rs746643048 Chr.6: 41921734-41921734 on Build GRCh38
rs781422076 Chr.6: 41921613-41921613 on Build GRCh38

View Full Product Details