Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 97582170-97582417 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD2
Entrez Gene ID
26287
Gene Name
ankyrin repeat domain 2
Gene Aliases
ARPP
UniGene
Hs.73708
Gene Chromosome Location
Chr.10: 97572441-97583884 on Build GRCh38
Transcripts
NM_001129981.2 NM_001291218.1 NM_001291219.1 NM_020349.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6225081 c.757G>A p.V253M Chr.10: 97582336-97582336 on Build GRCh38
7413800 c.812T>C p.L271P Chr.10: 97582391-97582391 on Build GRCh38
6208008 c.749C>T p.P250L Chr.10: 97582328-97582328 on Build GRCh38
4919991 c.769A>G p.T257A Chr.10: 97582348-97582348 on Build GRCh38
5453788 c.793C>T p.H265Y Chr.10: 97582372-97582372 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs538362754 Chr.10: 97582314-97582315 on Build GRCh38
rs747686350 Chr.10: 97582272-97582272 on Build GRCh38
rs760624001 Chr.10: 97582393-97582393 on Build GRCh38
rs371824013 Chr.10: 97582351-97582351 on Build GRCh38
rs867256679 Chr.10: 97582360-97582360 on Build GRCh38
rs537471247 Chr.10: 97582310-97582310 on Build GRCh38
rs185893932 Chr.10: 97582311-97582311 on Build GRCh38
rs988911251 Chr.10: 97582230-97582230 on Build GRCh38
rs778346703 Chr.10: 97582287-97582287 on Build GRCh38
rs892728433 Chr.10: 97582386-97582386 on Build GRCh38
rs908474037 Chr.10: 97582251-97582251 on Build GRCh38
rs926450988 Chr.10: 97582199-97582199 on Build GRCh38
rs367694142 Chr.10: 97582282-97582282 on Build GRCh38
rs145093754 Chr.10: 97582328-97582328 on Build GRCh38
rs766622514 Chr.10: 97582333-97582333 on Build GRCh38
rs1050532043 Chr.10: 97582303-97582303 on Build GRCh38
rs774616303 Chr.10: 97582336-97582336 on Build GRCh38
rs1047478852 Chr.10: 97582340-97582340 on Build GRCh38
rs759669168 Chr.10: 97582346-97582346 on Build GRCh38
rs930366826 Chr.10: 97582339-97582339 on Build GRCh38
rs375926537 Chr.10: 97582281-97582281 on Build GRCh38
rs745348892 Chr.10: 97582297-97582297 on Build GRCh38
rs757564823 Chr.10: 97582365-97582366 on Build GRCh38
rs940658328 Chr.10: 97582256-97582256 on Build GRCh38
rs770055356 Chr.10: 97582317-97582317 on Build GRCh38
rs201541205 Chr.10: 97582325-97582325 on Build GRCh38
rs376525356 Chr.10: 97582382-97582382 on Build GRCh38
rs534926101 Chr.10: 97582348-97582348 on Build GRCh38
rs960600526 Chr.10: 97582246-97582246 on Build GRCh38
rs957707895 Chr.10: 97582224-97582224 on Build GRCh38
rs769200028 Chr.10: 97582309-97582309 on Build GRCh38
rs551480864 Chr.10: 97582260-97582260 on Build GRCh38
rs992466480 Chr.10: 97582247-97582247 on Build GRCh38

View Full Product Details