Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 102075922-102076175 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf70
Entrez Gene ID
85016
Gene Name
chromosome 11 open reading frame 70
Gene Aliases
-
UniGene
Hs.98328
Gene Chromosome Location
Chr.11: 102047409-102084560 on Build GRCh38
Transcripts
NM_001195005.1 NM_032930.2 XM_005271713.3 XM_017018454.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6614101 c.517C>T p.H173Y Chr.11: 102075954-102075954 on Build GRCh38
6348179 c.493G>C p.E165Q Chr.11: 102075930-102075930 on Build GRCh38
4998426 c.586C>A p.L196I Chr.11: 102076023-102076023 on Build GRCh38
6067027 c.493G>T p.E165* Chr.11: 102075930-102075930 on Build GRCh38
685808 c.484G>T p.D162Y Chr.11: 102076035-102076035 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773698817 Chr.11: 102076069-102076069 on Build GRCh38
rs372015461 Chr.11: 102076081-102076081 on Build GRCh38
rs775160676 Chr.11: 102076089-102076089 on Build GRCh38
rs770229788 Chr.11: 102075958-102075958 on Build GRCh38
rs767227461 Chr.11: 102075983-102075983 on Build GRCh38
rs866788183 Chr.11: 102075990-102075990 on Build GRCh38
rs376842255 Chr.11: 102075965-102075965 on Build GRCh38
rs145316880 Chr.11: 102075991-102075991 on Build GRCh38
rs1040011315 Chr.11: 102076126-102076126 on Build GRCh38
rs915184576 Chr.11: 102076020-102076020 on Build GRCh38
rs758501351 Chr.11: 102076030-102076030 on Build GRCh38
rs571495259 Chr.11: 102076095-102076095 on Build GRCh38
rs759047947 Chr.11: 102075979-102075979 on Build GRCh38
rs563349596 Chr.11: 102075992-102075992 on Build GRCh38
rs755462053 Chr.11: 102076041-102076041 on Build GRCh38
rs556290819 Chr.11: 102076146-102076146 on Build GRCh38
rs944462401 Chr.11: 102076125-102076125 on Build GRCh38
rs529008192 Chr.11: 102076004-102076004 on Build GRCh38
rs747400996 Chr.11: 102075957-102075957 on Build GRCh38
rs770407389 Chr.11: 102075967-102075967 on Build GRCh38
rs750519685 Chr.11: 102076028-102076028 on Build GRCh38
rs777318658 Chr.11: 102075961-102075961 on Build GRCh38
rs773930289 Chr.11: 102075975-102075975 on Build GRCh38
rs371474452 Chr.11: 102076080-102076080 on Build GRCh38
rs763985305 Chr.11: 102076017-102076017 on Build GRCh38
rs771830038 Chr.11: 102075955-102075955 on Build GRCh38
rs1014240853 Chr.11: 102076129-102076129 on Build GRCh38
rs748610887 Chr.11: 102076056-102076056 on Build GRCh38
rs923646456 Chr.11: 102076083-102076087 on Build GRCh38
rs376333708 Chr.11: 102076022-102076022 on Build GRCh38
rs192420752 Chr.11: 102076107-102076107 on Build GRCh38
rs770342377 Chr.11: 102076057-102076057 on Build GRCh38
rs151324884 Chr.11: 102076034-102076034 on Build GRCh38
rs937913571 Chr.11: 102075999-102075999 on Build GRCh38
rs752435466 Chr.11: 102075993-102075993 on Build GRCh38
rs927291742 Chr.11: 102075962-102075962 on Build GRCh38
rs549559265 Chr.11: 102075959-102075959 on Build GRCh38
rs781762004 Chr.11: 102076052-102076052 on Build GRCh38

View Full Product Details