Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 77907738-77907971 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCK1
Entrez Gene ID
57143
Gene Name
aarF domain containing kinase 1
Gene Aliases
-
UniGene
Hs.413208
Gene Chromosome Location
Chr.14: 77800058-77933954 on Build GRCh38
Transcripts
NM_001142545.1 NM_020421.3 XM_005267885.3 XM_005267886.3 XM_011536985.1 XM_017021474.1 XM_017021475.1 XM_017021476.1 XM_017021477.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4508660 c.579C>T p.L193L Chr.14: 77907844-77907844 on Build GRCh38
6194877 c.824A>G p.D275G Chr.14: 77907885-77907885 on Build GRCh38
958232 c.545G>A p.R182Q Chr.14: 77907810-77907810 on Build GRCh38
5478771 c.643G>A p.D215N Chr.14: 77907908-77907908 on Build GRCh38
5821730 c.538-2A>T p.? Chr.14: 77907801-77907801 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749467779 Chr.14: 77907907-77907907 on Build GRCh38
rs764146384 Chr.14: 77907790-77907790 on Build GRCh38
rs780382366 Chr.14: 77907900-77907900 on Build GRCh38
rs780353968 Chr.14: 77907881-77907882 on Build GRCh38
rs767998199 Chr.14: 77907764-77907764 on Build GRCh38
rs200683533 Chr.14: 77907823-77907823 on Build GRCh38
rs969950590 Chr.14: 77907768-77907768 on Build GRCh38
rs751026962 Chr.14: 77907872-77907872 on Build GRCh38
rs17106595 Chr.14: 77907949-77907949 on Build GRCh38
rs200494082 Chr.14: 77907858-77907858 on Build GRCh38
rs764940937 Chr.14: 77907772-77907772 on Build GRCh38
rs115320309 Chr.14: 77907771-77907771 on Build GRCh38
rs34272020 Chr.14: 77907829-77907829 on Build GRCh38
rs149126432 Chr.14: 77907910-77907910 on Build GRCh38
rs55934334 Chr.14: 77907836-77907836 on Build GRCh38
rs372050102 Chr.14: 77907866-77907866 on Build GRCh38
rs374530596 Chr.14: 77907890-77907890 on Build GRCh38
rs905751375 Chr.14: 77907760-77907760 on Build GRCh38
rs752289467 Chr.14: 77907777-77907777 on Build GRCh38
rs373019674 Chr.14: 77907950-77907950 on Build GRCh38
rs758565964 Chr.14: 77907782-77907783 on Build GRCh38
rs767404674 Chr.14: 77907877-77907877 on Build GRCh38
rs764987326 Chr.14: 77907859-77907859 on Build GRCh38
rs200132502 Chr.14: 77907873-77907873 on Build GRCh38
rs145193842 Chr.14: 77907865-77907865 on Build GRCh38
rs773741399 Chr.14: 77907837-77907837 on Build GRCh38
rs377002610 Chr.14: 77907770-77907770 on Build GRCh38
rs772743037 Chr.14: 77907941-77907941 on Build GRCh38
rs773829514 Chr.14: 77907945-77907945 on Build GRCh38
rs757124062 Chr.14: 77907809-77907809 on Build GRCh38
rs774421723 Chr.14: 77907835-77907835 on Build GRCh38
rs774699503 Chr.14: 77907761-77907761 on Build GRCh38
rs1029255623 Chr.14: 77907871-77907871 on Build GRCh38
rs778880011 Chr.14: 77907908-77907908 on Build GRCh38
rs749434899 Chr.14: 77907831-77907831 on Build GRCh38
rs140603078 Chr.14: 77907810-77907810 on Build GRCh38
rs757820434 Chr.14: 77907785-77907785 on Build GRCh38
rs201937466 Chr.14: 77907879-77907879 on Build GRCh38
rs895920350 Chr.14: 77907786-77907786 on Build GRCh38
rs768606266 Chr.14: 77907832-77907832 on Build GRCh38
rs751452010 Chr.14: 77907796-77907796 on Build GRCh38
rs200934423 Chr.14: 77907911-77907911 on Build GRCh38
rs779716231 Chr.14: 77907913-77907913 on Build GRCh38
rs745616076 Chr.14: 77907816-77907816 on Build GRCh38

View Full Product Details