Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28758860-28759099 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM222B
Entrez Gene ID
55731
Gene Name
family with sequence similarity 222 member B
Gene Aliases
C17orf63
UniGene
Hs.564533
Gene Chromosome Location
Chr.17: 28755978-28855036 on Build GRCh38
Transcripts
NM_001077498.2 NM_001288631.1 NM_001288632.1 NM_001288633.1 NM_001288634.1 NM_001288635.1 NM_001288636.1 NM_001288637.1 NM_001288638.1 NM_001288639.1 NM_001288640.1 NM_018182.3 XM_011524986.2 XM_011524987.1 XM_011524988.2 XM_017024831.1 XM_017024832.1 XM_017024833.1 XM_017024834.1 XM_017024835.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4682953 c.989C>T p.T330I Chr.17: 28758970-28758970 on Build GRCh38
4801319 c.972C>T p.V324V Chr.17: 28758987-28758987 on Build GRCh38
4682952 c.989C>T p.T330I Chr.17: 28758970-28758970 on Build GRCh38
4652836 c.1044C>T p.G348G Chr.17: 28758915-28758915 on Build GRCh38
2800994 c.891C>T p.P297P Chr.17: 28759068-28759068 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766200026 Chr.17: 28759050-28759050 on Build GRCh38
rs375208934 Chr.17: 28758987-28758987 on Build GRCh38
rs775091691 Chr.17: 28759043-28759043 on Build GRCh38
rs199717133 Chr.17: 28758980-28758980 on Build GRCh38
rs200350361 Chr.17: 28758952-28758952 on Build GRCh38
rs200444762 Chr.17: 28758953-28758953 on Build GRCh38
rs201471823 Chr.17: 28759061-28759061 on Build GRCh38
rs773766356 Chr.17: 28758964-28758964 on Build GRCh38
rs201240329 Chr.17: 28759019-28759019 on Build GRCh38
rs755105411 Chr.17: 28758949-28758949 on Build GRCh38
rs554449532 Chr.17: 28758957-28758957 on Build GRCh38
rs898139397 Chr.17: 28758926-28758926 on Build GRCh38
rs765389691 Chr.17: 28759069-28759069 on Build GRCh38
rs760436935 Chr.17: 28759044-28759044 on Build GRCh38
rs752755670 Chr.17: 28759070-28759070 on Build GRCh38
rs34289421 Chr.17: 28758958-28758958 on Build GRCh38
rs904373574 Chr.17: 28758932-28758932 on Build GRCh38
rs748197585 Chr.17: 28759008-28759008 on Build GRCh38
rs376794567 Chr.17: 28758951-28758951 on Build GRCh38
rs764115309 Chr.17: 28758907-28758907 on Build GRCh38
rs755017318 Chr.17: 28759002-28759002 on Build GRCh38
rs372959774 Chr.17: 28759027-28759027 on Build GRCh38
rs749362624 Chr.17: 28758948-28758948 on Build GRCh38
rs767489400 Chr.17: 28759034-28759034 on Build GRCh38
rs776112666 Chr.17: 28758890-28758890 on Build GRCh38
rs758472939 Chr.17: 28759074-28759074 on Build GRCh38
rs747372365 Chr.17: 28759017-28759017 on Build GRCh38
rs112187222 Chr.17: 28759016-28759016 on Build GRCh38
rs759188442 Chr.17: 28758891-28758891 on Build GRCh38
rs779007369 Chr.17: 28759004-28759004 on Build GRCh38
rs773856870 Chr.17: 28759031-28759031 on Build GRCh38
rs534717471 Chr.17: 28758968-28758968 on Build GRCh38
rs750212243 Chr.17: 28758910-28758910 on Build GRCh38
rs1022923568 Chr.17: 28758969-28758969 on Build GRCh38
rs755973922 Chr.17: 28758979-28758979 on Build GRCh38
rs756276292 Chr.17: 28758938-28758938 on Build GRCh38
rs756972270 Chr.17: 28758908-28758908 on Build GRCh38
rs765004217 Chr.17: 28758900-28758900 on Build GRCh38
rs753324955 Chr.17: 28759002-28759004 on Build GRCh38
rs773576816 Chr.17: 28758954-28758954 on Build GRCh38
rs563800134 Chr.17: 28759077-28759077 on Build GRCh38
rs752194762 Chr.17: 28758905-28758905 on Build GRCh38
rs1028301133 Chr.17: 28759009-28759009 on Build GRCh38
rs761764271 Chr.17: 28758965-28758965 on Build GRCh38
rs780262327 Chr.17: 28758941-28758941 on Build GRCh38
rs748911334 Chr.17: 28759026-28759026 on Build GRCh38
rs767373332 Chr.17: 28758966-28758966 on Build GRCh38
rs555899896 Chr.17: 28759035-28759035 on Build GRCh38
rs773077335 Chr.17: 28759042-28759042 on Build GRCh38
rs753661690 Chr.17: 28759060-28759060 on Build GRCh38
rs761237014 Chr.17: 28759033-28759033 on Build GRCh38
rs372142554 Chr.17: 28758884-28758884 on Build GRCh38
rs771472912 Chr.17: 28759018-28759018 on Build GRCh38
rs369987412 Chr.17: 28758956-28758956 on Build GRCh38
rs758880501 Chr.17: 28759011-28759011 on Build GRCh38
rs772532734 Chr.17: 28758885-28758885 on Build GRCh38

View Full Product Details