Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 99708787-99709048 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRRS1
Entrez Gene ID
391059
Gene Name
ferric chelate reductase 1
Gene Aliases
SDFR2, SDR2
UniGene
Hs.454779
Gene Chromosome Location
Chr.1: 99708699-99766630 on Build GRCh38
Transcripts
NM_001013660.2 XM_005270861.3 XM_011541451.2 XM_017001271.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7430352 c.1825G>T p.D609Y Chr.1: 99708954-99708954 on Build GRCh38
423310 c.1853C>T p.S618L Chr.1: 99708926-99708926 on Build GRCh38
4935661 c.1781A>T p.Q594L Chr.1: 99708998-99708998 on Build GRCh38
332331 c.1881G>T p.*627Y Chr.1: 99708898-99708898 on Build GRCh38
892880 c.1791T>G p.I597M Chr.1: 99708988-99708988 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765740240 Chr.1: 99709001-99709001 on Build GRCh38
rs192583899 Chr.1: 99708899-99708899 on Build GRCh38
rs772725916 Chr.1: 99708887-99708887 on Build GRCh38
rs568801878 Chr.1: 99708950-99708950 on Build GRCh38
rs759959918 Chr.1: 99708986-99708986 on Build GRCh38
rs774424630 Chr.1: 99708949-99708949 on Build GRCh38
rs754438918 Chr.1: 99708985-99708985 on Build GRCh38
rs752449134 Chr.1: 99708868-99708868 on Build GRCh38
rs911585827 Chr.1: 99708969-99708969 on Build GRCh38
rs771979007 Chr.1: 99708952-99708952 on Build GRCh38
rs1051339353 Chr.1: 99708820-99708820 on Build GRCh38
rs146752240 Chr.1: 99708905-99708905 on Build GRCh38
rs761217556 Chr.1: 99708968-99708968 on Build GRCh38
rs1022306144 Chr.1: 99708955-99708955 on Build GRCh38
rs768250766 Chr.1: 99708874-99708874 on Build GRCh38
rs756799060 Chr.1: 99709023-99709023 on Build GRCh38
rs751112100 Chr.1: 99709017-99709017 on Build GRCh38
rs777097398 Chr.1: 99709003-99709003 on Build GRCh38
rs144949239 Chr.1: 99708962-99708962 on Build GRCh38
rs745630104 Chr.1: 99708926-99708926 on Build GRCh38
rs767028812 Chr.1: 99708977-99708977 on Build GRCh38
rs769624373 Chr.1: 99708945-99708945 on Build GRCh38
rs752221458 Chr.1: 99709000-99709000 on Build GRCh38
rs773619325 Chr.1: 99709022-99709022 on Build GRCh38
rs553200347 Chr.1: 99708998-99708998 on Build GRCh38
rs1008932456 Chr.1: 99708857-99708857 on Build GRCh38
rs935076079 Chr.1: 99708987-99708987 on Build GRCh38
rs12041864 Chr.1: 99708890-99708890 on Build GRCh38
rs878894759 Chr.1: 99708999-99708999 on Build GRCh38
rs900607627 Chr.1: 99708889-99708889 on Build GRCh38
rs757840417 Chr.1: 99708885-99708885 on Build GRCh38

View Full Product Details