Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 108156849-108157116 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL4A6
Entrez Gene ID
1288
Gene Name
collagen type IV alpha 6 chain
Gene Aliases
CXDELq22.3, DELXq22.3, DFNX6
UniGene
Hs.145586
Gene Chromosome Location
Chr.X: 108155607-108439497 on Build GRCh38
Transcripts
NM_001287758.1 NM_001287759.1 NM_001287760.1 NM_001847.3 NM_033641.3 XM_006724617.3 XM_011530852.2 XM_011530853.2

SNP Detail

SNP ID Chromosome Location
rs948457790 Chr.X: 108157078-108157078 on Build GRCh38
rs765597147 Chr.X: 108156977-108156977 on Build GRCh38
rs985953428 Chr.X: 108156884-108156884 on Build GRCh38
rs766540554 Chr.X: 108156961-108156961 on Build GRCh38
rs775020715 Chr.X: 108156997-108156997 on Build GRCh38
rs924514095 Chr.X: 108156929-108156929 on Build GRCh38
rs777670741 Chr.X: 108156969-108156969 on Build GRCh38
rs754744392 Chr.X: 108157021-108157021 on Build GRCh38
rs774969830 Chr.X: 108157006-108157006 on Build GRCh38
rs749659697 Chr.X: 108157001-108157001 on Build GRCh38
rs775038923 Chr.X: 108156873-108156873 on Build GRCh38
rs760675474 Chr.X: 108157040-108157040 on Build GRCh38
rs3765292 Chr.X: 108156906-108156906 on Build GRCh38
rs769425557 Chr.X: 108156995-108156995 on Build GRCh38
rs868419322 Chr.X: 108157083-108157083 on Build GRCh38
rs752469273 Chr.X: 108156968-108156968 on Build GRCh38
rs769211787 Chr.X: 108157002-108157002 on Build GRCh38
rs573679137 Chr.X: 108157092-108157092 on Build GRCh38
rs763913956 Chr.X: 108157064-108157064 on Build GRCh38
rs934084168 Chr.X: 108156983-108156983 on Build GRCh38
rs773301536 Chr.X: 108157037-108157037 on Build GRCh38
rs751471734 Chr.X: 108157079-108157079 on Build GRCh38
rs992499363 Chr.X: 108157004-108157004 on Build GRCh38
rs767249186 Chr.X: 108157080-108157080 on Build GRCh38
rs59121927 Chr.X: 108156930-108156930 on Build GRCh38
rs758181213 Chr.X: 108157063-108157063 on Build GRCh38
rs766585041 Chr.X: 108156911-108156911 on Build GRCh38
rs749619355 Chr.X: 108157029-108157029 on Build GRCh38
rs370369075 Chr.X: 108156987-108156987 on Build GRCh38
rs747013143 Chr.X: 108156972-108156972 on Build GRCh38
rs954122672 Chr.X: 108156880-108156880 on Build GRCh38
rs756310211 Chr.X: 108156960-108156960 on Build GRCh38
rs780799556 Chr.X: 108156991-108156991 on Build GRCh38
rs768884286 Chr.X: 108157055-108157055 on Build GRCh38
rs186343614 Chr.X: 108156889-108156889 on Build GRCh38
rs753313861 Chr.X: 108156990-108156991 on Build GRCh38
rs199981380 Chr.X: 108156993-108156993 on Build GRCh38
rs773285255 Chr.X: 108156897-108156897 on Build GRCh38

View Full Product Details