Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 50475338-50475563 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM71E1
Entrez Gene ID
112703
Gene Name
family with sequence similarity 71 member E1
Gene Aliases
-
UniGene
Hs.448941
Gene Chromosome Location
Chr.19: 50466643-50476746 on Build GRCh38
Transcripts
NM_001308429.1 NM_138411.2 XM_011526405.2 XM_011526406.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6151888 c.508G>A p.A170T Chr.19: 50475356-50475356 on Build GRCh38
4863130 c.481C>T p.P161S Chr.19: 50475383-50475383 on Build GRCh38
6999181 c.367C>T p.R123W Chr.19: 50475449-50475449 on Build GRCh38
713297 c.433C>T p.P145S Chr.19: 50475383-50475383 on Build GRCh38
3737228 c.464_465insTCCA p.A157fs*>93 Chr.19: 50475399-50475400 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs191020926 Chr.19: 50475508-50475508 on Build GRCh38
rs778301108 Chr.19: 50475516-50475517 on Build GRCh38
rs537093062 Chr.19: 50475398-50475398 on Build GRCh38
rs775252763 Chr.19: 50475362-50475362 on Build GRCh38
rs749603781 Chr.19: 50475490-50475490 on Build GRCh38
rs763452397 Chr.19: 50475382-50475382 on Build GRCh38
rs746692083 Chr.19: 50475479-50475479 on Build GRCh38
rs752839799 Chr.19: 50475361-50475361 on Build GRCh38
rs140490534 Chr.19: 50475429-50475429 on Build GRCh38
rs759658220 Chr.19: 50475404-50475404 on Build GRCh38
rs143306289 Chr.19: 50475480-50475480 on Build GRCh38
rs754093303 Chr.19: 50475516-50475516 on Build GRCh38
rs753537896 Chr.19: 50475448-50475448 on Build GRCh38
rs746641890 Chr.19: 50475363-50475363 on Build GRCh38
rs773652743 Chr.19: 50475489-50475489 on Build GRCh38
rs763961551 Chr.19: 50475444-50475444 on Build GRCh38
rs765122835 Chr.19: 50475525-50475525 on Build GRCh38
rs185418641 Chr.19: 50475467-50475467 on Build GRCh38
rs757282302 Chr.19: 50475459-50475459 on Build GRCh38
rs1049286345 Chr.19: 50475538-50475538 on Build GRCh38
rs764173618 Chr.19: 50475515-50475515 on Build GRCh38
rs757159568 Chr.19: 50475521-50475521 on Build GRCh38
rs940243941 Chr.19: 50475389-50475389 on Build GRCh38
rs753029642 Chr.19: 50475428-50475428 on Build GRCh38
rs867867680 Chr.19: 50475504-50475504 on Build GRCh38
rs773394430 Chr.19: 50475366-50475366 on Build GRCh38
rs1623859 Chr.19: 50475532-50475532 on Build GRCh38
rs201537879 Chr.19: 50475477-50475477 on Build GRCh38
rs973306279 Chr.19: 50475392-50475392 on Build GRCh38
rs758440891 Chr.19: 50475485-50475488 on Build GRCh38
rs200427417 Chr.19: 50475478-50475478 on Build GRCh38
rs549345876 Chr.19: 50475426-50475426 on Build GRCh38
rs939730170 Chr.19: 50475492-50475492 on Build GRCh38
rs199597534 Chr.19: 50475494-50475494 on Build GRCh38
rs895120268 Chr.19: 50475449-50475449 on Build GRCh38
rs866131132 Chr.19: 50475484-50475484 on Build GRCh38
rs539244786 Chr.19: 50475491-50475491 on Build GRCh38
rs987080557 Chr.19: 50475380-50475380 on Build GRCh38
rs372043427 Chr.19: 50475505-50475505 on Build GRCh38
rs368186220 Chr.19: 50475495-50475495 on Build GRCh38

View Full Product Details