Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 23565923-23566154 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CST9L
Entrez Gene ID
128821
Gene Name
cystatin 9-like
Gene Aliases
CTES7B, bA218C14.1
UniGene
Hs.121554
Gene Chromosome Location
Chr.20: 23564732-23568749 on Build GRCh38
Transcripts
NM_080610.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4943054 c.258A>G p.V86V Chr.20: 23566070-23566070 on Build GRCh38
1249352 c.255T>C p.T85T Chr.20: 23566073-23566073 on Build GRCh38
6200294 c.354+4A>C p.? Chr.20: 23565970-23565970 on Build GRCh38
4908423 c.269A>C p.E90A Chr.20: 23566059-23566059 on Build GRCh38
6637007 c.354+2T>C p.? Chr.20: 23565972-23565972 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs545793477 Chr.20: 23566018-23566018 on Build GRCh38
rs777612116 Chr.20: 23566005-23566005 on Build GRCh38
rs762096631 Chr.20: 23566041-23566041 on Build GRCh38
rs750815990 Chr.20: 23565962-23565962 on Build GRCh38
rs754292265 Chr.20: 23566061-23566061 on Build GRCh38
rs139795806 Chr.20: 23566093-23566093 on Build GRCh38
rs987413506 Chr.20: 23566062-23566062 on Build GRCh38
rs777047849 Chr.20: 23566122-23566122 on Build GRCh38
rs568122636 Chr.20: 23566063-23566063 on Build GRCh38
rs758624895 Chr.20: 23566076-23566076 on Build GRCh38
rs778983947 Chr.20: 23565995-23565995 on Build GRCh38
rs146538492 Chr.20: 23566004-23566004 on Build GRCh38
rs762660726 Chr.20: 23566023-23566023 on Build GRCh38
rs756626927 Chr.20: 23565985-23565985 on Build GRCh38
rs771226340 Chr.20: 23566119-23566119 on Build GRCh38
rs777898227 Chr.20: 23566077-23566077 on Build GRCh38
rs768246735 Chr.20: 23566037-23566037 on Build GRCh38
rs762529073 Chr.20: 23565948-23565948 on Build GRCh38
rs765355598 Chr.20: 23566070-23566070 on Build GRCh38
rs761066730 Chr.20: 23566045-23566045 on Build GRCh38
rs552619927 Chr.20: 23565988-23565988 on Build GRCh38
rs55722105 Chr.20: 23566085-23566085 on Build GRCh38
rs751842420 Chr.20: 23566082-23566082 on Build GRCh38
rs866980383 Chr.20: 23566048-23566048 on Build GRCh38
rs766760726 Chr.20: 23566053-23566053 on Build GRCh38
rs140293366 Chr.20: 23566089-23566089 on Build GRCh38
rs755009202 Chr.20: 23565992-23565992 on Build GRCh38
rs112407759 Chr.20: 23565989-23565989 on Build GRCh38
rs367979416 Chr.20: 23565944-23565944 on Build GRCh38
rs73904935 Chr.20: 23566019-23566019 on Build GRCh38
rs368473297 Chr.20: 23566011-23566011 on Build GRCh38
rs371780080 Chr.20: 23565990-23565990 on Build GRCh38
rs200712432 Chr.20: 23566088-23566088 on Build GRCh38
rs2295564 Chr.20: 23566002-23566002 on Build GRCh38
rs189798431 Chr.20: 23566109-23566109 on Build GRCh38
rs768323635 Chr.20: 23566102-23566102 on Build GRCh38
rs746828326 Chr.20: 23566010-23566010 on Build GRCh38
rs761475021 Chr.20: 23566111-23566111 on Build GRCh38
rs79890466 Chr.20: 23566072-23566072 on Build GRCh38
rs375095063 Chr.20: 23565956-23565956 on Build GRCh38
rs773601205 Chr.20: 23566042-23566042 on Build GRCh38
rs760025948 Chr.20: 23566128-23566128 on Build GRCh38
rs372129662 Chr.20: 23566084-23566084 on Build GRCh38
rs947627604 Chr.20: 23565979-23565979 on Build GRCh38

View Full Product Details